Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26794239-26794686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPA
Entrez Gene ID
1058
Gene Name
centromere protein A
Gene Aliases
CENP-A, CenH3
UniGene
Hs.1594
Gene Chromosome Location
Chr.2: 26786014-26794589 on Build GRCh38
Transcripts
NM_001042426.1 NM_001809.3

SNP Detail

SNP ID Chromosome Location
rs768550873 Chr.2: 26794523-26794523 on Build GRCh38
rs1049408123 Chr.2: 26794619-26794619 on Build GRCh38
rs747936052 Chr.2: 26794590-26794590 on Build GRCh38
rs961377642 Chr.2: 26794493-26794493 on Build GRCh38
rs1014679762 Chr.2: 26794654-26794657 on Build GRCh38
rs910370235 Chr.2: 26794658-26794658 on Build GRCh38
rs1009210047 Chr.2: 26794297-26794297 on Build GRCh38
rs535303655 Chr.2: 26794333-26794333 on Build GRCh38
rs1038298101 Chr.2: 26794565-26794565 on Build GRCh38
rs963954704 Chr.2: 26794359-26794359 on Build GRCh38
rs992404605 Chr.2: 26794428-26794428 on Build GRCh38
rs920019176 Chr.2: 26794501-26794501 on Build GRCh38
rs1007475621 Chr.2: 26794274-26794274 on Build GRCh38
rs937546834 Chr.2: 26794589-26794589 on Build GRCh38
rs940865255 Chr.2: 26794515-26794515 on Build GRCh38
rs931390002 Chr.2: 26794520-26794520 on Build GRCh38
rs959928815 Chr.2: 26794357-26794357 on Build GRCh38
rs545735548 Chr.2: 26794517-26794517 on Build GRCh38
rs1040470920 Chr.2: 26794276-26794276 on Build GRCh38
rs1033842533 Chr.2: 26794354-26794354 on Build GRCh38
rs972670396 Chr.2: 26794494-26794494 on Build GRCh38
rs895780722 Chr.2: 26794625-26794625 on Build GRCh38
rs746775475 Chr.2: 26794441-26794441 on Build GRCh38
rs920832619 Chr.2: 26794569-26794569 on Build GRCh38
rs541310134 Chr.2: 26794445-26794445 on Build GRCh38
rs1056076161 Chr.2: 26794600-26794600 on Build GRCh38
rs753592561 Chr.2: 26794334-26794334 on Build GRCh38
rs557088740 Chr.2: 26794335-26794335 on Build GRCh38
rs1041797797 Chr.2: 26794661-26794661 on Build GRCh38
rs575460553 Chr.2: 26794398-26794398 on Build GRCh38
rs908321026 Chr.2: 26794471-26794471 on Build GRCh38
rs376469103 Chr.2: 26794535-26794535 on Build GRCh38
rs901497554 Chr.2: 26794289-26794289 on Build GRCh38
rs975038469 Chr.2: 26794392-26794392 on Build GRCh38
rs766114039 Chr.2: 26794332-26794332 on Build GRCh38
rs889066533 Chr.2: 26794269-26794269 on Build GRCh38

View Full Product Details