Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27578810-27579308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf16
Entrez Gene ID
84226
Gene Name
chromosome 2 open reading frame 16
Gene Aliases
-
UniGene
Hs.131021
Gene Chromosome Location
Chr.2: 27576522-27582722 on Build GRCh38
Transcripts
NM_032266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3580703 c.2715G>A p.R905R Chr.2: 27579287-27579287 on Build GRCh38
442759 c.2574G>C p.G858G Chr.2: 27579146-27579146 on Build GRCh38
6615824 c.2290G>A p.A764T Chr.2: 27578862-27578862 on Build GRCh38
2940380 c.2315C>T p.T772I Chr.2: 27578887-27578887 on Build GRCh38
3839320 c.2403C>T p.S801S Chr.2: 27578975-27578975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924995246 Chr.2: 27579235-27579235 on Build GRCh38
rs751248828 Chr.2: 27579078-27579078 on Build GRCh38
rs201575164 Chr.2: 27579038-27579038 on Build GRCh38
rs768623287 Chr.2: 27579048-27579048 on Build GRCh38
rs759218200 Chr.2: 27578909-27578909 on Build GRCh38
rs775372150 Chr.2: 27578963-27578963 on Build GRCh38
rs372540355 Chr.2: 27579075-27579075 on Build GRCh38
rs769117383 Chr.2: 27579069-27579069 on Build GRCh38
rs754388407 Chr.2: 27579024-27579024 on Build GRCh38
rs746769493 Chr.2: 27579046-27579046 on Build GRCh38
rs781527575 Chr.2: 27578868-27578868 on Build GRCh38
rs201642055 Chr.2: 27578934-27578934 on Build GRCh38
rs755621889 Chr.2: 27578851-27578851 on Build GRCh38
rs750140390 Chr.2: 27578930-27578930 on Build GRCh38
rs865923015 Chr.2: 27579049-27579050 on Build GRCh38
rs764386714 Chr.2: 27579206-27579206 on Build GRCh38
rs759194820 Chr.2: 27579190-27579190 on Build GRCh38
rs769859103 Chr.2: 27578873-27578873 on Build GRCh38
rs267599325 Chr.2: 27579271-27579271 on Build GRCh38
rs373409429 Chr.2: 27579118-27579118 on Build GRCh38
rs762463627 Chr.2: 27579074-27579074 on Build GRCh38
rs1042007263 Chr.2: 27579180-27579180 on Build GRCh38
rs375613150 Chr.2: 27579098-27579098 on Build GRCh38
rs780302659 Chr.2: 27579124-27579124 on Build GRCh38
rs773284401 Chr.2: 27578881-27578881 on Build GRCh38
rs749397314 Chr.2: 27578953-27578953 on Build GRCh38
rs923498019 Chr.2: 27578895-27578895 on Build GRCh38
rs754279989 Chr.2: 27579102-27579102 on Build GRCh38
rs1919128 Chr.2: 27578892-27578892 on Build GRCh38
rs777204776 Chr.2: 27579096-27579098 on Build GRCh38
rs755283480 Chr.2: 27578932-27578932 on Build GRCh38
rs751389465 Chr.2: 27579094-27579094 on Build GRCh38
rs780518234 Chr.2: 27578849-27578849 on Build GRCh38
rs777006672 Chr.2: 27579210-27579210 on Build GRCh38
rs1012669839 Chr.2: 27579269-27579269 on Build GRCh38
rs771160816 Chr.2: 27578900-27578900 on Build GRCh38
rs905224548 Chr.2: 27579055-27579055 on Build GRCh38
rs776751704 Chr.2: 27578975-27578975 on Build GRCh38
rs917781573 Chr.2: 27579281-27579281 on Build GRCh38
rs751763614 Chr.2: 27579227-27579227 on Build GRCh38
rs199647806 Chr.2: 27579010-27579010 on Build GRCh38
rs142432422 Chr.2: 27578955-27578955 on Build GRCh38
rs367711029 Chr.2: 27578991-27578991 on Build GRCh38
rs746171023 Chr.2: 27578958-27578958 on Build GRCh38
rs1039900796 Chr.2: 27578862-27578862 on Build GRCh38
rs370300682 Chr.2: 27578992-27578992 on Build GRCh38
rs774154153 Chr.2: 27578907-27578907 on Build GRCh38
rs902861334 Chr.2: 27578915-27578915 on Build GRCh38
rs764858351 Chr.2: 27578984-27578984 on Build GRCh38
rs747367524 Chr.2: 27578851-27578851 on Build GRCh38
rs770467132 Chr.2: 27579284-27579284 on Build GRCh38
rs894126331 Chr.2: 27579250-27579250 on Build GRCh38
rs749238111 Chr.2: 27579279-27579279 on Build GRCh38
rs768745795 Chr.2: 27578971-27578971 on Build GRCh38
rs565322712 Chr.2: 27578916-27578916 on Build GRCh38
rs999836039 Chr.2: 27578941-27578941 on Build GRCh38
rs778219346 Chr.2: 27579013-27579013 on Build GRCh38
rs962211042 Chr.2: 27578879-27578879 on Build GRCh38
rs755263543 Chr.2: 27579243-27579243 on Build GRCh38
rs781084498 Chr.2: 27579244-27579244 on Build GRCh38
rs764051795 Chr.2: 27579091-27579091 on Build GRCh38
rs780961674 Chr.2: 27579185-27579185 on Build GRCh38
rs868020171 Chr.2: 27578893-27578893 on Build GRCh38
rs758135011 Chr.2: 27579086-27579086 on Build GRCh38
rs764497121 Chr.2: 27579100-27579100 on Build GRCh38
rs777777006 Chr.2: 27578951-27578951 on Build GRCh38
rs150880708 Chr.2: 27579037-27579037 on Build GRCh38
rs372328700 Chr.2: 27579179-27579179 on Build GRCh38
rs756626057 Chr.2: 27578943-27578943 on Build GRCh38
rs752625085 Chr.2: 27579018-27579018 on Build GRCh38
rs778814464 Chr.2: 27579117-27579117 on Build GRCh38
rs372494393 Chr.2: 27579246-27579246 on Build GRCh38
rs747294451 Chr.2: 27579127-27579127 on Build GRCh38
rs763644341 Chr.2: 27578927-27578927 on Build GRCh38
rs762191563 Chr.2: 27579213-27579213 on Build GRCh38
rs534860684 Chr.2: 27579153-27579153 on Build GRCh38
rs866537105 Chr.2: 27578869-27578869 on Build GRCh38
rs767714904 Chr.2: 27578995-27578995 on Build GRCh38
rs546819428 Chr.2: 27579157-27579157 on Build GRCh38
rs552477182 Chr.2: 27579071-27579071 on Build GRCh38
rs577064419 Chr.2: 27578938-27578938 on Build GRCh38
rs766635320 Chr.2: 27579226-27579226 on Build GRCh38
rs753508803 Chr.2: 27579143-27579144 on Build GRCh38
rs753123779 Chr.2: 27579000-27579000 on Build GRCh38
rs771036313 Chr.2: 27578954-27578954 on Build GRCh38
rs375417773 Chr.2: 27579034-27579034 on Build GRCh38
rs373280648 Chr.2: 27578841-27578841 on Build GRCh38
rs757778138 Chr.2: 27579110-27579110 on Build GRCh38
rs369073751 Chr.2: 27579220-27579220 on Build GRCh38
rs753422352 Chr.2: 27578928-27578928 on Build GRCh38
rs779056495 Chr.2: 27579030-27579030 on Build GRCh38
rs764953409 Chr.2: 27578929-27578929 on Build GRCh38
rs267599326 Chr.2: 27579282-27579282 on Build GRCh38
rs765744218 Chr.2: 27579216-27579216 on Build GRCh38
rs772554173 Chr.2: 27579175-27579175 on Build GRCh38
rs200959503 Chr.2: 27578965-27578965 on Build GRCh38
rs984160369 Chr.2: 27579067-27579067 on Build GRCh38
rs373095066 Chr.2: 27578917-27578917 on Build GRCh38
rs1054158213 Chr.2: 27579247-27579247 on Build GRCh38
rs757020776 Chr.2: 27579143-27579143 on Build GRCh38
rs200321448 Chr.2: 27578976-27578976 on Build GRCh38
rs566136500 Chr.2: 27579176-27579176 on Build GRCh38
rs139381292 Chr.2: 27579278-27579278 on Build GRCh38
rs761908059 Chr.2: 27579060-27579060 on Build GRCh38
rs769792298 Chr.2: 27579164-27579164 on Build GRCh38
rs756984328 Chr.2: 27578840-27578840 on Build GRCh38
rs755411579 Chr.2: 27578860-27578860 on Build GRCh38
rs1006754469 Chr.2: 27578847-27578847 on Build GRCh38
rs13416968 Chr.2: 27578872-27578872 on Build GRCh38
rs530692673 Chr.2: 27579052-27579052 on Build GRCh38

View Full Product Details