Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27890441-27890934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRE
Entrez Gene ID
9577
Gene Name
brain and reproductive organ-expressed (TNFRSF1A modulator)
Gene Aliases
BRCC4, BRCC45
UniGene
Hs.258314
Gene Chromosome Location
Chr.2: 27890615-28338901 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2 NR_028308.1

Gene Symbol
BRE-AS1
Entrez Gene ID
100302650
Gene Name
BRE antisense RNA 1
Gene Aliases
-
UniGene
Hs.729719
Gene Chromosome Location
Chr.2: 27889456-27891114 on Build GRCh38
Transcripts
NM_001261840.1 NM_004899.4 NM_199191.2 NM_199192.2 NM_199193.2 NM_199194.2 NR_028308.1

SNP Detail

SNP ID Chromosome Location
rs771306905 Chr.2: 27890767-27890767 on Build GRCh38
rs1018062248 Chr.2: 27890821-27890821 on Build GRCh38
rs944746447 Chr.2: 27890759-27890759 on Build GRCh38
rs1026371175 Chr.2: 27890806-27890806 on Build GRCh38
rs747173924 Chr.2: 27890495-27890495 on Build GRCh38
rs896953662 Chr.2: 27890789-27890789 on Build GRCh38
rs1055450071 Chr.2: 27890696-27890696 on Build GRCh38
rs565517635 Chr.2: 27890663-27890663 on Build GRCh38
rs900663799 Chr.2: 27890774-27890774 on Build GRCh38
rs1006203640 Chr.2: 27890812-27890812 on Build GRCh38
rs1028026113 Chr.2: 27890620-27890620 on Build GRCh38
rs920641345 Chr.2: 27890700-27890700 on Build GRCh38
rs952257754 Chr.2: 27890809-27890809 on Build GRCh38
rs1055219374 Chr.2: 27890726-27890726 on Build GRCh38
rs867816969 Chr.2: 27890881-27890881 on Build GRCh38
rs781359981 Chr.2: 27890506-27890507 on Build GRCh38
rs895058391 Chr.2: 27890756-27890756 on Build GRCh38
rs536218565 Chr.2: 27890764-27890764 on Build GRCh38
rs796407895 Chr.2: 27890885-27890885 on Build GRCh38
rs927904283 Chr.2: 27890553-27890553 on Build GRCh38
rs985210953 Chr.2: 27890496-27890496 on Build GRCh38
rs114146461 Chr.2: 27890466-27890466 on Build GRCh38
rs996473925 Chr.2: 27890785-27890785 on Build GRCh38
rs1042096542 Chr.2: 27890606-27890606 on Build GRCh38
rs538064099 Chr.2: 27890567-27890567 on Build GRCh38
rs538347533 Chr.2: 27890628-27890628 on Build GRCh38
rs574663262 Chr.2: 27890604-27890604 on Build GRCh38
rs976213732 Chr.2: 27890536-27890536 on Build GRCh38
rs549921090 Chr.2: 27890592-27890592 on Build GRCh38
rs996811507 Chr.2: 27890597-27890597 on Build GRCh38
rs571058235 Chr.2: 27890593-27890593 on Build GRCh38
rs1047836648 Chr.2: 27890578-27890578 on Build GRCh38
rs886490538 Chr.2: 27890581-27890581 on Build GRCh38
rs936335759 Chr.2: 27890710-27890710 on Build GRCh38
rs771142173 Chr.2: 27890502-27890502 on Build GRCh38
rs749514790 Chr.2: 27890667-27890667 on Build GRCh38
rs994972531 Chr.2: 27890571-27890571 on Build GRCh38
rs930640934 Chr.2: 27890706-27890707 on Build GRCh38
rs994022178 Chr.2: 27890791-27890791 on Build GRCh38
rs989070394 Chr.2: 27890650-27890650 on Build GRCh38
rs764142329 Chr.2: 27890853-27890853 on Build GRCh38
rs553765865 Chr.2: 27890652-27890652 on Build GRCh38
rs1046174635 Chr.2: 27890538-27890538 on Build GRCh38
rs964749228 Chr.2: 27890896-27890896 on Build GRCh38
rs927774172 Chr.2: 27890658-27890658 on Build GRCh38
rs903612266 Chr.2: 27890662-27890662 on Build GRCh38
rs1010546443 Chr.2: 27890825-27890825 on Build GRCh38
rs997077066 Chr.2: 27890554-27890567 on Build GRCh38
rs776816973 Chr.2: 27890570-27890570 on Build GRCh38
rs1013545667 Chr.2: 27890766-27890766 on Build GRCh38
rs910793515 Chr.2: 27890497-27890497 on Build GRCh38
rs893347372 Chr.2: 27890814-27890814 on Build GRCh38

View Full Product Details