Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30641241-30641779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
UniGene
Hs.660911
Gene Chromosome Location
Chr.2: 30610382-30809527 on Build GRCh38
Transcripts
XM_017005265.1 NM_001002257.2 NM_001304445.1 NM_001304446.1 NM_182551.4 XM_005264244.1 XM_005264245.3 XM_011532741.2 XM_011532742.2 XM_011532743.2 XM_011532744.2 XM_017003746.1

Gene Symbol
LCLAT1
Entrez Gene ID
253558
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
UniGene
Hs.468048
Gene Chromosome Location
Chr.2: 30447226-30644225 on Build GRCh38
Transcripts
XM_017005265.1 NM_001002257.2 NM_001304445.1 NM_001304446.1 NM_182551.4 XM_005264244.1 XM_005264245.3 XM_011532741.2 XM_011532742.2 XM_011532743.2 XM_011532744.2 XM_017003746.1

SNP Detail

SNP ID Chromosome Location
rs369614361 Chr.2: 30641512-30641512 on Build GRCh38
rs538263035 Chr.2: 30641739-30641739 on Build GRCh38
rs747620740 Chr.2: 30641584-30641584 on Build GRCh38
rs1010389689 Chr.2: 30641533-30641533 on Build GRCh38
rs1055010172 Chr.2: 30641492-30641492 on Build GRCh38
rs979797725 Chr.2: 30641754-30641754 on Build GRCh38
rs973608942 Chr.2: 30641365-30641366 on Build GRCh38
rs900552612 Chr.2: 30641477-30641477 on Build GRCh38
rs989041038 Chr.2: 30641269-30641270 on Build GRCh38
rs917527371 Chr.2: 30641632-30641632 on Build GRCh38
rs773382986 Chr.2: 30641674-30641674 on Build GRCh38
rs1047658164 Chr.2: 30641376-30641376 on Build GRCh38
rs80257598 Chr.2: 30641369-30641369 on Build GRCh38
rs535530870 Chr.2: 30641517-30641517 on Build GRCh38
rs17395619 Chr.2: 30641447-30641447 on Build GRCh38
rs948395836 Chr.2: 30641357-30641357 on Build GRCh38
rs1007540986 Chr.2: 30641481-30641481 on Build GRCh38
rs1057347920 Chr.2: 30641286-30641286 on Build GRCh38
rs925392872 Chr.2: 30641642-30641642 on Build GRCh38
rs891586336 Chr.2: 30641450-30641450 on Build GRCh38
rs929181486 Chr.2: 30641371-30641371 on Build GRCh38
rs2609945 Chr.2: 30641460-30641460 on Build GRCh38
rs992981469 Chr.2: 30641617-30641617 on Build GRCh38
rs200452584 Chr.2: 30641688-30641690 on Build GRCh38
rs569121973 Chr.2: 30641697-30641697 on Build GRCh38
rs925574204 Chr.2: 30641755-30641755 on Build GRCh38
rs537779945 Chr.2: 30641402-30641402 on Build GRCh38
rs1024504770 Chr.2: 30641550-30641550 on Build GRCh38
rs1033358258 Chr.2: 30641695-30641695 on Build GRCh38
rs926362033 Chr.2: 30641352-30641352 on Build GRCh38
rs911429962 Chr.2: 30641399-30641399 on Build GRCh38
rs186627062 Chr.2: 30641295-30641295 on Build GRCh38
rs981776680 Chr.2: 30641321-30641324 on Build GRCh38
rs935754478 Chr.2: 30641404-30641404 on Build GRCh38
rs139847393 Chr.2: 30641713-30641716 on Build GRCh38
rs948946907 Chr.2: 30641720-30641720 on Build GRCh38
rs192561099 Chr.2: 30641542-30641542 on Build GRCh38
rs756441068 Chr.2: 30641279-30641279 on Build GRCh38
rs971660261 Chr.2: 30641666-30641666 on Build GRCh38
rs561487161 Chr.2: 30641278-30641278 on Build GRCh38
rs1017558204 Chr.2: 30641497-30641497 on Build GRCh38
rs1039058251 Chr.2: 30641472-30641472 on Build GRCh38
rs558211278 Chr.2: 30641753-30641753 on Build GRCh38
rs541912935 Chr.2: 30641272-30641272 on Build GRCh38
rs550434062 Chr.2: 30641319-30641319 on Build GRCh38
rs1029507176 Chr.2: 30641465-30641465 on Build GRCh38
rs889688758 Chr.2: 30641480-30641480 on Build GRCh38
rs780265561 Chr.2: 30641391-30641391 on Build GRCh38

View Full Product Details