Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30723058-30723555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
UniGene
Hs.660911
Gene Chromosome Location
Chr.2: 30610382-30809527 on Build GRCh38
Transcripts
NM_144575.2 XM_017005264.1 XM_017005265.1

SNP Detail

SNP ID Chromosome Location
rs975894690 Chr.2: 30723499-30723499 on Build GRCh38
rs977396482 Chr.2: 30723230-30723230 on Build GRCh38
rs921323716 Chr.2: 30723521-30723521 on Build GRCh38
rs887327450 Chr.2: 30723334-30723334 on Build GRCh38
rs565971377 Chr.2: 30723174-30723174 on Build GRCh38
rs1033554433 Chr.2: 30723429-30723429 on Build GRCh38
rs1010702700 Chr.2: 30723448-30723448 on Build GRCh38
rs773399108 Chr.2: 30723409-30723409 on Build GRCh38
rs553487079 Chr.2: 30723276-30723276 on Build GRCh38
rs972230997 Chr.2: 30723131-30723132 on Build GRCh38
rs925054012 Chr.2: 30723186-30723186 on Build GRCh38
rs966589795 Chr.2: 30723456-30723456 on Build GRCh38
rs975631852 Chr.2: 30723281-30723281 on Build GRCh38
rs1879278 Chr.2: 30723482-30723482 on Build GRCh38
rs372495289 Chr.2: 30723318-30723318 on Build GRCh38
rs1049127760 Chr.2: 30723323-30723323 on Build GRCh38
rs186245207 Chr.2: 30723128-30723128 on Build GRCh38
rs967375931 Chr.2: 30723217-30723217 on Build GRCh38
rs535846327 Chr.2: 30723353-30723353 on Build GRCh38
rs534800945 Chr.2: 30723465-30723465 on Build GRCh38
rs912666214 Chr.2: 30723250-30723250 on Build GRCh38
rs998886743 Chr.2: 30723467-30723467 on Build GRCh38
rs931616416 Chr.2: 30723304-30723304 on Build GRCh38
rs955167155 Chr.2: 30723141-30723141 on Build GRCh38
rs1879279 Chr.2: 30723360-30723360 on Build GRCh38
rs372965267 Chr.2: 30723101-30723101 on Build GRCh38
rs1016282412 Chr.2: 30723394-30723394 on Build GRCh38
rs770909654 Chr.2: 30723327-30723327 on Build GRCh38
rs1021311069 Chr.2: 30723455-30723455 on Build GRCh38
rs893640961 Chr.2: 30723445-30723445 on Build GRCh38
rs944091722 Chr.2: 30723259-30723259 on Build GRCh38
rs921429598 Chr.2: 30723295-30723295 on Build GRCh38
rs1013881205 Chr.2: 30723255-30723255 on Build GRCh38
rs1002004240 Chr.2: 30723165-30723165 on Build GRCh38
rs988105646 Chr.2: 30723464-30723465 on Build GRCh38
rs1003799668 Chr.2: 30723114-30723114 on Build GRCh38
rs1026868023 Chr.2: 30723406-30723406 on Build GRCh38
rs558783512 Chr.2: 30723471-30723471 on Build GRCh38
rs936485842 Chr.2: 30723190-30723191 on Build GRCh38
rs987751666 Chr.2: 30723142-30723142 on Build GRCh38
rs1033914862 Chr.2: 30723170-30723170 on Build GRCh38
rs573420987 Chr.2: 30723277-30723277 on Build GRCh38
rs1020837527 Chr.2: 30723187-30723187 on Build GRCh38
rs72613845 Chr.2: 30723102-30723102 on Build GRCh38
rs143968883 Chr.2: 30723378-30723378 on Build GRCh38
rs779133747 Chr.2: 30723236-30723236 on Build GRCh38
rs865881370 Chr.2: 30723176-30723176 on Build GRCh38
rs147308148 Chr.2: 30723405-30723405 on Build GRCh38
rs552453604 Chr.2: 30723156-30723156 on Build GRCh38
rs748301421 Chr.2: 30723291-30723291 on Build GRCh38
rs72613846 Chr.2: 30723134-30723134 on Build GRCh38
rs541520620 Chr.2: 30723527-30723527 on Build GRCh38
rs533622953 Chr.2: 30723251-30723251 on Build GRCh38
rs939706270 Chr.2: 30723339-30723339 on Build GRCh38
rs886767116 Chr.2: 30723112-30723112 on Build GRCh38
rs912175480 Chr.2: 30723151-30723151 on Build GRCh38

View Full Product Details