Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 31266462-31266956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD3
Entrez Gene ID
30845
Gene Name
EH domain containing 3
Gene Aliases
PAST3
UniGene
Hs.368808
Gene Chromosome Location
Chr.2: 31234014-31269451 on Build GRCh38
Transcripts
NM_014600.2 XM_011532806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205076 c.1381A>G p.K461E Chr.2: 31266477-31266477 on Build GRCh38
4583467 c.1465G>A p.D489N Chr.2: 31266561-31266561 on Build GRCh38
1718724 c.1532A>C p.K511T Chr.2: 31266628-31266628 on Build GRCh38
167569 c.1498G>A p.E500K Chr.2: 31266594-31266594 on Build GRCh38
6324424 c.1440G>C p.V480V Chr.2: 31266536-31266536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879167307 Chr.2: 31266765-31266766 on Build GRCh38
rs367596478 Chr.2: 31266741-31266741 on Build GRCh38
rs762026491 Chr.2: 31266588-31266588 on Build GRCh38
rs962893406 Chr.2: 31266909-31266909 on Build GRCh38
rs145305384 Chr.2: 31266680-31266680 on Build GRCh38
rs984961496 Chr.2: 31266903-31266903 on Build GRCh38
rs766594592 Chr.2: 31266766-31266767 on Build GRCh38
rs201490817 Chr.2: 31266593-31266593 on Build GRCh38
rs771770948 Chr.2: 31266656-31266656 on Build GRCh38
rs757814503 Chr.2: 31266503-31266503 on Build GRCh38
rs774940677 Chr.2: 31266659-31266659 on Build GRCh38
rs530564469 Chr.2: 31266679-31266679 on Build GRCh38
rs770467922 Chr.2: 31266652-31266652 on Build GRCh38
rs144078061 Chr.2: 31266856-31266856 on Build GRCh38
rs192848173 Chr.2: 31266908-31266908 on Build GRCh38
rs535776415 Chr.2: 31266842-31266842 on Build GRCh38
rs770239969 Chr.2: 31266728-31266728 on Build GRCh38
rs767841259 Chr.2: 31266699-31266699 on Build GRCh38
rs779821173 Chr.2: 31266726-31266726 on Build GRCh38
rs1045200500 Chr.2: 31266918-31266918 on Build GRCh38
rs77999318 Chr.2: 31266709-31266709 on Build GRCh38
rs147406143 Chr.2: 31266529-31266529 on Build GRCh38
rs67643147 Chr.2: 31266762-31266763 on Build GRCh38
rs267599345 Chr.2: 31266688-31266688 on Build GRCh38
rs200504962 Chr.2: 31266514-31266514 on Build GRCh38
rs777410186 Chr.2: 31266716-31266716 on Build GRCh38
rs151181600 Chr.2: 31266653-31266653 on Build GRCh38
rs755269658 Chr.2: 31266633-31266633 on Build GRCh38
rs1030918366 Chr.2: 31266871-31266871 on Build GRCh38
rs770412826 Chr.2: 31266560-31266560 on Build GRCh38
rs150263934 Chr.2: 31266902-31266902 on Build GRCh38
rs923376075 Chr.2: 31266621-31266621 on Build GRCh38
rs924861066 Chr.2: 31266864-31266864 on Build GRCh38
rs372095794 Chr.2: 31266718-31266718 on Build GRCh38
rs370001880 Chr.2: 31266761-31266762 on Build GRCh38
rs878957323 Chr.2: 31266762-31266765 on Build GRCh38
rs35925176 Chr.2: 31266504-31266504 on Build GRCh38
rs869043033 Chr.2: 31266787-31266794 on Build GRCh38
rs774039230 Chr.2: 31266561-31266561 on Build GRCh38
rs1042031877 Chr.2: 31266862-31266862 on Build GRCh38
rs878890799 Chr.2: 31266762-31266775 on Build GRCh38
rs201820295 Chr.2: 31266738-31266738 on Build GRCh38
rs368656515 Chr.2: 31266698-31266698 on Build GRCh38
rs368461616 Chr.2: 31266694-31266694 on Build GRCh38
rs745334247 Chr.2: 31266654-31266654 on Build GRCh38
rs375673528 Chr.2: 31266779-31266794 on Build GRCh38
rs771584509 Chr.2: 31266579-31266579 on Build GRCh38
rs914536512 Chr.2: 31266645-31266645 on Build GRCh38
rs1003041015 Chr.2: 31266696-31266696 on Build GRCh38
rs762993184 Chr.2: 31266779-31266780 on Build GRCh38
rs541802949 Chr.2: 31266587-31266587 on Build GRCh38
rs766957331 Chr.2: 31266685-31266685 on Build GRCh38
rs17011170 Chr.2: 31266932-31266932 on Build GRCh38
rs771581623 Chr.2: 31266754-31266754 on Build GRCh38
rs375529279 Chr.2: 31266639-31266639 on Build GRCh38
rs3209660 Chr.2: 31266795-31266795 on Build GRCh38
rs868772924 Chr.2: 31266879-31266879 on Build GRCh38
rs370382338 Chr.2: 31266513-31266513 on Build GRCh38
rs777822714 Chr.2: 31266642-31266642 on Build GRCh38
rs200037726 Chr.2: 31266601-31266601 on Build GRCh38
rs758488379 Chr.2: 31266508-31266508 on Build GRCh38
rs200698516 Chr.2: 31266717-31266717 on Build GRCh38
rs546891895 Chr.2: 31266797-31266797 on Build GRCh38
rs553843907 Chr.2: 31266830-31266830 on Build GRCh38
rs965709531 Chr.2: 31266705-31266705 on Build GRCh38
rs751875498 Chr.2: 31266605-31266605 on Build GRCh38
rs557816559 Chr.2: 31266890-31266890 on Build GRCh38
rs188280370 Chr.2: 31266857-31266857 on Build GRCh38
rs777501539 Chr.2: 31266533-31266533 on Build GRCh38
rs538388340 Chr.2: 31266777-31266800 on Build GRCh38
rs567122967 Chr.2: 31266801-31266801 on Build GRCh38
rs374806899 Chr.2: 31266711-31266711 on Build GRCh38
rs764419322 Chr.2: 31266488-31266488 on Build GRCh38
rs752027133 Chr.2: 31266762-31266767 on Build GRCh38
rs746607553 Chr.2: 31266727-31266727 on Build GRCh38
rs961890359 Chr.2: 31266810-31266811 on Build GRCh38
rs895195763 Chr.2: 31266827-31266827 on Build GRCh38
rs182810757 Chr.2: 31266565-31266565 on Build GRCh38
rs141403133 Chr.2: 31266500-31266500 on Build GRCh38
rs975888686 Chr.2: 31266811-31266811 on Build GRCh38
rs1037315084 Chr.2: 31266791-31266791 on Build GRCh38
rs761084914 Chr.2: 31266485-31266485 on Build GRCh38
rs750963219 Chr.2: 31266591-31266591 on Build GRCh38
rs965678087 Chr.2: 31266876-31266876 on Build GRCh38
rs749601475 Chr.2: 31266744-31266744 on Build GRCh38
rs888671803 Chr.2: 31266526-31266526 on Build GRCh38
rs644926 Chr.2: 31266861-31266861 on Build GRCh38
rs765780879 Chr.2: 31266590-31266590 on Build GRCh38
rs3031889 Chr.2: 31266784-31266794 on Build GRCh38
rs756180065 Chr.2: 31266777-31266781 on Build GRCh38
rs73925220 Chr.2: 31266891-31266891 on Build GRCh38
rs757832265 Chr.2: 31266722-31266722 on Build GRCh38
rs770044816 Chr.2: 31266675-31266675 on Build GRCh38
rs755854068 Chr.2: 31266641-31266641 on Build GRCh38
rs748861373 Chr.2: 31266543-31266543 on Build GRCh38
rs139970694 Chr.2: 31266506-31266506 on Build GRCh38
rs894969230 Chr.2: 31266638-31266638 on Build GRCh38
rs541464935 Chr.2: 31266793-31266794 on Build GRCh38
rs267599346 Chr.2: 31266689-31266689 on Build GRCh38
rs561678984 Chr.2: 31266662-31266662 on Build GRCh38
rs774988428 Chr.2: 31266756-31266756 on Build GRCh38
rs749199744 Chr.2: 31266644-31266644 on Build GRCh38
rs370672845 Chr.2: 31266602-31266602 on Build GRCh38
rs1023317840 Chr.2: 31266760-31266760 on Build GRCh38
rs145066486 Chr.2: 31266532-31266532 on Build GRCh38
rs60629834 Chr.2: 31266794-31266795 on Build GRCh38
rs922757129 Chr.2: 31266486-31266486 on Build GRCh38
rs1057314080 Chr.2: 31266897-31266897 on Build GRCh38

View Full Product Details