Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36560604-36561114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ2
Entrez Gene ID
9637
Gene Name
fasciculation and elongation protein zeta 2
Gene Aliases
HUM3CL
UniGene
Hs.258563
Gene Chromosome Location
Chr.2: 36552239-36598190 on Build GRCh38
Transcripts
NM_001042548.1 NM_005102.2 XM_006712153.2

SNP Detail

SNP ID Chromosome Location
rs369596428 Chr.2: 36560819-36560819 on Build GRCh38
rs920801919 Chr.2: 36561049-36561049 on Build GRCh38
rs754628417 Chr.2: 36560792-36560792 on Build GRCh38
rs779321744 Chr.2: 36560810-36560810 on Build GRCh38
rs201859002 Chr.2: 36560756-36560756 on Build GRCh38
rs375896608 Chr.2: 36560874-36560874 on Build GRCh38
rs780624804 Chr.2: 36560949-36560949 on Build GRCh38
rs1029181013 Chr.2: 36560965-36560965 on Build GRCh38
rs908142284 Chr.2: 36560705-36560705 on Build GRCh38
rs958269863 Chr.2: 36560665-36560665 on Build GRCh38
rs940962630 Chr.2: 36561025-36561025 on Build GRCh38
rs766145457 Chr.2: 36560781-36560781 on Build GRCh38
rs920549068 Chr.2: 36560848-36560848 on Build GRCh38
rs563953341 Chr.2: 36560865-36560865 on Build GRCh38
rs780346971 Chr.2: 36560813-36560813 on Build GRCh38
rs750923190 Chr.2: 36560875-36560875 on Build GRCh38
rs982331853 Chr.2: 36560702-36560702 on Build GRCh38
rs375995625 Chr.2: 36560801-36560801 on Build GRCh38
rs770008552 Chr.2: 36560850-36560850 on Build GRCh38
rs767037891 Chr.2: 36560863-36560863 on Build GRCh38
rs774061037 Chr.2: 36560854-36560854 on Build GRCh38
rs572275130 Chr.2: 36560820-36560820 on Build GRCh38
rs201386130 Chr.2: 36560809-36560809 on Build GRCh38
rs906605947 Chr.2: 36560931-36560931 on Build GRCh38
rs767031234 Chr.2: 36560798-36560798 on Build GRCh38
rs543513119 Chr.2: 36560909-36560909 on Build GRCh38
rs192843973 Chr.2: 36560884-36560884 on Build GRCh38
rs1046943135 Chr.2: 36560920-36560920 on Build GRCh38
rs201451880 Chr.2: 36560852-36560852 on Build GRCh38
rs746151102 Chr.2: 36560751-36560751 on Build GRCh38
rs910966357 Chr.2: 36561084-36561084 on Build GRCh38
rs547509204 Chr.2: 36560963-36560963 on Build GRCh38
rs751222555 Chr.2: 36560787-36560787 on Build GRCh38
rs930520533 Chr.2: 36560849-36560849 on Build GRCh38
rs982110844 Chr.2: 36560981-36560981 on Build GRCh38
rs776626587 Chr.2: 36560767-36560767 on Build GRCh38
rs747158150 Chr.2: 36560841-36560841 on Build GRCh38
rs75497421 Chr.2: 36561079-36561079 on Build GRCh38
rs746190039 Chr.2: 36560749-36560749 on Build GRCh38
rs989687967 Chr.2: 36560688-36560688 on Build GRCh38
rs990948233 Chr.2: 36560957-36560957 on Build GRCh38
rs377511278 Chr.2: 36560919-36560919 on Build GRCh38
rs574473483 Chr.2: 36560679-36560679 on Build GRCh38
rs770730428 Chr.2: 36560853-36560853 on Build GRCh38
rs776558632 Chr.2: 36561016-36561017 on Build GRCh38
rs973550336 Chr.2: 36561046-36561046 on Build GRCh38
rs562087595 Chr.2: 36560937-36560937 on Build GRCh38
rs1031288356 Chr.2: 36560657-36560657 on Build GRCh38
rs878878566 Chr.2: 36561030-36561030 on Build GRCh38
rs372609415 Chr.2: 36560890-36560890 on Build GRCh38
rs762780271 Chr.2: 36560779-36560779 on Build GRCh38
rs762008568 Chr.2: 36560774-36560774 on Build GRCh38
rs748175643 Chr.2: 36560846-36560846 on Build GRCh38
rs781218152 Chr.2: 36560903-36560903 on Build GRCh38
rs759395676 Chr.2: 36560858-36560858 on Build GRCh38
rs940805932 Chr.2: 36560724-36560724 on Build GRCh38
rs569328230 Chr.2: 36561018-36561018 on Build GRCh38
rs529378665 Chr.2: 36560958-36560958 on Build GRCh38
rs958343737 Chr.2: 36560951-36560951 on Build GRCh38
rs1011128741 Chr.2: 36560962-36560962 on Build GRCh38
rs556249036 Chr.2: 36560652-36560652 on Build GRCh38
rs755652536 Chr.2: 36560807-36560807 on Build GRCh38
rs769990207 Chr.2: 36560915-36560915 on Build GRCh38
rs765231127 Chr.2: 36560778-36560778 on Build GRCh38
rs940388954 Chr.2: 36560811-36560811 on Build GRCh38
rs949655813 Chr.2: 36560631-36560631 on Build GRCh38
rs987643644 Chr.2: 36561067-36561067 on Build GRCh38
rs987602166 Chr.2: 36560759-36560759 on Build GRCh38
rs932156630 Chr.2: 36561077-36561077 on Build GRCh38
rs761758135 Chr.2: 36561015-36561015 on Build GRCh38
rs758940921 Chr.2: 36560883-36560883 on Build GRCh38
rs189078080 Chr.2: 36560753-36560753 on Build GRCh38
rs746383880 Chr.2: 36560812-36560812 on Build GRCh38
rs372339512 Chr.2: 36560873-36560873 on Build GRCh38

View Full Product Details