Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37084362-37084798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH11
Entrez Gene ID
253635
Gene Name
G-patch domain containing 11
Gene Aliases
CCDC75, CENP-Y, CENPY
UniGene
Hs.595250
Gene Chromosome Location
Chr.2: 37084451-37099244 on Build GRCh38
Transcripts
NM_001278505.1 NM_001322249.1 NM_174931.3 XM_017003748.1 XM_006712035.3

Gene Symbol
HEATR5B
Entrez Gene ID
54497
Gene Name
HEAT repeat containing 5B
Gene Aliases
-
UniGene
Hs.744084
Gene Chromosome Location
Chr.2: 36980999-37084384 on Build GRCh38
Transcripts
NM_001278505.1 NM_001322249.1 NM_174931.3 XM_017003748.1 XM_006712035.3

SNP Detail

SNP ID Chromosome Location
rs1040002488 Chr.2: 37084398-37084398 on Build GRCh38
rs561174560 Chr.2: 37084454-37084454 on Build GRCh38
rs958977323 Chr.2: 37084535-37084535 on Build GRCh38
rs1016694207 Chr.2: 37084466-37084466 on Build GRCh38
rs936028487 Chr.2: 37084388-37084388 on Build GRCh38
rs367928922 Chr.2: 37084714-37084714 on Build GRCh38
rs899728917 Chr.2: 37084412-37084412 on Build GRCh38
rs947894762 Chr.2: 37084552-37084552 on Build GRCh38
rs181623560 Chr.2: 37084704-37084704 on Build GRCh38
rs550169999 Chr.2: 37084551-37084551 on Build GRCh38
rs1003928114 Chr.2: 37084465-37084465 on Build GRCh38
rs748772645 Chr.2: 37084570-37084570 on Build GRCh38
rs559263737 Chr.2: 37084732-37084732 on Build GRCh38
rs1019225961 Chr.2: 37084503-37084503 on Build GRCh38
rs571230302 Chr.2: 37084530-37084530 on Build GRCh38
rs1014423092 Chr.2: 37084743-37084743 on Build GRCh38
rs897599552 Chr.2: 37084396-37084396 on Build GRCh38
rs374862659 Chr.2: 37084441-37084441 on Build GRCh38
rs753037254 Chr.2: 37084521-37084521 on Build GRCh38
rs993077165 Chr.2: 37084537-37084537 on Build GRCh38
rs531946654 Chr.2: 37084543-37084543 on Build GRCh38
rs564496340 Chr.2: 37084577-37084577 on Build GRCh38
rs957400301 Chr.2: 37084655-37084655 on Build GRCh38
rs772740672 Chr.2: 37084766-37084766 on Build GRCh38
rs745470188 Chr.2: 37084729-37084729 on Build GRCh38
rs745464732 Chr.2: 37084528-37084528 on Build GRCh38
rs888843082 Chr.2: 37084585-37084585 on Build GRCh38
rs778860664 Chr.2: 37084505-37084505 on Build GRCh38
rs995030431 Chr.2: 37084571-37084571 on Build GRCh38
rs1027414575 Chr.2: 37084572-37084572 on Build GRCh38
rs998131912 Chr.2: 37084433-37084433 on Build GRCh38
rs1036462039 Chr.2: 37084563-37084563 on Build GRCh38
rs895863856 Chr.2: 37084558-37084573 on Build GRCh38
rs756875720 Chr.2: 37084593-37084593 on Build GRCh38
rs953519547 Chr.2: 37084453-37084453 on Build GRCh38
rs989962767 Chr.2: 37084429-37084429 on Build GRCh38
rs962665688 Chr.2: 37084510-37084510 on Build GRCh38
rs771760584 Chr.2: 37084758-37084758 on Build GRCh38
rs994579196 Chr.2: 37084397-37084397 on Build GRCh38
rs185904752 Chr.2: 37084762-37084762 on Build GRCh38
rs966718159 Chr.2: 37084611-37084611 on Build GRCh38
rs568857434 Chr.2: 37084554-37084554 on Build GRCh38
rs377680228 Chr.2: 37084598-37084598 on Build GRCh38
rs960362774 Chr.2: 37084737-37084737 on Build GRCh38
rs143944589 Chr.2: 37084645-37084645 on Build GRCh38
rs1019110515 Chr.2: 37084607-37084607 on Build GRCh38
rs567082815 Chr.2: 37084646-37084646 on Build GRCh38
rs536388024 Chr.2: 37084561-37084561 on Build GRCh38
rs528356748 Chr.2: 37084484-37084484 on Build GRCh38
rs1033790635 Chr.2: 37084532-37084532 on Build GRCh38
rs202237272 Chr.2: 37084428-37084429 on Build GRCh38
rs770780127 Chr.2: 37084492-37084492 on Build GRCh38
rs778506374 Chr.2: 37084689-37084689 on Build GRCh38
rs556496869 Chr.2: 37084557-37084558 on Build GRCh38
rs1004064046 Chr.2: 37084695-37084695 on Build GRCh38
rs546517158 Chr.2: 37084501-37084501 on Build GRCh38
rs3084427 Chr.2: 37084531-37084532 on Build GRCh38
rs745957565 Chr.2: 37084525-37084525 on Build GRCh38
rs756103268 Chr.2: 37084386-37084386 on Build GRCh38
rs906475995 Chr.2: 37084559-37084559 on Build GRCh38
rs917373307 Chr.2: 37084540-37084540 on Build GRCh38
rs796288665 Chr.2: 37084531-37084531 on Build GRCh38
rs898227434 Chr.2: 37084712-37084712 on Build GRCh38
rs977697303 Chr.2: 37084625-37084626 on Build GRCh38
rs915366667 Chr.2: 37084434-37084434 on Build GRCh38
rs933893600 Chr.2: 37084599-37084599 on Build GRCh38
rs111750489 Chr.2: 37084523-37084523 on Build GRCh38

View Full Product Details