Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37090575-37090974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH11
Entrez Gene ID
253635
Gene Name
G-patch domain containing 11
Gene Aliases
CCDC75, CENP-Y, CENPY
UniGene
Hs.595250
Gene Chromosome Location
Chr.2: 37084451-37099244 on Build GRCh38
Transcripts
NM_001278505.1 NM_001322249.1 NM_174931.3 XM_017003748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407957 c.291A>G p.E97E Chr.2: 37090697-37090697 on Build GRCh38
6563051 c.301C>G p.L101V Chr.2: 37090707-37090707 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945949459 Chr.2: 37090919-37090919 on Build GRCh38
rs778713953 Chr.2: 37090847-37090847 on Build GRCh38
rs759051319 Chr.2: 37090690-37090690 on Build GRCh38
rs777274706 Chr.2: 37090766-37090766 on Build GRCh38
rs755219361 Chr.2: 37090611-37090611 on Build GRCh38
rs571016756 Chr.2: 37090850-37090850 on Build GRCh38
rs999916580 Chr.2: 37090717-37090717 on Build GRCh38
rs571898620 Chr.2: 37090767-37090767 on Build GRCh38
rs962277189 Chr.2: 37090848-37090848 on Build GRCh38
rs201403331 Chr.2: 37090803-37090804 on Build GRCh38
rs187648741 Chr.2: 37090802-37090802 on Build GRCh38
rs997837460 Chr.2: 37090739-37090739 on Build GRCh38
rs752196335 Chr.2: 37090686-37090686 on Build GRCh38
rs749090899 Chr.2: 37090681-37090681 on Build GRCh38
rs1011988044 Chr.2: 37090785-37090785 on Build GRCh38
rs889621942 Chr.2: 37090616-37090616 on Build GRCh38
rs904360750 Chr.2: 37090701-37090701 on Build GRCh38
rs183398888 Chr.2: 37090631-37090631 on Build GRCh38
rs954981555 Chr.2: 37090767-37090767 on Build GRCh38
rs548688624 Chr.2: 37090687-37090687 on Build GRCh38
rs200460070 Chr.2: 37090663-37090663 on Build GRCh38
rs963811240 Chr.2: 37090727-37090727 on Build GRCh38
rs927364925 Chr.2: 37090828-37090828 on Build GRCh38
rs4670655 Chr.2: 37090898-37090898 on Build GRCh38
rs973436555 Chr.2: 37090824-37090824 on Build GRCh38
rs552614113 Chr.2: 37090808-37090808 on Build GRCh38
rs984112160 Chr.2: 37090796-37090796 on Build GRCh38
rs963527279 Chr.2: 37090811-37090811 on Build GRCh38
rs4670654 Chr.2: 37090726-37090726 on Build GRCh38
rs893583660 Chr.2: 37090735-37090735 on Build GRCh38
rs13425846 Chr.2: 37090945-37090945 on Build GRCh38
rs970923021 Chr.2: 37090804-37090804 on Build GRCh38
rs149446111 Chr.2: 37090863-37090863 on Build GRCh38
rs746322698 Chr.2: 37090768-37090768 on Build GRCh38
rs1023770526 Chr.2: 37090788-37090788 on Build GRCh38
rs550875824 Chr.2: 37090940-37090940 on Build GRCh38
rs895722055 Chr.2: 37090623-37090623 on Build GRCh38
rs780879728 Chr.2: 37090683-37090683 on Build GRCh38

View Full Product Details