Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37106728-37107251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK2
Entrez Gene ID
5610
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases
EIF2AK1, PKR, PPP1R83, PRKR
UniGene
Hs.131431
Gene Chromosome Location
Chr.2: 37099210-37157065 on Build GRCh38
Transcripts
NM_001135651.2 NM_001135652.2 NM_002759.3 XM_011532987.2 XM_017004503.1

SNP Detail

SNP ID Chromosome Location
rs886245466 Chr.2: 37107015-37107015 on Build GRCh38
rs1033951774 Chr.2: 37106865-37106865 on Build GRCh38
rs148247112 Chr.2: 37106773-37106773 on Build GRCh38
rs767770189 Chr.2: 37107197-37107197 on Build GRCh38
rs1024774363 Chr.2: 37106989-37106989 on Build GRCh38
rs1016574625 Chr.2: 37107151-37107151 on Build GRCh38
rs895904126 Chr.2: 37106905-37106905 on Build GRCh38
rs577720765 Chr.2: 37106810-37106810 on Build GRCh38
rs966857451 Chr.2: 37106878-37106878 on Build GRCh38
rs1014256113 Chr.2: 37106934-37106934 on Build GRCh38
rs1004818496 Chr.2: 37107091-37107091 on Build GRCh38
rs925314078 Chr.2: 37106974-37106974 on Build GRCh38
rs773547953 Chr.2: 37106772-37106772 on Build GRCh38
rs745562740 Chr.2: 37107057-37107058 on Build GRCh38
rs544804617 Chr.2: 37106873-37106873 on Build GRCh38
rs913363338 Chr.2: 37106781-37106781 on Build GRCh38
rs201533392 Chr.2: 37107042-37107042 on Build GRCh38
rs987635141 Chr.2: 37106774-37106774 on Build GRCh38
rs556831739 Chr.2: 37107029-37107029 on Build GRCh38
rs752759198 Chr.2: 37107222-37107222 on Build GRCh38
rs979392918 Chr.2: 37106931-37106931 on Build GRCh38
rs573131592 Chr.2: 37106768-37106769 on Build GRCh38
rs397718360 Chr.2: 37107055-37107056 on Build GRCh38
rs770784762 Chr.2: 37106901-37106901 on Build GRCh38
rs1038904176 Chr.2: 37107218-37107218 on Build GRCh38
rs34203821 Chr.2: 37107042-37107043 on Build GRCh38
rs756098916 Chr.2: 37107227-37107227 on Build GRCh38
rs186609289 Chr.2: 37107173-37107173 on Build GRCh38
rs962944328 Chr.2: 37106822-37106822 on Build GRCh38
rs943270983 Chr.2: 37107184-37107184 on Build GRCh38
rs1006298826 Chr.2: 37106809-37106809 on Build GRCh38
rs755192926 Chr.2: 37107166-37107166 on Build GRCh38
rs995833347 Chr.2: 37107225-37107225 on Build GRCh38
rs992344172 Chr.2: 37106869-37106869 on Build GRCh38
rs776085688 Chr.2: 37107056-37107057 on Build GRCh38
rs1002482641 Chr.2: 37106830-37106830 on Build GRCh38
rs766309019 Chr.2: 37106867-37106867 on Build GRCh38
rs765719032 Chr.2: 37106770-37106770 on Build GRCh38
rs879652670 Chr.2: 37106831-37106831 on Build GRCh38
rs1043678250 Chr.2: 37106856-37106856 on Build GRCh38
rs1055842410 Chr.2: 37106875-37106875 on Build GRCh38

View Full Product Details