Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37644664-37645174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP3
Entrez Gene ID
10602
Gene Name
CDC42 effector protein 3
Gene Aliases
BORG2, CEP3, UB1
UniGene
Hs.369574
Gene Chromosome Location
Chr.2: 37641882-37672535 on Build GRCh38
Transcripts
NM_001270436.1 NM_001270437.1 NM_001270438.1 NM_006449.4 XM_011532482.2

SNP Detail

SNP ID Chromosome Location
rs532114923 Chr.2: 37644990-37644990 on Build GRCh38
rs985451058 Chr.2: 37644972-37644972 on Build GRCh38
rs1010187340 Chr.2: 37645148-37645148 on Build GRCh38
rs56719269 Chr.2: 37644757-37644757 on Build GRCh38
rs947494310 Chr.2: 37644762-37644762 on Build GRCh38
rs942286851 Chr.2: 37645033-37645034 on Build GRCh38
rs897407148 Chr.2: 37644834-37644834 on Build GRCh38
rs537123098 Chr.2: 37644876-37644883 on Build GRCh38
rs914570519 Chr.2: 37644760-37644760 on Build GRCh38
rs1051409214 Chr.2: 37645090-37645090 on Build GRCh38
rs934523994 Chr.2: 37644707-37644707 on Build GRCh38
rs1035972656 Chr.2: 37645104-37645104 on Build GRCh38
rs928558857 Chr.2: 37644706-37644706 on Build GRCh38
rs1005857964 Chr.2: 37645049-37645049 on Build GRCh38
rs1003120288 Chr.2: 37644934-37644934 on Build GRCh38
rs1005952500 Chr.2: 37644912-37644912 on Build GRCh38
rs191628111 Chr.2: 37645130-37645130 on Build GRCh38
rs1047878573 Chr.2: 37644709-37644709 on Build GRCh38
rs955977702 Chr.2: 37644946-37644946 on Build GRCh38
rs555231343 Chr.2: 37644887-37644899 on Build GRCh38
rs182607170 Chr.2: 37644878-37644878 on Build GRCh38
rs988821300 Chr.2: 37644710-37644710 on Build GRCh38
rs1002620497 Chr.2: 37645102-37645102 on Build GRCh38
rs1041195356 Chr.2: 37644783-37644783 on Build GRCh38
rs566418631 Chr.2: 37645015-37645015 on Build GRCh38
rs922178275 Chr.2: 37645080-37645080 on Build GRCh38
rs10694 Chr.2: 37644996-37644996 on Build GRCh38
rs372860359 Chr.2: 37644930-37644930 on Build GRCh38
rs1044025442 Chr.2: 37645070-37645070 on Build GRCh38
rs186884058 Chr.2: 37645009-37645009 on Build GRCh38
rs751540431 Chr.2: 37644895-37644895 on Build GRCh38
rs1034099608 Chr.2: 37644942-37644943 on Build GRCh38
rs997271674 Chr.2: 37644784-37644784 on Build GRCh38
rs888666847 Chr.2: 37645039-37645039 on Build GRCh38
rs909354831 Chr.2: 37645018-37645020 on Build GRCh38
rs1041642673 Chr.2: 37644995-37644995 on Build GRCh38
rs1043288660 Chr.2: 37645120-37645120 on Build GRCh38
rs572370645 Chr.2: 37644703-37644703 on Build GRCh38
rs917998466 Chr.2: 37644695-37644695 on Build GRCh38
rs143034717 Chr.2: 37644840-37644840 on Build GRCh38
rs969807023 Chr.2: 37644933-37644933 on Build GRCh38
rs929511020 Chr.2: 37644706-37644707 on Build GRCh38
rs562367954 Chr.2: 37644970-37644970 on Build GRCh38
rs944216725 Chr.2: 37644782-37644782 on Build GRCh38
rs564850520 Chr.2: 37644989-37644989 on Build GRCh38
rs968693819 Chr.2: 37644953-37644953 on Build GRCh38
rs905615599 Chr.2: 37645081-37645081 on Build GRCh38
rs911314292 Chr.2: 37644977-37644977 on Build GRCh38
rs942275809 Chr.2: 37644778-37644778 on Build GRCh38
rs988726004 Chr.2: 37644947-37644947 on Build GRCh38
rs143650183 Chr.2: 37644899-37644899 on Build GRCh38
rs911424364 Chr.2: 37644780-37644780 on Build GRCh38
rs914601755 Chr.2: 37644952-37644952 on Build GRCh38
rs144639735 Chr.2: 37644872-37644892 on Build GRCh38
rs1009959958 Chr.2: 37645119-37645119 on Build GRCh38
rs532311840 Chr.2: 37644887-37644887 on Build GRCh38
rs780146187 Chr.2: 37644779-37644779 on Build GRCh38
rs533565171 Chr.2: 37645059-37645059 on Build GRCh38
rs552244487 Chr.2: 37645106-37645106 on Build GRCh38
rs1035497863 Chr.2: 37644939-37644939 on Build GRCh38

View Full Product Details