Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38813761-38814280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX57
Entrez Gene ID
90957
Gene Name
DEAH-box helicase 57
Gene Aliases
DDX57
UniGene
Hs.468226
Gene Chromosome Location
Chr.2: 38797729-38875883 on Build GRCh38
Transcripts
NM_198963.1 XM_011533154.2 XM_011533155.2 XM_011533156.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094163 c.3667C>G p.P1223A Chr.2: 38813835-38813835 on Build GRCh38
1250063 c.3619G>A p.A1207T Chr.2: 38813883-38813883 on Build GRCh38
1637970 c.3639A>T p.I1213I Chr.2: 38813863-38813863 on Build GRCh38
4916661 c.3639A>G p.I1213M Chr.2: 38813863-38813863 on Build GRCh38
7232512 c.3622G>A p.E1208K Chr.2: 38813880-38813880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200596889 Chr.2: 38813803-38813803 on Build GRCh38
rs1034132465 Chr.2: 38813949-38813949 on Build GRCh38
rs780009363 Chr.2: 38813831-38813831 on Build GRCh38
rs370295178 Chr.2: 38813826-38813826 on Build GRCh38
rs571561326 Chr.2: 38813992-38813992 on Build GRCh38
rs1055116913 Chr.2: 38814146-38814146 on Build GRCh38
rs933623455 Chr.2: 38814155-38814155 on Build GRCh38
rs990194731 Chr.2: 38814094-38814094 on Build GRCh38
rs1002817744 Chr.2: 38814052-38814052 on Build GRCh38
rs921622505 Chr.2: 38814080-38814080 on Build GRCh38
rs766191440 Chr.2: 38814062-38814062 on Build GRCh38
rs758314882 Chr.2: 38813830-38813830 on Build GRCh38
rs117031610 Chr.2: 38814193-38814193 on Build GRCh38
rs902147065 Chr.2: 38813909-38813909 on Build GRCh38
rs765474041 Chr.2: 38813907-38813907 on Build GRCh38
rs899076244 Chr.2: 38814104-38814104 on Build GRCh38
rs748395705 Chr.2: 38813914-38813917 on Build GRCh38
rs773695856 Chr.2: 38813941-38813942 on Build GRCh38
rs1050747655 Chr.2: 38814158-38814158 on Build GRCh38
rs958570675 Chr.2: 38814040-38814040 on Build GRCh38
rs994390222 Chr.2: 38814225-38814225 on Build GRCh38
rs201516645 Chr.2: 38813816-38813816 on Build GRCh38
rs754122218 Chr.2: 38813811-38813811 on Build GRCh38
rs770298204 Chr.2: 38813942-38813943 on Build GRCh38
rs181107316 Chr.2: 38813899-38813899 on Build GRCh38
rs777128438 Chr.2: 38813898-38813898 on Build GRCh38
rs940469675 Chr.2: 38814098-38814098 on Build GRCh38
rs2122769 Chr.2: 38813999-38813999 on Build GRCh38
rs747864012 Chr.2: 38813862-38813862 on Build GRCh38
rs769476598 Chr.2: 38813868-38813868 on Build GRCh38
rs548777494 Chr.2: 38813809-38813809 on Build GRCh38
rs770310943 Chr.2: 38813875-38813875 on Build GRCh38
rs781024818 Chr.2: 38813933-38813933 on Build GRCh38
rs755765861 Chr.2: 38813937-38813937 on Build GRCh38
rs759126884 Chr.2: 38814177-38814177 on Build GRCh38
rs1032517147 Chr.2: 38814032-38814032 on Build GRCh38
rs375666982 Chr.2: 38813934-38813934 on Build GRCh38
rs758536629 Chr.2: 38813919-38813919 on Build GRCh38
rs573904486 Chr.2: 38814099-38814099 on Build GRCh38
rs987193575 Chr.2: 38814088-38814088 on Build GRCh38
rs551025245 Chr.2: 38814084-38814084 on Build GRCh38
rs1025034846 Chr.2: 38814004-38814004 on Build GRCh38
rs763085866 Chr.2: 38813942-38813942 on Build GRCh38
rs762695949 Chr.2: 38814034-38814034 on Build GRCh38
rs546958285 Chr.2: 38813972-38813972 on Build GRCh38
rs746909137 Chr.2: 38813843-38813843 on Build GRCh38
rs977677633 Chr.2: 38814131-38814131 on Build GRCh38
rs368699930 Chr.2: 38813912-38813912 on Build GRCh38
rs1034345583 Chr.2: 38814066-38814066 on Build GRCh38
rs112437127 Chr.2: 38814019-38814019 on Build GRCh38
rs184350209 Chr.2: 38814222-38814222 on Build GRCh38
rs555392829 Chr.2: 38814097-38814097 on Build GRCh38
rs923498449 Chr.2: 38814140-38814140 on Build GRCh38
rs766312892 Chr.2: 38813923-38813923 on Build GRCh38
rs764554764 Chr.2: 38813894-38813894 on Build GRCh38
rs910893075 Chr.2: 38814159-38814159 on Build GRCh38
rs756668586 Chr.2: 38814246-38814246 on Build GRCh38
rs780899834 Chr.2: 38813857-38813857 on Build GRCh38
rs966821338 Chr.2: 38814008-38814008 on Build GRCh38
rs539476356 Chr.2: 38813998-38813998 on Build GRCh38
rs537571375 Chr.2: 38814022-38814022 on Build GRCh38
rs937285895 Chr.2: 38814132-38814132 on Build GRCh38
rs754781303 Chr.2: 38813856-38813856 on Build GRCh38
rs4670897 Chr.2: 38814217-38814217 on Build GRCh38
rs940522076 Chr.2: 38813925-38813925 on Build GRCh38
rs776167951 Chr.2: 38813888-38813888 on Build GRCh38
rs1024981088 Chr.2: 38814209-38814209 on Build GRCh38
rs761201191 Chr.2: 38813889-38813889 on Build GRCh38
rs553282431 Chr.2: 38814215-38814215 on Build GRCh38
rs372332413 Chr.2: 38813926-38813926 on Build GRCh38
rs201931249 Chr.2: 38813945-38813945 on Build GRCh38
rs777516184 Chr.2: 38813942-38813942 on Build GRCh38

View Full Product Details