Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38919238-38919742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF33
Entrez Gene ID
100271715
Gene Name
Rho guanine nucleotide exchange factor 33
Gene Aliases
-
UniGene
Hs.99841
Gene Chromosome Location
Chr.2: 38919363-38975449 on Build GRCh38
Transcripts
NM_001145451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3729889 c.5delA p.T4fs*13 Chr.2: 38919452-38919452 on Build GRCh38
6908035 c.25+2T>C p.? Chr.2: 38919474-38919474 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs536389416 Chr.2: 38919309-38919309 on Build GRCh38
rs902982955 Chr.2: 38919593-38919593 on Build GRCh38
rs558822315 Chr.2: 38919558-38919558 on Build GRCh38
rs751079168 Chr.2: 38919467-38919467 on Build GRCh38
rs375480888 Chr.2: 38919444-38919444 on Build GRCh38
rs371189493 Chr.2: 38919441-38919441 on Build GRCh38
rs1055378302 Chr.2: 38919468-38919468 on Build GRCh38
rs929616463 Chr.2: 38919401-38919401 on Build GRCh38
rs938387470 Chr.2: 38919464-38919464 on Build GRCh38
rs1007593985 Chr.2: 38919712-38919712 on Build GRCh38
rs1011721464 Chr.2: 38919542-38919542 on Build GRCh38
rs766422335 Chr.2: 38919586-38919586 on Build GRCh38
rs559158182 Chr.2: 38919587-38919587 on Build GRCh38
rs779611535 Chr.2: 38919463-38919463 on Build GRCh38
rs942096104 Chr.2: 38919395-38919395 on Build GRCh38
rs973620151 Chr.2: 38919396-38919396 on Build GRCh38
rs562375562 Chr.2: 38919660-38919660 on Build GRCh38
rs754882170 Chr.2: 38919353-38919353 on Build GRCh38
rs750138898 Chr.2: 38919458-38919458 on Build GRCh38
rs371790109 Chr.2: 38919530-38919530 on Build GRCh38
rs1003455621 Chr.2: 38919501-38919501 on Build GRCh38
rs73930376 Chr.2: 38919599-38919599 on Build GRCh38
rs574364867 Chr.2: 38919690-38919690 on Build GRCh38
rs944239279 Chr.2: 38919348-38919348 on Build GRCh38
rs11680671 Chr.2: 38919683-38919683 on Build GRCh38
rs985604686 Chr.2: 38919317-38919317 on Build GRCh38
rs1021731258 Chr.2: 38919594-38919594 on Build GRCh38
rs891824321 Chr.2: 38919550-38919550 on Build GRCh38
rs375326777 Chr.2: 38919513-38919513 on Build GRCh38
rs965526553 Chr.2: 38919713-38919713 on Build GRCh38
rs532815086 Chr.2: 38919320-38919320 on Build GRCh38
rs954603128 Chr.2: 38919600-38919600 on Build GRCh38
rs781444055 Chr.2: 38919452-38919452 on Build GRCh38
rs1048165654 Chr.2: 38919423-38919423 on Build GRCh38
rs985637187 Chr.2: 38919686-38919686 on Build GRCh38
rs367866222 Chr.2: 38919453-38919453 on Build GRCh38
rs80087061 Chr.2: 38919349-38919349 on Build GRCh38
rs544896523 Chr.2: 38919612-38919612 on Build GRCh38
rs374488189 Chr.2: 38919490-38919490 on Build GRCh38
rs906461608 Chr.2: 38919499-38919499 on Build GRCh38
rs988945030 Chr.2: 38919265-38919265 on Build GRCh38
rs968938010 Chr.2: 38919295-38919295 on Build GRCh38
rs781134669 Chr.2: 38919700-38919700 on Build GRCh38
rs573211268 Chr.2: 38919409-38919409 on Build GRCh38
rs914629162 Chr.2: 38919267-38919267 on Build GRCh38

View Full Product Details