Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38950886-38951433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF33
Entrez Gene ID
100271715
Gene Name
Rho guanine nucleotide exchange factor 33
Gene Aliases
-
UniGene
Hs.99841
Gene Chromosome Location
Chr.2: 38919363-38975449 on Build GRCh38
Transcripts
NM_001145451.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5547889 c.1053+7delT p.? Chr.2: 38951128-38951128 on Build GRCh38
6199395 c.1006C>T p.R336C Chr.2: 38951074-38951074 on Build GRCh38
5719584 c.1039T>C p.L347L Chr.2: 38951107-38951107 on Build GRCh38
1408041 c.1048G>T p.D350Y Chr.2: 38951116-38951116 on Build GRCh38
1020862 c.1050C>T p.D350D Chr.2: 38951118-38951118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747168764 Chr.2: 38951158-38951158 on Build GRCh38
rs72916860 Chr.2: 38951377-38951377 on Build GRCh38
rs114922379 Chr.2: 38951095-38951095 on Build GRCh38
rs1045723896 Chr.2: 38951304-38951304 on Build GRCh38
rs761319512 Chr.2: 38951233-38951233 on Build GRCh38
rs757968570 Chr.2: 38951225-38951225 on Build GRCh38
rs777240629 Chr.2: 38951251-38951251 on Build GRCh38
rs773022020 Chr.2: 38951017-38951017 on Build GRCh38
rs1000470043 Chr.2: 38950963-38950963 on Build GRCh38
rs984005205 Chr.2: 38951122-38951122 on Build GRCh38
rs769518527 Chr.2: 38950972-38950972 on Build GRCh38
rs750485662 Chr.2: 38951118-38951118 on Build GRCh38
rs892136902 Chr.2: 38950976-38950976 on Build GRCh38
rs754864286 Chr.2: 38951169-38951169 on Build GRCh38
rs907305501 Chr.2: 38951310-38951310 on Build GRCh38
rs529950060 Chr.2: 38951245-38951246 on Build GRCh38
rs971438743 Chr.2: 38950956-38950956 on Build GRCh38
rs1028123911 Chr.2: 38951069-38951069 on Build GRCh38
rs957796824 Chr.2: 38951331-38951331 on Build GRCh38
rs1012930413 Chr.2: 38951288-38951288 on Build GRCh38
rs775131530 Chr.2: 38951173-38951173 on Build GRCh38
rs931147459 Chr.2: 38951236-38951236 on Build GRCh38
rs772627967 Chr.2: 38950987-38950987 on Build GRCh38
rs992867793 Chr.2: 38951160-38951160 on Build GRCh38
rs367684576 Chr.2: 38951369-38951369 on Build GRCh38
rs1032236164 Chr.2: 38950998-38950998 on Build GRCh38
rs1011745663 Chr.2: 38951335-38951335 on Build GRCh38
rs571317945 Chr.2: 38951272-38951272 on Build GRCh38
rs34037582 Chr.2: 38951152-38951153 on Build GRCh38
rs751480297 Chr.2: 38951159-38951159 on Build GRCh38
rs776050646 Chr.2: 38951029-38951029 on Build GRCh38
rs150260831 Chr.2: 38950969-38950969 on Build GRCh38
rs184408864 Chr.2: 38951038-38951038 on Build GRCh38
rs762264172 Chr.2: 38951281-38951281 on Build GRCh38
rs1027989502 Chr.2: 38951338-38951338 on Build GRCh38
rs983953122 Chr.2: 38951353-38951353 on Build GRCh38
rs975117127 Chr.2: 38951059-38951059 on Build GRCh38
rs1036843795 Chr.2: 38951187-38951187 on Build GRCh38
rs533709743 Chr.2: 38951161-38951161 on Build GRCh38
rs749080563 Chr.2: 38950980-38950980 on Build GRCh38
rs776933047 Chr.2: 38951082-38951082 on Build GRCh38
rs764421530 Chr.2: 38951058-38951058 on Build GRCh38
rs441869 Chr.2: 38951352-38951352 on Build GRCh38
rs558451815 Chr.2: 38951259-38951259 on Build GRCh38
rs777989444 Chr.2: 38951127-38951128 on Build GRCh38
rs747776456 Chr.2: 38950970-38950970 on Build GRCh38
rs770589282 Chr.2: 38951033-38951033 on Build GRCh38
rs554200497 Chr.2: 38951044-38951044 on Build GRCh38
rs538286183 Chr.2: 38950911-38950911 on Build GRCh38
rs549898183 Chr.2: 38951192-38951192 on Build GRCh38
rs1031745050 Chr.2: 38951327-38951327 on Build GRCh38
rs919725636 Chr.2: 38951197-38951197 on Build GRCh38
rs990837827 Chr.2: 38951016-38951016 on Build GRCh38
rs758493918 Chr.2: 38951139-38951139 on Build GRCh38
rs192361017 Chr.2: 38950947-38950947 on Build GRCh38
rs900804593 Chr.2: 38951289-38951289 on Build GRCh38
rs764673292 Chr.2: 38951250-38951250 on Build GRCh38
rs373984297 Chr.2: 38951002-38951002 on Build GRCh38
rs546754970 Chr.2: 38951208-38951208 on Build GRCh38
rs904663896 Chr.2: 38950952-38950952 on Build GRCh38
rs957659948 Chr.2: 38951004-38951004 on Build GRCh38
rs894494633 Chr.2: 38950926-38950926 on Build GRCh38
rs965178750 Chr.2: 38951043-38951043 on Build GRCh38
rs1024431568 Chr.2: 38950953-38950953 on Build GRCh38
rs759052337 Chr.2: 38951142-38951142 on Build GRCh38
rs762057719 Chr.2: 38951083-38951083 on Build GRCh38
rs545112765 Chr.2: 38951324-38951324 on Build GRCh38
rs1019694281 Chr.2: 38951021-38951021 on Build GRCh38

View Full Product Details