Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 39225746-39226287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL4
Entrez Gene ID
344387
Gene Name
cyclin dependent kinase like 4
Gene Aliases
-
UniGene
Hs.403201
Gene Chromosome Location
Chr.2: 39174421-39246815 on Build GRCh38
Transcripts
NM_001009565.1 XM_011532815.2 XM_011532816.2 XM_011532817.2 XM_011532819.2 XM_017003981.1 XM_017003982.1 XM_017003983.1 XM_017003984.1 XM_017003985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5905475 c.241G>A p.E81K Chr.2: 39225888-39225888 on Build GRCh38
4816055 c.187C>G p.L63V Chr.2: 39225942-39225942 on Build GRCh38
3426441 c.253C>A p.H85N Chr.2: 39225876-39225876 on Build GRCh38
6410196 c.187C>T p.L63F Chr.2: 39225942-39225942 on Build GRCh38
6408165 c.290G>T p.G97V Chr.2: 39225839-39225839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753956046 Chr.2: 39225951-39225951 on Build GRCh38
rs939884407 Chr.2: 39225867-39225867 on Build GRCh38
rs1028381380 Chr.2: 39226134-39226134 on Build GRCh38
rs548777060 Chr.2: 39226046-39226046 on Build GRCh38
rs776210677 Chr.2: 39225981-39225981 on Build GRCh38
rs372955336 Chr.2: 39225854-39225854 on Build GRCh38
rs767729102 Chr.2: 39225925-39225925 on Build GRCh38
rs199555201 Chr.2: 39225959-39225959 on Build GRCh38
rs936165378 Chr.2: 39226222-39226222 on Build GRCh38
rs996685363 Chr.2: 39225935-39225935 on Build GRCh38
rs972573873 Chr.2: 39226246-39226246 on Build GRCh38
rs772797993 Chr.2: 39226161-39226161 on Build GRCh38
rs776084989 Chr.2: 39225906-39225906 on Build GRCh38
rs766767021 Chr.2: 39225823-39225823 on Build GRCh38
rs760193087 Chr.2: 39225894-39225894 on Build GRCh38
rs757353052 Chr.2: 39225960-39225960 on Build GRCh38
rs150026360 Chr.2: 39225886-39225886 on Build GRCh38
rs745760496 Chr.2: 39225963-39225963 on Build GRCh38
rs917498727 Chr.2: 39226154-39226154 on Build GRCh38
rs534402528 Chr.2: 39226062-39226062 on Build GRCh38
rs759928283 Chr.2: 39225917-39225917 on Build GRCh38
rs999065328 Chr.2: 39226051-39226051 on Build GRCh38
rs777693618 Chr.2: 39225861-39225861 on Build GRCh38
rs781485493 Chr.2: 39225841-39225841 on Build GRCh38
rs749279866 Chr.2: 39225862-39225862 on Build GRCh38
rs200316561 Chr.2: 39225971-39225971 on Build GRCh38
rs989903689 Chr.2: 39226082-39226082 on Build GRCh38
rs771696927 Chr.2: 39225882-39225882 on Build GRCh38
rs751911303 Chr.2: 39225832-39225832 on Build GRCh38
rs61744520 Chr.2: 39225858-39225858 on Build GRCh38
rs957078003 Chr.2: 39226068-39226068 on Build GRCh38
rs774865930 Chr.2: 39225996-39225996 on Build GRCh38
rs538642009 Chr.2: 39225769-39225769 on Build GRCh38
rs768187251 Chr.2: 39225976-39225976 on Build GRCh38
rs778907747 Chr.2: 39225876-39225876 on Build GRCh38
rs751999567 Chr.2: 39225910-39225910 on Build GRCh38
rs373664586 Chr.2: 39225795-39225795 on Build GRCh38
rs953748286 Chr.2: 39226143-39226143 on Build GRCh38
rs753013168 Chr.2: 39225928-39225928 on Build GRCh38
rs778997560 Chr.2: 39225961-39225961 on Build GRCh38
rs201395955 Chr.2: 39225972-39225972 on Build GRCh38
rs961320809 Chr.2: 39226021-39226021 on Build GRCh38
rs536914953 Chr.2: 39225868-39225868 on Build GRCh38
rs756259785 Chr.2: 39225936-39225936 on Build GRCh38
rs771918384 Chr.2: 39225966-39225966 on Build GRCh38
rs932519916 Chr.2: 39225829-39225829 on Build GRCh38
rs777977186 Chr.2: 39225948-39225948 on Build GRCh38
rs971955402 Chr.2: 39226047-39226047 on Build GRCh38
rs376028533 Chr.2: 39225810-39225810 on Build GRCh38
rs745711704 Chr.2: 39226018-39226018 on Build GRCh38
rs558683330 Chr.2: 39226065-39226065 on Build GRCh38
rs138920388 Chr.2: 39225987-39225987 on Build GRCh38
rs780963885 Chr.2: 39226007-39226007 on Build GRCh38
rs745670477 Chr.2: 39225879-39225879 on Build GRCh38
rs577042383 Chr.2: 39226118-39226118 on Build GRCh38
rs1044550472 Chr.2: 39226178-39226178 on Build GRCh38
rs145051446 Chr.2: 39225907-39225907 on Build GRCh38
rs939853504 Chr.2: 39226183-39226183 on Build GRCh38
rs10174296 Chr.2: 39226163-39226163 on Build GRCh38
rs370894516 Chr.2: 39226001-39226001 on Build GRCh38
rs774682653 Chr.2: 39225981-39225982 on Build GRCh38
rs369296581 Chr.2: 39225836-39225836 on Build GRCh38
rs377382590 Chr.2: 39225926-39225926 on Build GRCh38
rs769257384 Chr.2: 39225992-39225992 on Build GRCh38
rs768258427 Chr.2: 39225902-39225902 on Build GRCh38
rs550587595 Chr.2: 39225813-39225813 on Build GRCh38

View Full Product Details