Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42351226-42351693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX7A2L
Entrez Gene ID
9167
Gene Name
cytochrome c oxidase subunit 7A2 like
Gene Aliases
COX7AR, COX7RP, EB1, SIG81
UniGene
Hs.339639
Gene Chromosome Location
Chr.2: 42335561-42368957 on Build GRCh38
Transcripts
NM_001319036.1 NM_001319037.1 NM_001319038.1 NM_001319040.1 NM_004718.3 NR_134947.1 NR_134948.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327080 c.215G>A p.G72D Chr.2: 42351349-42351349 on Build GRCh38
1690512 c.221_222CC>TT p.P74L Chr.2: 42351342-42351343 on Build GRCh38
7115220 c.236G>A p.R79Q Chr.2: 42351328-42351328 on Build GRCh38
3839548 c.298T>C p.Y100H Chr.2: 42351266-42351266 on Build GRCh38
721855 c.223G>A p.V75I Chr.2: 42351341-42351341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964720947 Chr.2: 42351582-42351582 on Build GRCh38
rs371934332 Chr.2: 42351289-42351289 on Build GRCh38
rs752710138 Chr.2: 42351254-42351254 on Build GRCh38
rs760419829 Chr.2: 42351302-42351302 on Build GRCh38
rs145811069 Chr.2: 42351343-42351343 on Build GRCh38
rs11547561 Chr.2: 42351314-42351314 on Build GRCh38
rs984343103 Chr.2: 42351466-42351466 on Build GRCh38
rs576754196 Chr.2: 42351311-42351311 on Build GRCh38
rs61754513 Chr.2: 42351329-42351329 on Build GRCh38
rs150597431 Chr.2: 42351269-42351269 on Build GRCh38
rs189919281 Chr.2: 42351414-42351414 on Build GRCh38
rs760326700 Chr.2: 42351657-42351657 on Build GRCh38
rs952982354 Chr.2: 42351465-42351465 on Build GRCh38
rs769051701 Chr.2: 42351515-42351515 on Build GRCh38
rs746298147 Chr.2: 42351279-42351279 on Build GRCh38
rs185444762 Chr.2: 42351341-42351341 on Build GRCh38
rs1021476087 Chr.2: 42351361-42351361 on Build GRCh38
rs751713037 Chr.2: 42351346-42351346 on Build GRCh38
rs563282755 Chr.2: 42351436-42351436 on Build GRCh38
rs763655573 Chr.2: 42351633-42351633 on Build GRCh38
rs966473316 Chr.2: 42351338-42351338 on Build GRCh38
rs1011138109 Chr.2: 42351601-42351601 on Build GRCh38
rs893763975 Chr.2: 42351596-42351596 on Build GRCh38
rs778980599 Chr.2: 42351252-42351252 on Build GRCh38
rs562260529 Chr.2: 42351648-42351648 on Build GRCh38
rs371849558 Chr.2: 42351404-42351404 on Build GRCh38
rs752319064 Chr.2: 42351360-42351360 on Build GRCh38
rs771427147 Chr.2: 42351383-42351383 on Build GRCh38
rs770186642 Chr.2: 42351285-42351285 on Build GRCh38
rs765050381 Chr.2: 42351335-42351335 on Build GRCh38
rs367796767 Chr.2: 42351435-42351435 on Build GRCh38
rs373859176 Chr.2: 42351342-42351342 on Build GRCh38
rs922580592 Chr.2: 42351539-42351539 on Build GRCh38
rs772857167 Chr.2: 42351386-42351386 on Build GRCh38
rs767554073 Chr.2: 42351281-42351284 on Build GRCh38
rs1041123387 Chr.2: 42351511-42351511 on Build GRCh38
rs757464832 Chr.2: 42351349-42351349 on Build GRCh38
rs551704458 Chr.2: 42351602-42351602 on Build GRCh38
rs766213187 Chr.2: 42351309-42351309 on Build GRCh38
rs940137938 Chr.2: 42351492-42351492 on Build GRCh38
rs893598209 Chr.2: 42351312-42351312 on Build GRCh38
rs1050101366 Chr.2: 42351579-42351579 on Build GRCh38
rs746194691 Chr.2: 42351364-42351364 on Build GRCh38
rs908612122 Chr.2: 42351477-42351477 on Build GRCh38
rs780385935 Chr.2: 42351380-42351380 on Build GRCh38
rs62142636 Chr.2: 42351563-42351563 on Build GRCh38
rs1034772804 Chr.2: 42351664-42351664 on Build GRCh38
rs1009041158 Chr.2: 42351578-42351578 on Build GRCh38
rs992113063 Chr.2: 42351433-42351433 on Build GRCh38
rs542393260 Chr.2: 42351610-42351610 on Build GRCh38
rs755650234 Chr.2: 42351379-42351379 on Build GRCh38
rs367985723 Chr.2: 42351382-42351382 on Build GRCh38
rs533082726 Chr.2: 42351558-42351558 on Build GRCh38
rs2534616 Chr.2: 42351653-42351653 on Build GRCh38
rs1010774893 Chr.2: 42351328-42351328 on Build GRCh38
rs566645191 Chr.2: 42351620-42351620 on Build GRCh38
rs781451266 Chr.2: 42351356-42351356 on Build GRCh38
rs775474123 Chr.2: 42351543-42351543 on Build GRCh38
rs776114196 Chr.2: 42351288-42351288 on Build GRCh38
rs758542580 Chr.2: 42351255-42351255 on Build GRCh38
rs772952010 Chr.2: 42351293-42351293 on Build GRCh38
rs368268918 Chr.2: 42351270-42351270 on Build GRCh38
rs1003273263 Chr.2: 42351636-42351636 on Build GRCh38
rs916171473 Chr.2: 42351446-42351446 on Build GRCh38
rs531317984 Chr.2: 42351514-42351514 on Build GRCh38
rs756543123 Chr.2: 42351368-42351368 on Build GRCh38
rs747252178 Chr.2: 42351256-42351256 on Build GRCh38

View Full Product Details