Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43824129-43824638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG5
Entrez Gene ID
64240
Gene Name
ATP binding cassette subfamily G member 5
Gene Aliases
STSL
UniGene
Hs.132992
Gene Chromosome Location
Chr.2: 43806154-43838840 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
UniGene
Hs.371597
Gene Chromosome Location
Chr.2: 43774039-43828492 on Build GRCh38
Transcripts
NM_022436.2 XM_005264480.3 XM_006712073.3 XM_006712074.3 XM_011533024.2 XM_011533025.2 XM_011533026.2 XM_011533027.2 XM_011533028.2 XM_005264364.4 XM_005264365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658482 c.997G>A p.A333T Chr.2: 43824340-43824340 on Build GRCh38
384930 c.1038A>T p.K346N Chr.2: 43824299-43824299 on Build GRCh38
4583521 c.1049C>G p.T350R Chr.2: 43824288-43824288 on Build GRCh38
1614864 c.1117A>T p.R373W Chr.2: 43824220-43824220 on Build GRCh38
4166877 c.940C>T p.R314W Chr.2: 43824397-43824397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753248302 Chr.2: 43824447-43824447 on Build GRCh38
rs746632134 Chr.2: 43824224-43824224 on Build GRCh38
rs761733742 Chr.2: 43824408-43824408 on Build GRCh38
rs371319986 Chr.2: 43824293-43824293 on Build GRCh38
rs374156612 Chr.2: 43824187-43824187 on Build GRCh38
rs183257361 Chr.2: 43824202-43824202 on Build GRCh38
rs780744160 Chr.2: 43824492-43824492 on Build GRCh38
rs778765045 Chr.2: 43824484-43824484 on Build GRCh38
rs764153556 Chr.2: 43824283-43824283 on Build GRCh38
rs780359213 Chr.2: 43824191-43824191 on Build GRCh38
rs553464142 Chr.2: 43824338-43824338 on Build GRCh38
rs971595317 Chr.2: 43824228-43824228 on Build GRCh38
rs1039775024 Chr.2: 43824489-43824489 on Build GRCh38
rs754395947 Chr.2: 43824341-43824341 on Build GRCh38
rs780811654 Chr.2: 43824457-43824457 on Build GRCh38
rs774304589 Chr.2: 43824404-43824404 on Build GRCh38
rs773296588 Chr.2: 43824422-43824422 on Build GRCh38
rs373926312 Chr.2: 43824417-43824417 on Build GRCh38
rs866998437 Chr.2: 43824529-43824529 on Build GRCh38
rs774717550 Chr.2: 43824419-43824462 on Build GRCh38
rs751766325 Chr.2: 43824179-43824179 on Build GRCh38
rs755828901 Chr.2: 43824472-43824472 on Build GRCh38
rs766912458 Chr.2: 43824333-43824333 on Build GRCh38
rs371609600 Chr.2: 43824206-43824206 on Build GRCh38
rs1006096668 Chr.2: 43824534-43824534 on Build GRCh38
rs1024561998 Chr.2: 43824553-43824553 on Build GRCh38
rs887691711 Chr.2: 43824528-43824528 on Build GRCh38
rs868003691 Chr.2: 43824249-43824249 on Build GRCh38
rs971222247 Chr.2: 43824576-43824576 on Build GRCh38
rs759998693 Chr.2: 43824442-43824442 on Build GRCh38
rs866292096 Chr.2: 43824277-43824277 on Build GRCh38
rs979836358 Chr.2: 43824614-43824614 on Build GRCh38
rs374949576 Chr.2: 43824163-43824163 on Build GRCh38
rs756376137 Chr.2: 43824322-43824322 on Build GRCh38
rs368123646 Chr.2: 43824292-43824292 on Build GRCh38
rs146131711 Chr.2: 43824380-43824380 on Build GRCh38
rs775348848 Chr.2: 43824270-43824270 on Build GRCh38
rs544541406 Chr.2: 43824396-43824396 on Build GRCh38
rs756661544 Chr.2: 43824453-43824453 on Build GRCh38
rs745606275 Chr.2: 43824269-43824269 on Build GRCh38
rs371613832 Chr.2: 43824180-43824180 on Build GRCh38
rs754306032 Chr.2: 43824435-43824435 on Build GRCh38
rs201302611 Chr.2: 43824357-43824357 on Build GRCh38
rs545795625 Chr.2: 43824546-43824546 on Build GRCh38
rs913359583 Chr.2: 43824494-43824494 on Build GRCh38
rs780639375 Chr.2: 43824379-43824379 on Build GRCh38
rs374824368 Chr.2: 43824294-43824294 on Build GRCh38
rs142037828 Chr.2: 43824434-43824435 on Build GRCh38
rs142971825 Chr.2: 43824397-43824397 on Build GRCh38
rs776365033 Chr.2: 43824255-43824255 on Build GRCh38
rs369775776 Chr.2: 43824368-43824368 on Build GRCh38
rs551472825 Chr.2: 43824480-43824480 on Build GRCh38
rs957544603 Chr.2: 43824479-43824479 on Build GRCh38
rs554246754 Chr.2: 43824503-43824503 on Build GRCh38
rs148131742 Chr.2: 43824288-43824288 on Build GRCh38
rs867011983 Chr.2: 43824274-43824274 on Build GRCh38
rs772704001 Chr.2: 43824478-43824481 on Build GRCh38
rs771202498 Chr.2: 43824345-43824347 on Build GRCh38
rs201653460 Chr.2: 43824173-43824173 on Build GRCh38
rs1036177097 Chr.2: 43824542-43824542 on Build GRCh38
rs77969446 Chr.2: 43824482-43824482 on Build GRCh38
rs946118309 Chr.2: 43824510-43824510 on Build GRCh38
rs903868361 Chr.2: 43824493-43824493 on Build GRCh38
rs550689927 Chr.2: 43824192-43824192 on Build GRCh38
rs770535982 Chr.2: 43824227-43824227 on Build GRCh38
rs777402726 Chr.2: 43824363-43824363 on Build GRCh38
rs768533843 Chr.2: 43824401-43824401 on Build GRCh38
rs752658278 Chr.2: 43824165-43824165 on Build GRCh38
rs891741806 Chr.2: 43824525-43824525 on Build GRCh38
rs143740796 Chr.2: 43824423-43824423 on Build GRCh38
rs1014715108 Chr.2: 43824350-43824350 on Build GRCh38
rs867691050 Chr.2: 43824345-43824345 on Build GRCh38
rs765796752 Chr.2: 43824445-43824445 on Build GRCh38
rs750705703 Chr.2: 43824182-43824182 on Build GRCh38
rs1050778822 Chr.2: 43824527-43824527 on Build GRCh38
rs187858673 Chr.2: 43824461-43824461 on Build GRCh38
rs374919194 Chr.2: 43824271-43824271 on Build GRCh38
rs1030996136 Chr.2: 43824515-43824515 on Build GRCh38
rs767648807 Chr.2: 43824415-43824415 on Build GRCh38
rs897605263 Chr.2: 43824591-43824591 on Build GRCh38
rs762934862 Chr.2: 43824272-43824272 on Build GRCh38
rs923284254 Chr.2: 43824517-43824517 on Build GRCh38
rs750663975 Chr.2: 43824312-43824312 on Build GRCh38
rs751087839 Chr.2: 43824287-43824287 on Build GRCh38
rs869025350 Chr.2: 43824359-43824359 on Build GRCh38
rs749813197 Chr.2: 43824552-43824552 on Build GRCh38
rs759493504 Chr.2: 43824155-43824155 on Build GRCh38
rs749191249 Chr.2: 43824398-43824398 on Build GRCh38
rs764172009 Chr.2: 43824178-43824178 on Build GRCh38

View Full Product Details