Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 43871879-43872418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG8
Entrez Gene ID
64241
Gene Name
ATP binding cassette subfamily G member 8
Gene Aliases
GBD4, STSL
UniGene
Hs.413931
Gene Chromosome Location
Chr.2: 43831942-43880051 on Build GRCh38
Transcripts
NM_022437.2 XM_005264483.3 XM_011533029.2 XM_011533030.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542542 c.1118G>T p.C373F Chr.2: 43872129-43872129 on Build GRCh38
4437717 c.1041C>T p.A347A Chr.2: 43872052-43872052 on Build GRCh38
1021061 c.1170G>A p.T390T Chr.2: 43872265-43872265 on Build GRCh38
5973801 c.965-1G>C p.? Chr.2: 43871975-43871975 on Build GRCh38
3372789 c.1017G>A p.R339R Chr.2: 43872028-43872028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111740641 Chr.2: 43872191-43872191 on Build GRCh38
rs778148799 Chr.2: 43872001-43872001 on Build GRCh38
rs776792168 Chr.2: 43872297-43872297 on Build GRCh38
rs139902223 Chr.2: 43872265-43872265 on Build GRCh38
rs780503856 Chr.2: 43872337-43872338 on Build GRCh38
rs950137376 Chr.2: 43872386-43872386 on Build GRCh38
rs765369343 Chr.2: 43871995-43871995 on Build GRCh38
rs369131115 Chr.2: 43872295-43872295 on Build GRCh38
rs148836748 Chr.2: 43872280-43872280 on Build GRCh38
rs762956509 Chr.2: 43872115-43872115 on Build GRCh38
rs746704103 Chr.2: 43872143-43872143 on Build GRCh38
rs768605516 Chr.2: 43872230-43872230 on Build GRCh38
rs548328895 Chr.2: 43872333-43872333 on Build GRCh38
rs771245445 Chr.2: 43872246-43872246 on Build GRCh38
rs1020491963 Chr.2: 43872216-43872216 on Build GRCh38
rs892978337 Chr.2: 43872171-43872171 on Build GRCh38
rs150654176 Chr.2: 43872255-43872255 on Build GRCh38
rs574933503 Chr.2: 43872148-43872148 on Build GRCh38
rs981127727 Chr.2: 43872008-43872008 on Build GRCh38
rs774941972 Chr.2: 43871961-43871961 on Build GRCh38
rs765841884 Chr.2: 43872257-43872257 on Build GRCh38
rs770886402 Chr.2: 43872098-43872098 on Build GRCh38
rs773369936 Chr.2: 43872242-43872242 on Build GRCh38
rs748783589 Chr.2: 43872020-43872020 on Build GRCh38
rs1041007620 Chr.2: 43872122-43872122 on Build GRCh38
rs575261912 Chr.2: 43872121-43872121 on Build GRCh38
rs747887630 Chr.2: 43872023-43872023 on Build GRCh38
rs775349904 Chr.2: 43872164-43872164 on Build GRCh38
rs568651603 Chr.2: 43872256-43872256 on Build GRCh38
rs115207336 Chr.2: 43872249-43872249 on Build GRCh38
rs142346631 Chr.2: 43872072-43872072 on Build GRCh38
rs55741639 Chr.2: 43871918-43871918 on Build GRCh38
rs771449085 Chr.2: 43871955-43871955 on Build GRCh38
rs758869767 Chr.2: 43872217-43872218 on Build GRCh38
rs761465379 Chr.2: 43872264-43872264 on Build GRCh38
rs532711934 Chr.2: 43872193-43872193 on Build GRCh38
rs1056029960 Chr.2: 43872040-43872040 on Build GRCh38
rs568899040 Chr.2: 43872313-43872313 on Build GRCh38
rs957176669 Chr.2: 43871975-43871975 on Build GRCh38
rs750047912 Chr.2: 43872208-43872210 on Build GRCh38
rs144200355 Chr.2: 43872296-43872296 on Build GRCh38
rs781662653 Chr.2: 43872071-43872071 on Build GRCh38
rs115515507 Chr.2: 43872348-43872348 on Build GRCh38
rs755929853 Chr.2: 43872226-43872226 on Build GRCh38
rs376947530 Chr.2: 43872199-43872199 on Build GRCh38
rs775572172 Chr.2: 43872114-43872114 on Build GRCh38
rs763648421 Chr.2: 43872197-43872197 on Build GRCh38
rs373153710 Chr.2: 43872194-43872194 on Build GRCh38
rs781004722 Chr.2: 43872219-43872219 on Build GRCh38
rs557394376 Chr.2: 43872374-43872377 on Build GRCh38
rs371876826 Chr.2: 43872168-43872168 on Build GRCh38
rs1041083656 Chr.2: 43872301-43872301 on Build GRCh38
rs769649113 Chr.2: 43872110-43872110 on Build GRCh38
rs202030766 Chr.2: 43871988-43871988 on Build GRCh38
rs145992603 Chr.2: 43872281-43872281 on Build GRCh38
rs369031361 Chr.2: 43872175-43872175 on Build GRCh38
rs371968769 Chr.2: 43872136-43872136 on Build GRCh38
rs886643200 Chr.2: 43871900-43871900 on Build GRCh38
rs928894885 Chr.2: 43872015-43872015 on Build GRCh38
rs760207833 Chr.2: 43872337-43872337 on Build GRCh38
rs770976468 Chr.2: 43872044-43872044 on Build GRCh38
rs764228571 Chr.2: 43872069-43872069 on Build GRCh38
rs546556227 Chr.2: 43872223-43872223 on Build GRCh38
rs1028794575 Chr.2: 43871973-43871973 on Build GRCh38
rs377097108 Chr.2: 43872152-43872152 on Build GRCh38
rs773022612 Chr.2: 43872029-43872029 on Build GRCh38
rs886056033 Chr.2: 43872298-43872298 on Build GRCh38
rs531331182 Chr.2: 43871901-43871901 on Build GRCh38
rs769713721 Chr.2: 43872163-43872163 on Build GRCh38
rs144221914 Chr.2: 43872074-43872074 on Build GRCh38
rs537925205 Chr.2: 43871917-43871917 on Build GRCh38
rs140778634 Chr.2: 43872105-43872105 on Build GRCh38
rs749233960 Chr.2: 43872229-43872229 on Build GRCh38
rs753700259 Chr.2: 43872279-43872279 on Build GRCh38
rs748219341 Chr.2: 43872287-43872287 on Build GRCh38
rs371617725 Chr.2: 43872260-43872260 on Build GRCh38
rs137852987 Chr.2: 43872094-43872094 on Build GRCh38
rs768280644 Chr.2: 43872021-43872021 on Build GRCh38
rs192112817 Chr.2: 43872274-43872274 on Build GRCh38
rs762135880 Chr.2: 43872119-43872119 on Build GRCh38
rs756992294 Chr.2: 43872146-43872146 on Build GRCh38
rs763192521 Chr.2: 43872063-43872063 on Build GRCh38
rs548297738 Chr.2: 43872243-43872243 on Build GRCh38
rs774622976 Chr.2: 43872118-43872118 on Build GRCh38
rs375807484 Chr.2: 43872028-43872028 on Build GRCh38
rs370167161 Chr.2: 43872052-43872052 on Build GRCh38
rs761976411 Chr.2: 43872172-43872172 on Build GRCh38
rs967135220 Chr.2: 43872251-43872251 on Build GRCh38
rs778440872 Chr.2: 43872079-43872079 on Build GRCh38
rs751866338 Chr.2: 43872070-43872070 on Build GRCh38
rs553899742 Chr.2: 43872059-43872059 on Build GRCh38
rs568259494 Chr.2: 43871979-43871979 on Build GRCh38
rs200436078 Chr.2: 43872235-43872235 on Build GRCh38
rs774111515 Chr.2: 43871974-43871974 on Build GRCh38
rs372660816 Chr.2: 43871985-43871985 on Build GRCh38
rs757910244 Chr.2: 43872138-43872138 on Build GRCh38
rs756901097 Chr.2: 43871949-43871950 on Build GRCh38
rs746791819 Chr.2: 43872095-43872095 on Build GRCh38
rs886056034 Chr.2: 43872305-43872305 on Build GRCh38
rs1019316295 Chr.2: 43871923-43871923 on Build GRCh38
rs4148217 Chr.2: 43872294-43872294 on Build GRCh38
rs554680915 Chr.2: 43872128-43872128 on Build GRCh38
rs759992462 Chr.2: 43872305-43872305 on Build GRCh38
rs893049658 Chr.2: 43872342-43872342 on Build GRCh38
rs756606650 Chr.2: 43872126-43872126 on Build GRCh38
rs982970978 Chr.2: 43872284-43872284 on Build GRCh38
rs757696341 Chr.2: 43872014-43872014 on Build GRCh38
rs761376730 Chr.2: 43872315-43872315 on Build GRCh38
rs773031649 Chr.2: 43872209-43872209 on Build GRCh38
rs772227858 Chr.2: 43872240-43872240 on Build GRCh38
rs996066077 Chr.2: 43872161-43872161 on Build GRCh38
rs755917160 Chr.2: 43872270-43872270 on Build GRCh38
rs778401901 Chr.2: 43872022-43872022 on Build GRCh38
rs748235816 Chr.2: 43872238-43872238 on Build GRCh38
rs758860195 Chr.2: 43871930-43871930 on Build GRCh38
rs768240854 Chr.2: 43871966-43871966 on Build GRCh38
rs201357179 Chr.2: 43872192-43872192 on Build GRCh38
rs752742682 Chr.2: 43872057-43872057 on Build GRCh38
rs1008196282 Chr.2: 43871919-43871919 on Build GRCh38
rs747599796 Chr.2: 43871947-43871947 on Build GRCh38
rs774376327 Chr.2: 43872233-43872233 on Build GRCh38
rs768764678 Chr.2: 43872165-43872165 on Build GRCh38
rs201268110 Chr.2: 43872316-43872316 on Build GRCh38
rs778687007 Chr.2: 43872154-43872155 on Build GRCh38
rs754828544 Chr.2: 43872355-43872355 on Build GRCh38
rs115578276 Chr.2: 43871902-43871902 on Build GRCh38
rs751950437 Chr.2: 43872003-43872003 on Build GRCh38
rs754428212 Chr.2: 43872077-43872077 on Build GRCh38
rs748803910 Chr.2: 43871965-43871965 on Build GRCh38
rs912945197 Chr.2: 43871999-43871999 on Build GRCh38
rs766986994 Chr.2: 43872330-43872330 on Build GRCh38
rs1056572250 Chr.2: 43872147-43872147 on Build GRCh38
rs962289791 Chr.2: 43872366-43872366 on Build GRCh38
rs753003049 Chr.2: 43871998-43871998 on Build GRCh38
rs114404835 Chr.2: 43872104-43872104 on Build GRCh38
rs745663864 Chr.2: 43872225-43872225 on Build GRCh38
rs756902565 Chr.2: 43872200-43872200 on Build GRCh38
rs759654526 Chr.2: 43872047-43872047 on Build GRCh38
rs368297023 Chr.2: 43872157-43872157 on Build GRCh38
rs747645864 Chr.2: 43872091-43872091 on Build GRCh38
rs375507330 Chr.2: 43872017-43872017 on Build GRCh38
rs766062233 Chr.2: 43872345-43872345 on Build GRCh38
rs374156864 Chr.2: 43872275-43872275 on Build GRCh38
rs562447483 Chr.2: 43872227-43872227 on Build GRCh38
rs995990808 Chr.2: 43871964-43871964 on Build GRCh38
rs982420514 Chr.2: 43872336-43872336 on Build GRCh38
rs770410308 Chr.2: 43872306-43872306 on Build GRCh38
rs148185316 Chr.2: 43872117-43872117 on Build GRCh38
rs948186748 Chr.2: 43872154-43872154 on Build GRCh38
rs147360326 Chr.2: 43872125-43872125 on Build GRCh38
rs1008068220 Chr.2: 43872080-43872080 on Build GRCh38
rs778109703 Chr.2: 43871936-43871936 on Build GRCh38
rs145373839 Chr.2: 43872035-43872035 on Build GRCh38
rs758668360 Chr.2: 43872000-43872000 on Build GRCh38
rs773501975 Chr.2: 43872189-43872189 on Build GRCh38

View Full Product Details