Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55870803-55871304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP1
Entrez Gene ID
2202
Gene Name
EGF containing fibulin like extracellular matrix protein 1
Gene Aliases
DHRD, DRAD, FBLN3, FBNL, FIBL-3, MLVT, MTLV, S1-5
UniGene
Hs.732348
Gene Chromosome Location
Chr.2: 55865962-55924163 on Build GRCh38
Transcripts
NM_001039348.2 NM_001039349.2 XM_005264205.4 XM_017003586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021905 c.1136G>A p.C379Y Chr.2: 55870904-55870904 on Build GRCh38
3582356 c.1203A>T p.R401R Chr.2: 55870837-55870837 on Build GRCh38
3582355 c.1205C>T p.S402F Chr.2: 55870835-55870835 on Build GRCh38
4094930 c.1073G>A p.R358H Chr.2: 55871051-55871051 on Build GRCh38
77633 c.1033C>T p.R345W Chr.2: 55871091-55871091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029380408 Chr.2: 55871227-55871227 on Build GRCh38
rs192404906 Chr.2: 55870929-55870929 on Build GRCh38
rs1017206119 Chr.2: 55871238-55871238 on Build GRCh38
rs45535043 Chr.2: 55871137-55871137 on Build GRCh38
rs146446706 Chr.2: 55870880-55870880 on Build GRCh38
rs775080680 Chr.2: 55870838-55870838 on Build GRCh38
rs745343709 Chr.2: 55870933-55870933 on Build GRCh38
rs773568598 Chr.2: 55871150-55871150 on Build GRCh38
rs766866113 Chr.2: 55870882-55870882 on Build GRCh38
rs780590305 Chr.2: 55870931-55870931 on Build GRCh38
rs148823921 Chr.2: 55871005-55871005 on Build GRCh38
rs760845391 Chr.2: 55871017-55871017 on Build GRCh38
rs371175216 Chr.2: 55871142-55871142 on Build GRCh38
rs576071059 Chr.2: 55871088-55871088 on Build GRCh38
rs144184617 Chr.2: 55871232-55871232 on Build GRCh38
rs1012296244 Chr.2: 55870848-55870848 on Build GRCh38
rs184366921 Chr.2: 55871163-55871163 on Build GRCh38
rs747086611 Chr.2: 55870941-55870941 on Build GRCh38
rs772284064 Chr.2: 55871016-55871016 on Build GRCh38
rs765589888 Chr.2: 55871052-55871052 on Build GRCh38
rs777178356 Chr.2: 55871028-55871028 on Build GRCh38
rs755710198 Chr.2: 55871092-55871092 on Build GRCh38
rs771729716 Chr.2: 55871135-55871135 on Build GRCh38
rs776624123 Chr.2: 55871146-55871146 on Build GRCh38
rs769156471 Chr.2: 55870942-55870942 on Build GRCh38
rs779233876 Chr.2: 55870913-55870913 on Build GRCh38
rs781367716 Chr.2: 55870967-55870967 on Build GRCh38
rs868281931 Chr.2: 55871228-55871228 on Build GRCh38
rs767503998 Chr.2: 55870850-55870850 on Build GRCh38
rs574538454 Chr.2: 55871263-55871263 on Build GRCh38
rs962600213 Chr.2: 55871239-55871239 on Build GRCh38
rs762126695 Chr.2: 55870841-55870841 on Build GRCh38
rs754909186 Chr.2: 55871121-55871121 on Build GRCh38
rs756890736 Chr.2: 55871074-55871074 on Build GRCh38
rs768641741 Chr.2: 55870990-55870990 on Build GRCh38
rs367901228 Chr.2: 55870890-55870890 on Build GRCh38
rs374684475 Chr.2: 55870972-55870972 on Build GRCh38
rs145654150 Chr.2: 55871265-55871265 on Build GRCh38
rs1055074015 Chr.2: 55871058-55871058 on Build GRCh38
rs1025522294 Chr.2: 55870934-55870934 on Build GRCh38
rs749245292 Chr.2: 55871120-55871120 on Build GRCh38
rs997820272 Chr.2: 55871198-55871198 on Build GRCh38
rs369049569 Chr.2: 55871039-55871039 on Build GRCh38
rs762143333 Chr.2: 55871006-55871006 on Build GRCh38
rs765694007 Chr.2: 55871161-55871161 on Build GRCh38
rs754310502 Chr.2: 55870885-55870885 on Build GRCh38
rs915936853 Chr.2: 55870862-55870862 on Build GRCh38
rs747183634 Chr.2: 55871139-55871139 on Build GRCh38
rs372748426 Chr.2: 55871051-55871051 on Build GRCh38
rs751279285 Chr.2: 55870920-55870920 on Build GRCh38
rs770059658 Chr.2: 55871144-55871144 on Build GRCh38
rs747909115 Chr.2: 55871130-55871130 on Build GRCh38
rs371636553 Chr.2: 55870944-55870944 on Build GRCh38
rs113138222 Chr.2: 55870999-55870999 on Build GRCh38
rs779708532 Chr.2: 55871108-55871108 on Build GRCh38
rs868466101 Chr.2: 55871002-55871002 on Build GRCh38
rs567386311 Chr.2: 55870830-55870830 on Build GRCh38
rs753001473 Chr.2: 55871053-55871053 on Build GRCh38
rs139142559 Chr.2: 55870829-55870829 on Build GRCh38
rs750022923 Chr.2: 55871090-55871090 on Build GRCh38
rs121434491 Chr.2: 55871091-55871091 on Build GRCh38
rs950047658 Chr.2: 55870876-55870876 on Build GRCh38
rs923282282 Chr.2: 55871136-55871136 on Build GRCh38
rs774225644 Chr.2: 55871004-55871004 on Build GRCh38
rs561554487 Chr.2: 55871249-55871249 on Build GRCh38
rs778868978 Chr.2: 55871126-55871126 on Build GRCh38
rs750455273 Chr.2: 55870875-55870875 on Build GRCh38
rs762613133 Chr.2: 55871003-55871003 on Build GRCh38
rs991475191 Chr.2: 55870834-55870834 on Build GRCh38
rs766343862 Chr.2: 55871018-55871018 on Build GRCh38
rs936807142 Chr.2: 55871203-55871203 on Build GRCh38

View Full Product Details