Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 62706812-62707317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1
Entrez Gene ID
23301
Gene Name
EH domain binding protein 1
Gene Aliases
HPC12, NACSIN
UniGene
Hs.271667
Gene Chromosome Location
Chr.2: 62673851-63046487 on Build GRCh38
Transcripts
NM_001142614.1 NM_001142615.2 NM_001142616.1 NM_015252.3 XM_005264225.2 XM_005264226.2 XM_005264227.1 XM_005264228.2 XM_006711971.2 XM_011532713.2 XM_011532714.2 XM_011532715.2 XM_011532716.2 XM_011532717.1 XM_011532718.2 XM_017003643.1 XM_017003644.1 XM_017003645.1 XM_017003646.1 XM_017003647.1 XM_017003648.1 XM_017003649.1 XM_017003650.1 XM_017003651.1 XM_017003652.1 XM_017003653.1 XM_017003654.1 XM_017003655.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164559 c.104+10A>C p.? Chr.2: 62707305-62707305 on Build GRCh38
308019 c.47C>T p.S16F Chr.2: 62707238-62707238 on Build GRCh38
1668976 c.91T>C p.C31R Chr.2: 62707282-62707282 on Build GRCh38
477554 c.21A>G p.R7R Chr.2: 62707212-62707212 on Build GRCh38
3839804 c.65C>T p.A22V Chr.2: 62707256-62707256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776715825 Chr.2: 62707173-62707173 on Build GRCh38
rs994138176 Chr.2: 62707021-62707021 on Build GRCh38
rs772377878 Chr.2: 62707142-62707142 on Build GRCh38
rs756024050 Chr.2: 62707223-62707223 on Build GRCh38
rs778693154 Chr.2: 62707266-62707266 on Build GRCh38
rs769062629 Chr.2: 62706927-62706927 on Build GRCh38
rs938836452 Chr.2: 62707265-62707265 on Build GRCh38
rs957724297 Chr.2: 62706870-62706870 on Build GRCh38
rs753367206 Chr.2: 62707261-62707261 on Build GRCh38
rs141267895 Chr.2: 62707194-62707194 on Build GRCh38
rs760982227 Chr.2: 62706923-62706923 on Build GRCh38
rs766464893 Chr.2: 62707242-62707242 on Build GRCh38
rs747477985 Chr.2: 62707163-62707163 on Build GRCh38
rs1025881124 Chr.2: 62707029-62707029 on Build GRCh38
rs1042273881 Chr.2: 62706899-62706899 on Build GRCh38
rs907156083 Chr.2: 62707125-62707125 on Build GRCh38
rs1020440423 Chr.2: 62706912-62706912 on Build GRCh38
rs761659610 Chr.2: 62707191-62707191 on Build GRCh38
rs940559825 Chr.2: 62707055-62707055 on Build GRCh38
rs955262765 Chr.2: 62707067-62707067 on Build GRCh38
rs754602388 Chr.2: 62707263-62707263 on Build GRCh38
rs911520462 Chr.2: 62706984-62706984 on Build GRCh38
rs944396505 Chr.2: 62706998-62706998 on Build GRCh38
rs745782595 Chr.2: 62707166-62707167 on Build GRCh38
rs946306777 Chr.2: 62706896-62706896 on Build GRCh38
rs886290768 Chr.2: 62707053-62707053 on Build GRCh38
rs775758047 Chr.2: 62707189-62707189 on Build GRCh38
rs113042275 Chr.2: 62707193-62707193 on Build GRCh38
rs201188594 Chr.2: 62707183-62707183 on Build GRCh38
rs375031846 Chr.2: 62707171-62707171 on Build GRCh38
rs990268400 Chr.2: 62706903-62706903 on Build GRCh38
rs930415164 Chr.2: 62707030-62707030 on Build GRCh38
rs1048906489 Chr.2: 62707047-62707047 on Build GRCh38
rs1006203139 Chr.2: 62706969-62706969 on Build GRCh38
rs548583605 Chr.2: 62706845-62706845 on Build GRCh38
rs1034796338 Chr.2: 62707156-62707156 on Build GRCh38
rs989738412 Chr.2: 62707182-62707182 on Build GRCh38
rs369378909 Chr.2: 62707200-62707200 on Build GRCh38
rs550453529 Chr.2: 62707117-62707117 on Build GRCh38
rs894578321 Chr.2: 62706953-62706953 on Build GRCh38
rs1017953775 Chr.2: 62707132-62707132 on Build GRCh38
rs773711991 Chr.2: 62707143-62707143 on Build GRCh38
rs778031853 Chr.2: 62707289-62707289 on Build GRCh38
rs747501545 Chr.2: 62707293-62707293 on Build GRCh38
rs370041943 Chr.2: 62707176-62707176 on Build GRCh38
rs1055913878 Chr.2: 62706928-62706928 on Build GRCh38
rs747767778 Chr.2: 62707281-62707281 on Build GRCh38
rs373652898 Chr.2: 62706989-62706989 on Build GRCh38
rs914785219 Chr.2: 62706889-62706889 on Build GRCh38
rs986979725 Chr.2: 62707274-62707274 on Build GRCh38
rs1045685851 Chr.2: 62707107-62707107 on Build GRCh38
rs371675047 Chr.2: 62706948-62706948 on Build GRCh38
rs776976870 Chr.2: 62706978-62706978 on Build GRCh38
rs1008489914 Chr.2: 62707068-62707068 on Build GRCh38
rs367901222 Chr.2: 62706852-62706852 on Build GRCh38
rs149177405 Chr.2: 62707286-62707286 on Build GRCh38

View Full Product Details