Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 64568974-64569475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
UniGene
Hs.655167
Gene Chromosome Location
Chr.2: 64524305-64593005 on Build GRCh38
Transcripts
NM_001002243.2 NM_017657.4 NM_203437.3 XM_005264380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182176 c.2207G>A p.R736Q Chr.2: 64569211-64569211 on Build GRCh38
4645468 c.2108C>T p.T703I Chr.2: 64569112-64569112 on Build GRCh38
1022244 c.2147A>G p.Y716C Chr.2: 64569151-64569151 on Build GRCh38
1182177 c.2207G>A p.R736Q Chr.2: 64569211-64569211 on Build GRCh38
4660260 c.2153G>A p.W718* Chr.2: 64569157-64569157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032229863 Chr.2: 64569340-64569340 on Build GRCh38
rs939896911 Chr.2: 64569046-64569046 on Build GRCh38
rs983738525 Chr.2: 64569387-64569387 on Build GRCh38
rs769860662 Chr.2: 64569217-64569217 on Build GRCh38
rs369689397 Chr.2: 64569130-64569130 on Build GRCh38
rs953747741 Chr.2: 64569359-64569359 on Build GRCh38
rs1045617028 Chr.2: 64569037-64569037 on Build GRCh38
rs574942779 Chr.2: 64569015-64569015 on Build GRCh38
rs754391594 Chr.2: 64569116-64569116 on Build GRCh38
rs747798962 Chr.2: 64569131-64569131 on Build GRCh38
rs777224794 Chr.2: 64569050-64569050 on Build GRCh38
rs6546087 Chr.2: 64569345-64569345 on Build GRCh38
rs936089634 Chr.2: 64569378-64569378 on Build GRCh38
rs746444763 Chr.2: 64569191-64569191 on Build GRCh38
rs1014600956 Chr.2: 64569086-64569086 on Build GRCh38
rs112260349 Chr.2: 64569448-64569448 on Build GRCh38
rs557314408 Chr.2: 64569067-64569067 on Build GRCh38
rs554714646 Chr.2: 64569000-64569000 on Build GRCh38
rs546012151 Chr.2: 64569423-64569423 on Build GRCh38
rs181627667 Chr.2: 64569067-64569067 on Build GRCh38
rs540740913 Chr.2: 64569036-64569036 on Build GRCh38
rs774584290 Chr.2: 64569251-64569251 on Build GRCh38
rs1037254232 Chr.2: 64569048-64569048 on Build GRCh38
rs200541567 Chr.2: 64569099-64569099 on Build GRCh38
rs950042014 Chr.2: 64569442-64569442 on Build GRCh38
rs977033134 Chr.2: 64569103-64569103 on Build GRCh38
rs764249417 Chr.2: 64569248-64569248 on Build GRCh38
rs763135763 Chr.2: 64569246-64569246 on Build GRCh38
rs999053468 Chr.2: 64569278-64569278 on Build GRCh38
rs771493374 Chr.2: 64569352-64569352 on Build GRCh38
rs758165281 Chr.2: 64569160-64569160 on Build GRCh38
rs778261896 Chr.2: 64569161-64569161 on Build GRCh38
rs776943966 Chr.2: 64569183-64569183 on Build GRCh38
rs868261565 Chr.2: 64569208-64569208 on Build GRCh38
rs186702663 Chr.2: 64569069-64569069 on Build GRCh38
rs926109971 Chr.2: 64569356-64569356 on Build GRCh38
rs1045648284 Chr.2: 64569447-64569447 on Build GRCh38
rs977369399 Chr.2: 64569349-64569349 on Build GRCh38
rs376674777 Chr.2: 64569176-64569176 on Build GRCh38
rs755957936 Chr.2: 64569098-64569098 on Build GRCh38
rs373938301 Chr.2: 64569140-64569140 on Build GRCh38
rs191234922 Chr.2: 64569288-64569288 on Build GRCh38
rs747289920 Chr.2: 64569162-64569162 on Build GRCh38
rs750264271 Chr.2: 64569078-64569078 on Build GRCh38
rs768199746 Chr.2: 64569014-64569014 on Build GRCh38
rs759465754 Chr.2: 64569054-64569054 on Build GRCh38
rs775583842 Chr.2: 64569245-64569245 on Build GRCh38
rs760137744 Chr.2: 64569430-64569430 on Build GRCh38
rs905733575 Chr.2: 64569065-64569065 on Build GRCh38
rs76658175 Chr.2: 64569076-64569076 on Build GRCh38
rs760322292 Chr.2: 64569256-64569256 on Build GRCh38
rs1053208627 Chr.2: 64569398-64569398 on Build GRCh38
rs766213691 Chr.2: 64569269-64569269 on Build GRCh38
rs753821744 Chr.2: 64569107-64569107 on Build GRCh38
rs909627429 Chr.2: 64569024-64569024 on Build GRCh38

View Full Product Details