Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69320314-69320827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT1
Entrez Gene ID
2673
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
UniGene
Hs.580300
Gene Chromosome Location
Chr.2: 69319769-69387254 on Build GRCh38
Transcripts
NM_001244710.1 NM_002056.3

SNP Detail

SNP ID Chromosome Location
rs992202446 Chr.2: 69320555-69320555 on Build GRCh38
rs963871918 Chr.2: 69320398-69320398 on Build GRCh38
rs940728850 Chr.2: 69320799-69320799 on Build GRCh38
rs1033824657 Chr.2: 69320515-69320515 on Build GRCh38
rs879750913 Chr.2: 69320788-69320788 on Build GRCh38
rs537761645 Chr.2: 69320413-69320413 on Build GRCh38
rs1001738837 Chr.2: 69320434-69320434 on Build GRCh38
rs917593121 Chr.2: 69320591-69320591 on Build GRCh38
rs111295641 Chr.2: 69320783-69320783 on Build GRCh38
rs1051676273 Chr.2: 69320741-69320741 on Build GRCh38
rs1044785 Chr.2: 69320392-69320392 on Build GRCh38
rs200306923 Chr.2: 69320802-69320802 on Build GRCh38
rs183623225 Chr.2: 69320702-69320702 on Build GRCh38
rs1044784 Chr.2: 69320426-69320426 on Build GRCh38
rs886056233 Chr.2: 69320757-69320757 on Build GRCh38
rs1039232128 Chr.2: 69320780-69320780 on Build GRCh38
rs758312130 Chr.2: 69320647-69320648 on Build GRCh38
rs886056234 Chr.2: 69320787-69320788 on Build GRCh38
rs921980900 Chr.2: 69320713-69320713 on Build GRCh38
rs936485410 Chr.2: 69320689-69320689 on Build GRCh38
rs886056232 Chr.2: 69320675-69320675 on Build GRCh38
rs565495497 Chr.2: 69320796-69320796 on Build GRCh38
rs1038882218 Chr.2: 69320710-69320710 on Build GRCh38
rs189850080 Chr.2: 69320707-69320707 on Build GRCh38
rs115361740 Chr.2: 69320725-69320725 on Build GRCh38
rs925222543 Chr.2: 69320667-69320667 on Build GRCh38
rs187793362 Chr.2: 69320427-69320427 on Build GRCh38
rs1016508768 Chr.2: 69320351-69320351 on Build GRCh38
rs966407746 Chr.2: 69320658-69320658 on Build GRCh38
rs568780403 Chr.2: 69320409-69320409 on Build GRCh38
rs533594691 Chr.2: 69320559-69320559 on Build GRCh38
rs773373681 Chr.2: 69320710-69320710 on Build GRCh38
rs767679164 Chr.2: 69320703-69320703 on Build GRCh38
rs191479634 Chr.2: 69320516-69320516 on Build GRCh38
rs959165367 Chr.2: 69320524-69320524 on Build GRCh38
rs886438707 Chr.2: 69320767-69320767 on Build GRCh38
rs900165415 Chr.2: 69320787-69320787 on Build GRCh38
rs977997400 Chr.2: 69320664-69320664 on Build GRCh38

View Full Product Details