Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69458300-69458795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
UniGene
Hs.468878
Gene Chromosome Location
Chr.2: 69457995-69643845 on Build GRCh38
Transcripts
NM_014911.3

SNP Detail

SNP ID Chromosome Location
rs928033670 Chr.2: 69458602-69458602 on Build GRCh38
rs951680820 Chr.2: 69458574-69458574 on Build GRCh38
rs933627929 Chr.2: 69458760-69458760 on Build GRCh38
rs982116823 Chr.2: 69458567-69458567 on Build GRCh38
rs900126539 Chr.2: 69458745-69458745 on Build GRCh38
rs552901088 Chr.2: 69458666-69458666 on Build GRCh38
rs752699617 Chr.2: 69458488-69458488 on Build GRCh38
rs999080219 Chr.2: 69458474-69458474 on Build GRCh38
rs766524955 Chr.2: 69458482-69458482 on Build GRCh38
rs1044161196 Chr.2: 69458404-69458404 on Build GRCh38
rs957448272 Chr.2: 69458499-69458499 on Build GRCh38
rs1044112340 Chr.2: 69458707-69458707 on Build GRCh38
rs1048999639 Chr.2: 69458324-69458324 on Build GRCh38
rs570640488 Chr.2: 69458711-69458711 on Build GRCh38
rs968614824 Chr.2: 69458471-69458471 on Build GRCh38
rs548660535 Chr.2: 69458465-69458465 on Build GRCh38
rs910036786 Chr.2: 69458619-69458619 on Build GRCh38
rs1039973613 Chr.2: 69458727-69458727 on Build GRCh38
rs1035374086 Chr.2: 69458442-69458442 on Build GRCh38
rs572797665 Chr.2: 69458680-69458680 on Build GRCh38
rs561879564 Chr.2: 69458551-69458551 on Build GRCh38
rs1047257162 Chr.2: 69458321-69458321 on Build GRCh38
rs773388817 Chr.2: 69458449-69458449 on Build GRCh38
rs997887909 Chr.2: 69458447-69458447 on Build GRCh38
rs765579799 Chr.2: 69458552-69458552 on Build GRCh38
rs1039982344 Chr.2: 69458415-69458415 on Build GRCh38
rs930048616 Chr.2: 69458658-69458658 on Build GRCh38
rs888627790 Chr.2: 69458378-69458378 on Build GRCh38
rs899467189 Chr.2: 69458420-69458420 on Build GRCh38
rs918615605 Chr.2: 69458572-69458573 on Build GRCh38
rs181724194 Chr.2: 69458622-69458622 on Build GRCh38
rs1057162998 Chr.2: 69458728-69458729 on Build GRCh38
rs550880792 Chr.2: 69458709-69458709 on Build GRCh38
rs1010561817 Chr.2: 69458765-69458766 on Build GRCh38
rs769173300 Chr.2: 69458576-69458577 on Build GRCh38
rs759432290 Chr.2: 69458709-69458710 on Build GRCh38
rs139723881 Chr.2: 69458707-69458708 on Build GRCh38
rs888626551 Chr.2: 69458323-69458323 on Build GRCh38
rs537042229 Chr.2: 69458393-69458393 on Build GRCh38
rs891470188 Chr.2: 69458729-69458729 on Build GRCh38
rs991380145 Chr.2: 69458523-69458523 on Build GRCh38
rs530694648 Chr.2: 69458681-69458681 on Build GRCh38
rs766817389 Chr.2: 69458438-69458438 on Build GRCh38
rs536412862 Chr.2: 69458623-69458623 on Build GRCh38
rs796361957 Chr.2: 69458708-69458709 on Build GRCh38
rs1002522823 Chr.2: 69458432-69458432 on Build GRCh38
rs1050779831 Chr.2: 69458765-69458765 on Build GRCh38
rs969352217 Chr.2: 69458553-69458553 on Build GRCh38

View Full Product Details