Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69464710-69465204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
UniGene
Hs.468878
Gene Chromosome Location
Chr.2: 69457995-69643845 on Build GRCh38
Transcripts
NM_014911.3

SNP Detail

SNP ID Chromosome Location
rs920892764 Chr.2: 69464813-69464813 on Build GRCh38
rs928596986 Chr.2: 69465163-69465163 on Build GRCh38
rs1042912064 Chr.2: 69464825-69464825 on Build GRCh38
rs1047069652 Chr.2: 69465146-69465146 on Build GRCh38
rs73934946 Chr.2: 69465124-69465124 on Build GRCh38
rs190503839 Chr.2: 69464809-69464809 on Build GRCh38
rs941795756 Chr.2: 69464770-69464770 on Build GRCh38
rs1044328341 Chr.2: 69464901-69464901 on Build GRCh38
rs924389984 Chr.2: 69464834-69464834 on Build GRCh38
rs955196840 Chr.2: 69464992-69464992 on Build GRCh38
rs904581435 Chr.2: 69464865-69464865 on Build GRCh38
rs557113083 Chr.2: 69465153-69465153 on Build GRCh38
rs778294307 Chr.2: 69464906-69464906 on Build GRCh38
rs887333390 Chr.2: 69465156-69465156 on Build GRCh38
rs867501516 Chr.2: 69464876-69464876 on Build GRCh38
rs73934947 Chr.2: 69465125-69465125 on Build GRCh38
rs986513874 Chr.2: 69465011-69465011 on Build GRCh38
rs779524330 Chr.2: 69464821-69464821 on Build GRCh38
rs772545161 Chr.2: 69464879-69464879 on Build GRCh38
rs962929641 Chr.2: 69464798-69464798 on Build GRCh38
rs549723688 Chr.2: 69464958-69464958 on Build GRCh38
rs762598542 Chr.2: 69465077-69465077 on Build GRCh38
rs948605696 Chr.2: 69464893-69464893 on Build GRCh38
rs564951590 Chr.2: 69464790-69464790 on Build GRCh38
rs934484416 Chr.2: 69464848-69464848 on Build GRCh38
rs997497176 Chr.2: 69464977-69464977 on Build GRCh38
rs78946376 Chr.2: 69465034-69465034 on Build GRCh38
rs748598083 Chr.2: 69464878-69464878 on Build GRCh38
rs998387207 Chr.2: 69464918-69464918 on Build GRCh38
rs933708038 Chr.2: 69464816-69464816 on Build GRCh38
rs992501999 Chr.2: 69464985-69464985 on Build GRCh38
rs771323739 Chr.2: 69465014-69465014 on Build GRCh38
rs1007397449 Chr.2: 69464867-69464875 on Build GRCh38
rs528458797 Chr.2: 69465035-69465035 on Build GRCh38
rs887161566 Chr.2: 69464924-69464924 on Build GRCh38
rs1042881331 Chr.2: 69465042-69465042 on Build GRCh38
rs925806964 Chr.2: 69465083-69465085 on Build GRCh38
rs951505981 Chr.2: 69464760-69464760 on Build GRCh38
rs143501020 Chr.2: 69465118-69465118 on Build GRCh38
rs747309100 Chr.2: 69464913-69464913 on Build GRCh38
rs960008867 Chr.2: 69464984-69464984 on Build GRCh38
rs183659203 Chr.2: 69464925-69464925 on Build GRCh38
rs1035150332 Chr.2: 69464753-69464753 on Build GRCh38
rs998104975 Chr.2: 69465140-69465140 on Build GRCh38
rs775231630 Chr.2: 69465037-69465037 on Build GRCh38
rs938593795 Chr.2: 69464793-69464793 on Build GRCh38
rs887107469 Chr.2: 69465175-69465175 on Build GRCh38
rs962917318 Chr.2: 69464971-69464971 on Build GRCh38
rs1005654444 Chr.2: 69464935-69464935 on Build GRCh38
rs531528488 Chr.2: 69464980-69464980 on Build GRCh38
rs544627076 Chr.2: 69464778-69464778 on Build GRCh38
rs1017072543 Chr.2: 69464952-69464952 on Build GRCh38
rs979763802 Chr.2: 69464777-69464777 on Build GRCh38
rs1056455479 Chr.2: 69465172-69465172 on Build GRCh38
rs1017533696 Chr.2: 69464944-69464944 on Build GRCh38
rs530523695 Chr.2: 69464808-69464808 on Build GRCh38
rs529593288 Chr.2: 69464933-69464933 on Build GRCh38
rs1003568751 Chr.2: 69464908-69464908 on Build GRCh38
rs1039692397 Chr.2: 69464875-69464875 on Build GRCh38
rs73934953 Chr.2: 69465164-69465164 on Build GRCh38
rs1054190440 Chr.2: 69464871-69464871 on Build GRCh38
rs963148386 Chr.2: 69464957-69464957 on Build GRCh38
rs908928112 Chr.2: 69464756-69464756 on Build GRCh38
rs982977183 Chr.2: 69464761-69464761 on Build GRCh38
rs1044685516 Chr.2: 69465148-69465148 on Build GRCh38

View Full Product Details