Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69465496-69465996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
UniGene
Hs.468878
Gene Chromosome Location
Chr.2: 69457995-69643845 on Build GRCh38
Transcripts
NM_014911.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506788 c.721C>G p.H241D Chr.2: 69465772-69465772 on Build GRCh38
5416311 c.892G>C p.E298Q Chr.2: 69465601-69465601 on Build GRCh38
5572999 c.969T>A p.S323S Chr.2: 69465524-69465524 on Build GRCh38
7270193 c.527T>C p.M176T Chr.2: 69465966-69465966 on Build GRCh38
6991953 c.773C>G p.S258C Chr.2: 69465720-69465720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939991654 Chr.2: 69465716-69465716 on Build GRCh38
rs574593797 Chr.2: 69465914-69465914 on Build GRCh38
rs528799890 Chr.2: 69465595-69465595 on Build GRCh38
rs941613413 Chr.2: 69465601-69465601 on Build GRCh38
rs969810139 Chr.2: 69465635-69465635 on Build GRCh38
rs373774938 Chr.2: 69465621-69465621 on Build GRCh38
rs184211617 Chr.2: 69465887-69465887 on Build GRCh38
rs754413422 Chr.2: 69465885-69465885 on Build GRCh38
rs934059631 Chr.2: 69465823-69465823 on Build GRCh38
rs1033306031 Chr.2: 69465794-69465794 on Build GRCh38
rs565174367 Chr.2: 69465616-69465616 on Build GRCh38
rs754038893 Chr.2: 69465840-69465840 on Build GRCh38
rs376375156 Chr.2: 69465809-69465809 on Build GRCh38
rs536007779 Chr.2: 69465741-69465741 on Build GRCh38
rs773847459 Chr.2: 69465869-69465869 on Build GRCh38
rs377251388 Chr.2: 69465739-69465739 on Build GRCh38
rs907817910 Chr.2: 69465684-69465684 on Build GRCh38
rs79314499 Chr.2: 69465944-69465944 on Build GRCh38
rs554669966 Chr.2: 69465912-69465912 on Build GRCh38
rs147076921 Chr.2: 69465877-69465877 on Build GRCh38
rs749159269 Chr.2: 69465797-69465797 on Build GRCh38
rs975624657 Chr.2: 69465801-69465801 on Build GRCh38
rs1008401254 Chr.2: 69465588-69465588 on Build GRCh38
rs982870062 Chr.2: 69465831-69465831 on Build GRCh38
rs748436986 Chr.2: 69465712-69465712 on Build GRCh38
rs758706052 Chr.2: 69465947-69465947 on Build GRCh38
rs374273327 Chr.2: 69465788-69465788 on Build GRCh38
rs200551979 Chr.2: 69465602-69465602 on Build GRCh38
rs9309428 Chr.2: 69465874-69465874 on Build GRCh38
rs1020078081 Chr.2: 69465907-69465907 on Build GRCh38
rs941332305 Chr.2: 69465863-69465863 on Build GRCh38
rs891460261 Chr.2: 69465615-69465615 on Build GRCh38
rs759642384 Chr.2: 69465953-69465955 on Build GRCh38
rs1035493131 Chr.2: 69465649-69465649 on Build GRCh38
rs1014780579 Chr.2: 69465943-69465943 on Build GRCh38
rs1014039902 Chr.2: 69465759-69465759 on Build GRCh38
rs959840565 Chr.2: 69465902-69465902 on Build GRCh38
rs1035885515 Chr.2: 69465898-69465898 on Build GRCh38
rs983313878 Chr.2: 69465926-69465926 on Build GRCh38
rs140524324 Chr.2: 69465775-69465775 on Build GRCh38
rs376431257 Chr.2: 69465870-69465870 on Build GRCh38
rs1049900569 Chr.2: 69465553-69465553 on Build GRCh38
rs760102968 Chr.2: 69465913-69465913 on Build GRCh38
rs1000248284 Chr.2: 69465948-69465948 on Build GRCh38
rs868601883 Chr.2: 69465591-69465591 on Build GRCh38
rs891292330 Chr.2: 69465575-69465575 on Build GRCh38
rs375748068 Chr.2: 69465648-69465648 on Build GRCh38
rs964015097 Chr.2: 69465798-69465800 on Build GRCh38
rs766626115 Chr.2: 69465878-69465878 on Build GRCh38
rs201931021 Chr.2: 69465969-69465969 on Build GRCh38
rs1050279595 Chr.2: 69465803-69465803 on Build GRCh38
rs371344053 Chr.2: 69465643-69465643 on Build GRCh38
rs1019942178 Chr.2: 69465760-69465760 on Build GRCh38
rs368873583 Chr.2: 69465669-69465669 on Build GRCh38
rs540025469 Chr.2: 69465965-69465965 on Build GRCh38
rs944249924 Chr.2: 69465854-69465854 on Build GRCh38
rs191294987 Chr.2: 69465608-69465608 on Build GRCh38
rs938288632 Chr.2: 69465871-69465871 on Build GRCh38
rs531021526 Chr.2: 69465622-69465622 on Build GRCh38
rs997355275 Chr.2: 69465528-69465528 on Build GRCh38
rs898610298 Chr.2: 69465527-69465527 on Build GRCh38
rs755145042 Chr.2: 69465627-69465627 on Build GRCh38
rs983362975 Chr.2: 69465658-69465658 on Build GRCh38
rs773474226 Chr.2: 69465876-69465876 on Build GRCh38
rs905532138 Chr.2: 69465881-69465881 on Build GRCh38
rs781137571 Chr.2: 69465752-69465752 on Build GRCh38
rs148323049 Chr.2: 69465650-69465650 on Build GRCh38
rs1032694543 Chr.2: 69465954-69465954 on Build GRCh38
rs974042708 Chr.2: 69465765-69465765 on Build GRCh38
rs1014008646 Chr.2: 69465890-69465890 on Build GRCh38
rs769641063 Chr.2: 69465785-69465785 on Build GRCh38
rs895537968 Chr.2: 69465686-69465686 on Build GRCh38

View Full Product Details