Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70165004-70165516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
UniGene
Hs.720049
Gene Chromosome Location
Chr.2: 70149885-70191071 on Build GRCh38
Transcripts
NM_017880.1 XM_005264389.2 XM_005264390.2 XM_005264391.2 XM_011532937.2 XM_017004398.1 XM_017004399.1 XM_017004400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1306954 c.1351G>A p.E451K Chr.2: 70165094-70165094 on Build GRCh38
5862763 c.1346C>T p.T449I Chr.2: 70165099-70165099 on Build GRCh38
237079 c.1253-8C>T p.? Chr.2: 70165200-70165200 on Build GRCh38
1306955 c.1283G>C p.G428A Chr.2: 70165162-70165162 on Build GRCh38
7172190 c.1262G>A p.R421Q Chr.2: 70165183-70165183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045081624 Chr.2: 70165383-70165383 on Build GRCh38
rs1053085242 Chr.2: 70165374-70165374 on Build GRCh38
rs946674601 Chr.2: 70165380-70165380 on Build GRCh38
rs532641604 Chr.2: 70165398-70165398 on Build GRCh38
rs1008910223 Chr.2: 70165423-70165423 on Build GRCh38
rs757053964 Chr.2: 70165322-70165322 on Build GRCh38
rs751847621 Chr.2: 70165061-70165061 on Build GRCh38
rs766608711 Chr.2: 70165172-70165172 on Build GRCh38
rs975332651 Chr.2: 70165444-70165444 on Build GRCh38
rs770566016 Chr.2: 70165164-70165164 on Build GRCh38
rs767826237 Chr.2: 70165483-70165483 on Build GRCh38
rs764329859 Chr.2: 70165221-70165221 on Build GRCh38
rs149872836 Chr.2: 70165490-70165490 on Build GRCh38
rs990816906 Chr.2: 70165073-70165073 on Build GRCh38
rs764225817 Chr.2: 70165054-70165054 on Build GRCh38
rs200955734 Chr.2: 70165081-70165081 on Build GRCh38
rs905233275 Chr.2: 70165387-70165387 on Build GRCh38
rs752866309 Chr.2: 70165046-70165046 on Build GRCh38
rs894062020 Chr.2: 70165379-70165379 on Build GRCh38
rs531927154 Chr.2: 70165491-70165491 on Build GRCh38
rs546538568 Chr.2: 70165199-70165199 on Build GRCh38
rs962664121 Chr.2: 70165441-70165441 on Build GRCh38
rs975049841 Chr.2: 70165245-70165245 on Build GRCh38
rs780796984 Chr.2: 70165415-70165415 on Build GRCh38
rs779190163 Chr.2: 70165033-70165033 on Build GRCh38
rs1003612618 Chr.2: 70165394-70165394 on Build GRCh38
rs933732248 Chr.2: 70165248-70165248 on Build GRCh38
rs777205183 Chr.2: 70165144-70165144 on Build GRCh38
rs758427783 Chr.2: 70165049-70165049 on Build GRCh38
rs774540688 Chr.2: 70165213-70165213 on Build GRCh38
rs567442826 Chr.2: 70165167-70165167 on Build GRCh38
rs770142151 Chr.2: 70165179-70165179 on Build GRCh38
rs1017222442 Chr.2: 70165430-70165430 on Build GRCh38
rs543780485 Chr.2: 70165301-70165301 on Build GRCh38
rs1004066125 Chr.2: 70165429-70165429 on Build GRCh38
rs778738862 Chr.2: 70165089-70165089 on Build GRCh38
rs757541790 Chr.2: 70165063-70165063 on Build GRCh38
rs748080155 Chr.2: 70165103-70165103 on Build GRCh38
rs771346298 Chr.2: 70165203-70165203 on Build GRCh38
rs746719463 Chr.2: 70165159-70165159 on Build GRCh38
rs563756756 Chr.2: 70165326-70165326 on Build GRCh38
rs753548446 Chr.2: 70165078-70165078 on Build GRCh38
rs11126263 Chr.2: 70165247-70165247 on Build GRCh38
rs986552560 Chr.2: 70165290-70165290 on Build GRCh38
rs763255504 Chr.2: 70165057-70165058 on Build GRCh38
rs962667346 Chr.2: 70165149-70165149 on Build GRCh38
rs1049430304 Chr.2: 70165408-70165408 on Build GRCh38
rs74660649 Chr.2: 70165200-70165200 on Build GRCh38
rs555438555 Chr.2: 70165183-70165183 on Build GRCh38
rs140651209 Chr.2: 70165071-70165071 on Build GRCh38
rs1027942350 Chr.2: 70165479-70165479 on Build GRCh38
rs377348055 Chr.2: 70165242-70165242 on Build GRCh38
rs890021591 Chr.2: 70165413-70165413 on Build GRCh38
rs947026569 Chr.2: 70165407-70165407 on Build GRCh38
rs751690032 Chr.2: 70165231-70165231 on Build GRCh38
rs892930721 Chr.2: 70165384-70165384 on Build GRCh38
rs1019894986 Chr.2: 70165445-70165445 on Build GRCh38
rs200219021 Chr.2: 70165486-70165486 on Build GRCh38
rs540609698 Chr.2: 70165293-70165293 on Build GRCh38

View Full Product Details