Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70181076-70181583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
-
UniGene
Hs.720049
Gene Chromosome Location
Chr.2: 70149885-70191071 on Build GRCh38
Transcripts
NM_017880.1 XM_005264389.2 XM_005264390.2 XM_005264391.2 XM_011532937.2 XM_017004398.1 XM_017004399.1 XM_017004400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095331 c.403C>T p.H135Y Chr.2: 70181583-70181583 on Build GRCh38
5623907 c.659G>T p.R220L Chr.2: 70181327-70181327 on Build GRCh38
4095329 c.662G>A p.R221H Chr.2: 70181324-70181324 on Build GRCh38
4095330 c.641G>T p.G214V Chr.2: 70181345-70181345 on Build GRCh38
3056386 c.665T>G p.F222C Chr.2: 70181321-70181321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777996451 Chr.2: 70181293-70181305 on Build GRCh38
rs376164589 Chr.2: 70181498-70181498 on Build GRCh38
rs768064949 Chr.2: 70181372-70181372 on Build GRCh38
rs765290214 Chr.2: 70181325-70181325 on Build GRCh38
rs779889295 Chr.2: 70181366-70181366 on Build GRCh38
rs992669437 Chr.2: 70181163-70181163 on Build GRCh38
rs749184211 Chr.2: 70181367-70181367 on Build GRCh38
rs753832427 Chr.2: 70181168-70181168 on Build GRCh38
rs771684373 Chr.2: 70181407-70181407 on Build GRCh38
rs147608350 Chr.2: 70181464-70181464 on Build GRCh38
rs773618316 Chr.2: 70181408-70181408 on Build GRCh38
rs747637155 Chr.2: 70181525-70181525 on Build GRCh38
rs749910015 Chr.2: 70181343-70181343 on Build GRCh38
rs761213773 Chr.2: 70181314-70181314 on Build GRCh38
rs756124141 Chr.2: 70181359-70181359 on Build GRCh38
rs116516305 Chr.2: 70181524-70181524 on Build GRCh38
rs1018264565 Chr.2: 70181145-70181145 on Build GRCh38
rs765291466 Chr.2: 70181460-70181460 on Build GRCh38
rs767807128 Chr.2: 70181152-70181152 on Build GRCh38
rs142004715 Chr.2: 70181312-70181312 on Build GRCh38
rs762199615 Chr.2: 70181143-70181143 on Build GRCh38
rs769610686 Chr.2: 70181265-70181265 on Build GRCh38
rs781072229 Chr.2: 70181246-70181246 on Build GRCh38
rs1018546263 Chr.2: 70181513-70181513 on Build GRCh38
rs751794327 Chr.2: 70181482-70181482 on Build GRCh38
rs1029740356 Chr.2: 70181250-70181250 on Build GRCh38
rs559141927 Chr.2: 70181535-70181535 on Build GRCh38
rs763620602 Chr.2: 70181133-70181133 on Build GRCh38
rs781199379 Chr.2: 70181328-70181328 on Build GRCh38
rs202165117 Chr.2: 70181465-70181465 on Build GRCh38
rs201828830 Chr.2: 70181504-70181504 on Build GRCh38
rs368806668 Chr.2: 70181321-70181321 on Build GRCh38
rs777005211 Chr.2: 70181430-70181430 on Build GRCh38
rs528190102 Chr.2: 70181563-70181563 on Build GRCh38
rs1005820327 Chr.2: 70181142-70181142 on Build GRCh38
rs866552727 Chr.2: 70181559-70181559 on Build GRCh38
rs749710534 Chr.2: 70181413-70181415 on Build GRCh38
rs752819943 Chr.2: 70181462-70181462 on Build GRCh38
rs750941568 Chr.2: 70181156-70181156 on Build GRCh38
rs750211207 Chr.2: 70181335-70181335 on Build GRCh38
rs146107703 Chr.2: 70181311-70181311 on Build GRCh38
rs941482806 Chr.2: 70181405-70181405 on Build GRCh38
rs577479936 Chr.2: 70181403-70181403 on Build GRCh38
rs373189922 Chr.2: 70181421-70181421 on Build GRCh38
rs774846597 Chr.2: 70181116-70181116 on Build GRCh38
rs748339283 Chr.2: 70181232-70181232 on Build GRCh38
rs754319328 Chr.2: 70181322-70181322 on Build GRCh38
rs564526178 Chr.2: 70181134-70181134 on Build GRCh38
rs745854692 Chr.2: 70181264-70181264 on Build GRCh38
rs769333261 Chr.2: 70181293-70181293 on Build GRCh38
rs148471734 Chr.2: 70181459-70181459 on Build GRCh38
rs778380987 Chr.2: 70181374-70181374 on Build GRCh38
rs760967013 Chr.2: 70181413-70181413 on Build GRCh38
rs748773824 Chr.2: 70181280-70181280 on Build GRCh38
rs761397237 Chr.2: 70181310-70181310 on Build GRCh38
rs978577289 Chr.2: 70181242-70181242 on Build GRCh38
rs758679774 Chr.2: 70181327-70181327 on Build GRCh38
rs756601319 Chr.2: 70181164-70181164 on Build GRCh38
rs977208870 Chr.2: 70181206-70181206 on Build GRCh38
rs868572578 Chr.2: 70181549-70181549 on Build GRCh38
rs768383241 Chr.2: 70181424-70181424 on Build GRCh38
rs755868066 Chr.2: 70181497-70181497 on Build GRCh38
rs139242568 Chr.2: 70181324-70181324 on Build GRCh38
rs775103656 Chr.2: 70181268-70181268 on Build GRCh38
rs967908176 Chr.2: 70181254-70181254 on Build GRCh38
rs747865624 Chr.2: 70181383-70181383 on Build GRCh38
rs1002283484 Chr.2: 70181511-70181511 on Build GRCh38
rs753778563 Chr.2: 70181502-70181502 on Build GRCh38
rs762730554 Chr.2: 70181117-70181117 on Build GRCh38
rs773883102 Chr.2: 70181294-70181294 on Build GRCh38
rs755274550 Chr.2: 70181196-70181196 on Build GRCh38
rs371588735 Chr.2: 70181530-70181530 on Build GRCh38
rs148177382 Chr.2: 70181203-70181203 on Build GRCh38
rs199928301 Chr.2: 70181495-70181495 on Build GRCh38

View Full Product Details