Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70984928-70985416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD53
Entrez Gene ID
79998
Gene Name
ankyrin repeat domain 53
Gene Aliases
-
UniGene
Hs.512744
Gene Chromosome Location
Chr.2: 70977183-70985499 on Build GRCh38
Transcripts
NM_001115116.1 NM_024933.3 XM_005264569.4 XM_011533114.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1409242 c.1452G>A p.R484R Chr.2: 70985159-70985159 on Build GRCh38
6319408 c.1486G>T p.D496Y Chr.2: 70985193-70985193 on Build GRCh38
6369194 c.1288G>T p.G430W Chr.2: 70984995-70984995 on Build GRCh38
3758339 c.1292A>T p.H431L Chr.2: 70984999-70984999 on Build GRCh38
1022579 c.1511A>G p.D504G Chr.2: 70985218-70985218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556018051 Chr.2: 70985010-70985010 on Build GRCh38
rs782330743 Chr.2: 70985099-70985099 on Build GRCh38
rs553757539 Chr.2: 70985162-70985162 on Build GRCh38
rs782773700 Chr.2: 70985204-70985204 on Build GRCh38
rs782394926 Chr.2: 70985303-70985303 on Build GRCh38
rs965399073 Chr.2: 70985328-70985328 on Build GRCh38
rs782219809 Chr.2: 70985096-70985096 on Build GRCh38
rs1025170524 Chr.2: 70985056-70985056 on Build GRCh38
rs781959053 Chr.2: 70984996-70984996 on Build GRCh38
rs11673798 Chr.2: 70985206-70985206 on Build GRCh38
rs961237001 Chr.2: 70985060-70985060 on Build GRCh38
rs782741308 Chr.2: 70985337-70985337 on Build GRCh38
rs782521839 Chr.2: 70985119-70985119 on Build GRCh38
rs944059619 Chr.2: 70985164-70985164 on Build GRCh38
rs527348428 Chr.2: 70985290-70985290 on Build GRCh38
rs538639855 Chr.2: 70985115-70985115 on Build GRCh38
rs75182953 Chr.2: 70985158-70985158 on Build GRCh38
rs61732288 Chr.2: 70985250-70985250 on Build GRCh38
rs781927317 Chr.2: 70985121-70985121 on Build GRCh38
rs375306837 Chr.2: 70985374-70985374 on Build GRCh38
rs782151063 Chr.2: 70985003-70985003 on Build GRCh38
rs993976275 Chr.2: 70985272-70985272 on Build GRCh38
rs538081467 Chr.2: 70985009-70985009 on Build GRCh38
rs782705065 Chr.2: 70985181-70985181 on Build GRCh38
rs1026862494 Chr.2: 70985091-70985091 on Build GRCh38
rs782630721 Chr.2: 70985093-70985093 on Build GRCh38
rs944147222 Chr.2: 70984946-70984946 on Build GRCh38
rs782570025 Chr.2: 70985362-70985362 on Build GRCh38
rs781996081 Chr.2: 70985288-70985288 on Build GRCh38
rs542795003 Chr.2: 70985051-70985051 on Build GRCh38
rs62145101 Chr.2: 70985372-70985372 on Build GRCh38
rs1010860983 Chr.2: 70984976-70984976 on Build GRCh38
rs1041554007 Chr.2: 70984954-70984954 on Build GRCh38
rs565572661 Chr.2: 70985245-70985245 on Build GRCh38
rs1055971718 Chr.2: 70985382-70985382 on Build GRCh38
rs377451472 Chr.2: 70985213-70985213 on Build GRCh38
rs782272530 Chr.2: 70985077-70985077 on Build GRCh38
rs575931933 Chr.2: 70985335-70985335 on Build GRCh38
rs782516129 Chr.2: 70985090-70985090 on Build GRCh38
rs544056308 Chr.2: 70985229-70985229 on Build GRCh38
rs782297188 Chr.2: 70985133-70985133 on Build GRCh38
rs61732279 Chr.2: 70985275-70985275 on Build GRCh38
rs781803085 Chr.2: 70985028-70985028 on Build GRCh38
rs782364072 Chr.2: 70984962-70984962 on Build GRCh38
rs782404739 Chr.2: 70985161-70985161 on Build GRCh38
rs1043281295 Chr.2: 70984995-70984995 on Build GRCh38
rs769409340 Chr.2: 70985312-70985312 on Build GRCh38
rs782717988 Chr.2: 70985027-70985027 on Build GRCh38
rs561067507 Chr.2: 70985324-70985324 on Build GRCh38
rs782129061 Chr.2: 70984961-70984961 on Build GRCh38
rs985355704 Chr.2: 70985320-70985320 on Build GRCh38
rs946202648 Chr.2: 70985187-70985187 on Build GRCh38
rs782259367 Chr.2: 70985247-70985247 on Build GRCh38
rs952623303 Chr.2: 70985116-70985116 on Build GRCh38
rs782431005 Chr.2: 70985050-70985050 on Build GRCh38
rs782540199 Chr.2: 70985195-70985195 on Build GRCh38
rs765365246 Chr.2: 70984955-70984955 on Build GRCh38
rs782575253 Chr.2: 70985137-70985137 on Build GRCh38
rs56204570 Chr.2: 70985163-70985163 on Build GRCh38
rs781812237 Chr.2: 70985339-70985339 on Build GRCh38
rs61732224 Chr.2: 70985000-70985000 on Build GRCh38
rs781952405 Chr.2: 70985173-70985173 on Build GRCh38
rs985427998 Chr.2: 70985132-70985132 on Build GRCh38
rs782043458 Chr.2: 70985002-70985002 on Build GRCh38
rs3796100 Chr.2: 70984999-70984999 on Build GRCh38
rs782498916 Chr.2: 70985020-70985020 on Build GRCh38
rs370566632 Chr.2: 70985176-70985176 on Build GRCh38
rs141581897 Chr.2: 70985251-70985251 on Build GRCh38
rs10460571 Chr.2: 70985302-70985302 on Build GRCh38
rs61735674 Chr.2: 70985170-70985170 on Build GRCh38
rs780587806 Chr.2: 70985134-70985134 on Build GRCh38
rs946141795 Chr.2: 70985357-70985357 on Build GRCh38
rs755507704 Chr.2: 70985235-70985235 on Build GRCh38
rs926111853 Chr.2: 70985366-70985366 on Build GRCh38

View Full Product Details