Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73243919-73244434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT7
Entrez Gene ID
10574
Gene Name
chaperonin containing TCP1 subunit 7
Gene Aliases
CCTETA, CCTH, NIP7-1, TCP1ETA
UniGene
Hs.368149
Gene Chromosome Location
Chr.2: 73233431-73253022 on Build GRCh38
Transcripts
NM_001009570.2 NM_001166284.1 NM_001166285.1 NM_006429.3 NR_029402.1 NR_029403.1 XM_011532478.2 XM_011532479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4967931 c.446+8T>G p.? Chr.2: 73244057-73244057 on Build GRCh38
6534495 c.426C>G p.T142T Chr.2: 73244029-73244029 on Build GRCh38
4426780 c.406A>G p.I136V Chr.2: 73244009-73244009 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759297454 Chr.2: 73244072-73244072 on Build GRCh38
rs755169051 Chr.2: 73244128-73244128 on Build GRCh38
rs796999950 Chr.2: 73244393-73244395 on Build GRCh38
rs373239788 Chr.2: 73244030-73244030 on Build GRCh38
rs755908763 Chr.2: 73244066-73244066 on Build GRCh38
rs779963174 Chr.2: 73244189-73244189 on Build GRCh38
rs1024878581 Chr.2: 73244071-73244072 on Build GRCh38
rs772376447 Chr.2: 73244066-73244068 on Build GRCh38
rs936087073 Chr.2: 73244345-73244345 on Build GRCh38
rs867801371 Chr.2: 73244040-73244040 on Build GRCh38
rs530074604 Chr.2: 73244287-73244287 on Build GRCh38
rs749763591 Chr.2: 73244066-73244067 on Build GRCh38
rs758948153 Chr.2: 73243972-73243972 on Build GRCh38
rs543597673 Chr.2: 73244205-73244205 on Build GRCh38
rs770450268 Chr.2: 73243951-73243951 on Build GRCh38
rs1864487 Chr.2: 73244223-73244223 on Build GRCh38
rs760929700 Chr.2: 73244082-73244083 on Build GRCh38
rs750704833 Chr.2: 73244048-73244048 on Build GRCh38
rs769560544 Chr.2: 73244013-73244013 on Build GRCh38
rs548703746 Chr.2: 73244307-73244307 on Build GRCh38
rs77843395 Chr.2: 73244393-73244393 on Build GRCh38
rs555638723 Chr.2: 73244197-73244197 on Build GRCh38
rs79785287 Chr.2: 73244085-73244085 on Build GRCh38
rs770812191 Chr.2: 73244069-73244069 on Build GRCh38
rs199840482 Chr.2: 73244029-73244029 on Build GRCh38
rs144024955 Chr.2: 73244190-73244190 on Build GRCh38
rs919444839 Chr.2: 73244263-73244264 on Build GRCh38
rs780169209 Chr.2: 73244060-73244060 on Build GRCh38
rs756146978 Chr.2: 73244058-73244058 on Build GRCh38
rs571913796 Chr.2: 73244119-73244119 on Build GRCh38
rs189131778 Chr.2: 73244339-73244339 on Build GRCh38
rs559710738 Chr.2: 73244285-73244285 on Build GRCh38
rs907187574 Chr.2: 73244102-73244102 on Build GRCh38
rs376089308 Chr.2: 73244061-73244061 on Build GRCh38
rs764877727 Chr.2: 73243980-73243980 on Build GRCh38
rs997264390 Chr.2: 73244268-73244268 on Build GRCh38
rs769412209 Chr.2: 73244079-73244079 on Build GRCh38
rs973104711 Chr.2: 73244242-73244242 on Build GRCh38
rs373353976 Chr.2: 73244094-73244094 on Build GRCh38
rs781406155 Chr.2: 73244316-73244316 on Build GRCh38
rs775078329 Chr.2: 73244081-73244081 on Build GRCh38
rs900274508 Chr.2: 73244265-73244265 on Build GRCh38
rs746736128 Chr.2: 73244068-73244068 on Build GRCh38
rs943560091 Chr.2: 73244308-73244309 on Build GRCh38
rs535954387 Chr.2: 73244162-73244162 on Build GRCh38
rs916013280 Chr.2: 73244065-73244065 on Build GRCh38
rs776369183 Chr.2: 73243964-73243964 on Build GRCh38
rs577441855 Chr.2: 73244276-73244276 on Build GRCh38
rs774509232 Chr.2: 73244065-73244066 on Build GRCh38
rs763611879 Chr.2: 73244011-73244011 on Build GRCh38
rs762060774 Chr.2: 73244091-73244091 on Build GRCh38
rs77312787 Chr.2: 73244086-73244086 on Build GRCh38
rs866148955 Chr.2: 73244229-73244229 on Build GRCh38
rs990689890 Chr.2: 73244379-73244379 on Build GRCh38
rs115773553 Chr.2: 73244395-73244395 on Build GRCh38
rs373030520 Chr.2: 73244089-73244089 on Build GRCh38
rs966382245 Chr.2: 73244320-73244320 on Build GRCh38
rs541333731 Chr.2: 73244279-73244279 on Build GRCh38
rs762552228 Chr.2: 73244082-73244082 on Build GRCh38
rs1017726590 Chr.2: 73244249-73244249 on Build GRCh38
rs867274129 Chr.2: 73244021-73244021 on Build GRCh38
rs531570896 Chr.2: 73244371-73244371 on Build GRCh38
rs1039059944 Chr.2: 73244338-73244338 on Build GRCh38
rs367760592 Chr.2: 73244357-73244357 on Build GRCh38
rs967059671 Chr.2: 73243967-73243967 on Build GRCh38
rs781220213 Chr.2: 73244037-73244037 on Build GRCh38
rs200159565 Chr.2: 73244007-73244007 on Build GRCh38
rs766537164 Chr.2: 73244090-73244090 on Build GRCh38
rs776118657 Chr.2: 73244071-73244071 on Build GRCh38
rs756272386 Chr.2: 73244095-73244095 on Build GRCh38
rs978002662 Chr.2: 73244326-73244326 on Build GRCh38
rs887955369 Chr.2: 73244222-73244222 on Build GRCh38
rs752128286 Chr.2: 73244087-73244087 on Build GRCh38
rs929560831 Chr.2: 73244188-73244188 on Build GRCh38
rs11289471 Chr.2: 73244066-73244066 on Build GRCh38
rs62149570 Chr.2: 73244063-73244063 on Build GRCh38

View Full Product Details