Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73946557-73947073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGUOK
Entrez Gene ID
1716
Gene Name
deoxyguanosine kinase
Gene Aliases
MTDPS3, dGK
UniGene
Hs.469022
Gene Chromosome Location
Chr.2: 73926826-73958961 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3364828 c.198T>A p.T66T Chr.2: 73946727-73946727 on Build GRCh38
4832619 c.288G>A p.L96L Chr.2: 73946751-73946751 on Build GRCh38
4832620 c.222G>A p.L74L Chr.2: 73946751-73946751 on Build GRCh38
4975140 c.304G>T p.E102* Chr.2: 73946833-73946833 on Build GRCh38
1532530 c.379C>T p.P127S Chr.2: 73946842-73946842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776244110 Chr.2: 73946777-73946777 on Build GRCh38
rs954458839 Chr.2: 73946778-73946778 on Build GRCh38
rs868412130 Chr.2: 73946677-73946677 on Build GRCh38
rs892630465 Chr.2: 73946650-73946650 on Build GRCh38
rs780959742 Chr.2: 73946855-73946855 on Build GRCh38
rs905247088 Chr.2: 73947002-73947002 on Build GRCh38
rs746603685 Chr.2: 73946762-73946762 on Build GRCh38
rs146274998 Chr.2: 73946947-73946947 on Build GRCh38
rs762169684 Chr.2: 73946897-73946897 on Build GRCh38
rs777348606 Chr.2: 73946760-73946760 on Build GRCh38
rs1015951530 Chr.2: 73946779-73946779 on Build GRCh38
rs371883253 Chr.2: 73946758-73946758 on Build GRCh38
rs762146219 Chr.2: 73946891-73946891 on Build GRCh38
rs940297 Chr.2: 73947018-73947018 on Build GRCh38
rs774437024 Chr.2: 73946811-73946811 on Build GRCh38
rs770463233 Chr.2: 73946783-73946783 on Build GRCh38
rs753439485 Chr.2: 73946602-73946602 on Build GRCh38
rs191541507 Chr.2: 73946689-73946689 on Build GRCh38
rs767604650 Chr.2: 73946815-73946815 on Build GRCh38
rs776060646 Chr.2: 73946785-73946785 on Build GRCh38
rs759093548 Chr.2: 73946781-73946781 on Build GRCh38
rs917899937 Chr.2: 73946989-73946989 on Build GRCh38
rs138683121 Chr.2: 73946728-73946728 on Build GRCh38
rs199645258 Chr.2: 73946829-73946829 on Build GRCh38
rs745436352 Chr.2: 73946857-73946857 on Build GRCh38
rs752581622 Chr.2: 73946955-73946955 on Build GRCh38
rs540511330 Chr.2: 73946661-73946661 on Build GRCh38
rs777710215 Chr.2: 73946845-73946845 on Build GRCh38
rs947344769 Chr.2: 73946599-73946599 on Build GRCh38
rs147540040 Chr.2: 73946812-73946812 on Build GRCh38
rs750602924 Chr.2: 73946693-73946695 on Build GRCh38
rs138535698 Chr.2: 73946664-73946664 on Build GRCh38
rs910349788 Chr.2: 73946591-73946592 on Build GRCh38
rs568816337 Chr.2: 73946713-73946713 on Build GRCh38
rs777559041 Chr.2: 73946833-73946833 on Build GRCh38
rs757017019 Chr.2: 73946850-73946850 on Build GRCh38
rs932394897 Chr.2: 73946830-73946830 on Build GRCh38
rs182536755 Chr.2: 73946928-73946928 on Build GRCh38
rs1804599 Chr.2: 73946886-73946886 on Build GRCh38
rs367550768 Chr.2: 73946683-73946683 on Build GRCh38
rs756158114 Chr.2: 73946824-73946824 on Build GRCh38
rs144587099 Chr.2: 73946642-73946642 on Build GRCh38
rs199531052 Chr.2: 73946712-73946712 on Build GRCh38
rs753051426 Chr.2: 73946711-73946711 on Build GRCh38
rs374557161 Chr.2: 73946696-73946696 on Build GRCh38
rs377009447 Chr.2: 73946733-73946733 on Build GRCh38
rs368033322 Chr.2: 73946740-73946740 on Build GRCh38
rs747937764 Chr.2: 73946743-73946743 on Build GRCh38
rs924541257 Chr.2: 73946614-73946616 on Build GRCh38
rs1050768690 Chr.2: 73946589-73946589 on Build GRCh38
rs769333517 Chr.2: 73946865-73946865 on Build GRCh38
rs368995530 Chr.2: 73946807-73946807 on Build GRCh38
rs543582938 Chr.2: 73946662-73946662 on Build GRCh38
rs974091914 Chr.2: 73946714-73946714 on Build GRCh38
rs752671697 Chr.2: 73946827-73946827 on Build GRCh38
rs750553734 Chr.2: 73946819-73946819 on Build GRCh38
rs753800926 Chr.2: 73946731-73946731 on Build GRCh38
rs749733889 Chr.2: 73946874-73946874 on Build GRCh38
rs764457214 Chr.2: 73946806-73946806 on Build GRCh38
rs368635864 Chr.2: 73946707-73946707 on Build GRCh38
rs768929581 Chr.2: 73946878-73946878 on Build GRCh38
rs898075148 Chr.2: 73946715-73946715 on Build GRCh38
rs754601584 Chr.2: 73946617-73946617 on Build GRCh38
rs781178702 Chr.2: 73946730-73946730 on Build GRCh38
rs770571614 Chr.2: 73946764-73946764 on Build GRCh38
rs139220446 Chr.2: 73946986-73946986 on Build GRCh38
rs141810774 Chr.2: 73946800-73946800 on Build GRCh38
rs775871024 Chr.2: 73946868-73946868 on Build GRCh38
rs587780587 Chr.2: 73946750-73946750 on Build GRCh38
rs200132518 Chr.2: 73946606-73946606 on Build GRCh38
rs1002136730 Chr.2: 73946684-73946684 on Build GRCh38
rs201519508 Chr.2: 73946699-73946699 on Build GRCh38
rs373983566 Chr.2: 73946753-73946753 on Build GRCh38
rs538005973 Chr.2: 73947042-73947042 on Build GRCh38
rs1052682825 Chr.2: 73946904-73946904 on Build GRCh38
rs104893631 Chr.2: 73946888-73946888 on Build GRCh38
rs104893630 Chr.2: 73946776-73946776 on Build GRCh38
rs1009162635 Chr.2: 73946640-73946640 on Build GRCh38
rs150678946 Chr.2: 73946816-73946816 on Build GRCh38
rs982717858 Chr.2: 73946738-73946738 on Build GRCh38
rs758552684 Chr.2: 73946881-73946881 on Build GRCh38
rs979112534 Chr.2: 73946604-73946604 on Build GRCh38
rs550390955 Chr.2: 73946818-73946818 on Build GRCh38
rs767770283 Chr.2: 73946922-73946922 on Build GRCh38
rs534254321 Chr.2: 73946930-73946930 on Build GRCh38
rs1011514211 Chr.2: 73946655-73946655 on Build GRCh38

View Full Product Details