Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74522772-74523268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
UniGene
Hs.191705
Gene Chromosome Location
Chr.2: 74518131-74526281 on Build GRCh38
Transcripts
NM_133637.2 XM_011532644.1 XM_011532645.1 XM_011532646.2 XM_017003520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583649 c.915A>G p.A305A Chr.2: 74522890-74522890 on Build GRCh38
4679346 c.1255_1256insA p.R419fs*36 Chr.2: 74522903-74522904 on Build GRCh38
6528365 c.782T>G p.F261C Chr.2: 74523127-74523127 on Build GRCh38
6564173 c.865G>A p.E289K Chr.2: 74522940-74522940 on Build GRCh38
6649190 c.701C>T p.P234L Chr.2: 74523208-74523208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769454015 Chr.2: 74523209-74523220 on Build GRCh38
rs889634899 Chr.2: 74522933-74522933 on Build GRCh38
rs760402229 Chr.2: 74522924-74522924 on Build GRCh38
rs1038259637 Chr.2: 74522964-74522964 on Build GRCh38
rs777001695 Chr.2: 74523232-74523232 on Build GRCh38
rs780651175 Chr.2: 74522887-74522887 on Build GRCh38
rs779712190 Chr.2: 74523047-74523047 on Build GRCh38
rs747733841 Chr.2: 74522904-74522904 on Build GRCh38
rs141416881 Chr.2: 74522882-74522882 on Build GRCh38
rs753250249 Chr.2: 74523143-74523143 on Build GRCh38
rs758410172 Chr.2: 74522824-74522824 on Build GRCh38
rs749233518 Chr.2: 74523209-74523209 on Build GRCh38
rs746573993 Chr.2: 74523162-74523162 on Build GRCh38
rs771621967 Chr.2: 74523061-74523061 on Build GRCh38
rs780082894 Chr.2: 74522797-74522803 on Build GRCh38
rs969556431 Chr.2: 74523104-74523104 on Build GRCh38
rs754214204 Chr.2: 74523130-74523130 on Build GRCh38
rs376345068 Chr.2: 74523215-74523215 on Build GRCh38
rs761737636 Chr.2: 74522897-74522898 on Build GRCh38
rs766747892 Chr.2: 74523129-74523129 on Build GRCh38
rs759370075 Chr.2: 74523094-74523094 on Build GRCh38
rs190151140 Chr.2: 74522912-74522912 on Build GRCh38
rs775521069 Chr.2: 74523103-74523103 on Build GRCh38
rs753784679 Chr.2: 74522822-74522822 on Build GRCh38
rs769646302 Chr.2: 74523200-74523200 on Build GRCh38
rs534122179 Chr.2: 74523153-74523153 on Build GRCh38
rs374979720 Chr.2: 74522994-74522994 on Build GRCh38
rs749806738 Chr.2: 74522890-74522890 on Build GRCh38
rs759458187 Chr.2: 74522943-74522943 on Build GRCh38
rs766253248 Chr.2: 74522814-74522814 on Build GRCh38
rs994488412 Chr.2: 74522992-74522992 on Build GRCh38
rs370443282 Chr.2: 74522827-74522827 on Build GRCh38
rs376136178 Chr.2: 74523112-74523112 on Build GRCh38
rs769579657 Chr.2: 74523100-74523100 on Build GRCh38
rs546999902 Chr.2: 74523194-74523194 on Build GRCh38
rs759749434 Chr.2: 74523236-74523236 on Build GRCh38
rs765627628 Chr.2: 74523238-74523238 on Build GRCh38
rs200302410 Chr.2: 74523199-74523199 on Build GRCh38
rs778264602 Chr.2: 74522897-74522897 on Build GRCh38
rs370698471 Chr.2: 74523090-74523092 on Build GRCh38
rs371529330 Chr.2: 74523069-74523069 on Build GRCh38
rs770715047 Chr.2: 74522938-74522938 on Build GRCh38
rs757402658 Chr.2: 74522859-74522859 on Build GRCh38
rs750524824 Chr.2: 74523125-74523125 on Build GRCh38
rs532949134 Chr.2: 74523022-74523022 on Build GRCh38
rs372058865 Chr.2: 74522961-74522961 on Build GRCh38
rs750713785 Chr.2: 74522878-74522878 on Build GRCh38
rs368436046 Chr.2: 74523219-74523219 on Build GRCh38
rs868387733 Chr.2: 74522988-74522988 on Build GRCh38
rs974613002 Chr.2: 74522908-74522908 on Build GRCh38
rs761984659 Chr.2: 74522968-74522968 on Build GRCh38
rs575699401 Chr.2: 74523240-74523240 on Build GRCh38
rs777327109 Chr.2: 74523146-74523146 on Build GRCh38
rs771580472 Chr.2: 74522910-74522910 on Build GRCh38
rs781518203 Chr.2: 74522866-74522866 on Build GRCh38
rs747557924 Chr.2: 74523059-74523059 on Build GRCh38
rs781159473 Chr.2: 74523018-74523019 on Build GRCh38
rs775225024 Chr.2: 74523207-74523207 on Build GRCh38
rs751550844 Chr.2: 74522804-74522805 on Build GRCh38
rs772848624 Chr.2: 74522802-74522802 on Build GRCh38
rs930202995 Chr.2: 74522973-74522973 on Build GRCh38
rs767587992 Chr.2: 74522970-74522970 on Build GRCh38
rs1025676516 Chr.2: 74523023-74523023 on Build GRCh38
rs760545850 Chr.2: 74522804-74522804 on Build GRCh38
rs150420269 Chr.2: 74523140-74523140 on Build GRCh38
rs75644451 Chr.2: 74523228-74523228 on Build GRCh38
rs551563920 Chr.2: 74523084-74523084 on Build GRCh38
rs756469372 Chr.2: 74523002-74523002 on Build GRCh38
rs756876809 Chr.2: 74523178-74523178 on Build GRCh38
rs200086801 Chr.2: 74522911-74522911 on Build GRCh38
rs754500864 Chr.2: 74523213-74523213 on Build GRCh38
rs367740434 Chr.2: 74523060-74523060 on Build GRCh38
rs566651196 Chr.2: 74523123-74523123 on Build GRCh38
rs755509943 Chr.2: 74523032-74523032 on Build GRCh38
rs892281613 Chr.2: 74523172-74523172 on Build GRCh38
rs73949679 Chr.2: 74523214-74523214 on Build GRCh38
rs562873082 Chr.2: 74523087-74523089 on Build GRCh38
rs1052830647 Chr.2: 74522841-74522841 on Build GRCh38
rs944040539 Chr.2: 74523119-74523119 on Build GRCh38
rs766655961 Chr.2: 74523230-74523230 on Build GRCh38
rs765211532 Chr.2: 74522952-74522952 on Build GRCh38
rs767765043 Chr.2: 74522839-74522839 on Build GRCh38
rs777221645 Chr.2: 74523068-74523068 on Build GRCh38
rs780550990 Chr.2: 74523015-74523015 on Build GRCh38
rs146840492 Chr.2: 74523126-74523126 on Build GRCh38
rs769342695 Chr.2: 74522895-74522895 on Build GRCh38
rs770629683 Chr.2: 74523082-74523082 on Build GRCh38
rs758892134 Chr.2: 74523145-74523145 on Build GRCh38

View Full Product Details