Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74523232-74523739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
UniGene
Hs.191705
Gene Chromosome Location
Chr.2: 74518131-74526281 on Build GRCh38
Transcripts
NM_133637.2 XM_011532644.1 XM_011532645.1 XM_011532646.2 XM_017003520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
364501 c.599G>A p.R200Q Chr.2: 74523401-74523401 on Build GRCh38
4679348 c.949G>A p.A317T Chr.2: 74523405-74523405 on Build GRCh38
2157373 c.626_627delCT p.A209fs*26 Chr.2: 74523373-74523374 on Build GRCh38
576007 c.535G>A p.V179I Chr.2: 74523465-74523465 on Build GRCh38
5789256 c.921C>G p.A307A Chr.2: 74523433-74523433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760123965 Chr.2: 74523480-74523480 on Build GRCh38
rs540562599 Chr.2: 74523432-74523432 on Build GRCh38
rs147197958 Chr.2: 74523377-74523377 on Build GRCh38
rs774765935 Chr.2: 74523444-74523444 on Build GRCh38
rs146254354 Chr.2: 74523433-74523433 on Build GRCh38
rs780255814 Chr.2: 74523570-74523570 on Build GRCh38
rs778834235 Chr.2: 74523531-74523531 on Build GRCh38
rs746815800 Chr.2: 74523334-74523334 on Build GRCh38
rs754741249 Chr.2: 74523524-74523524 on Build GRCh38
rs777890921 Chr.2: 74523558-74523558 on Build GRCh38
rs910938008 Chr.2: 74523278-74523278 on Build GRCh38
rs573274390 Chr.2: 74523438-74523438 on Build GRCh38
rs749076346 Chr.2: 74523309-74523309 on Build GRCh38
rs879198421 Chr.2: 74523589-74523589 on Build GRCh38
rs774452490 Chr.2: 74523315-74523315 on Build GRCh38
rs1046985528 Chr.2: 74523674-74523674 on Build GRCh38
rs753980493 Chr.2: 74523467-74523467 on Build GRCh38
rs977969760 Chr.2: 74523452-74523452 on Build GRCh38
rs957077033 Chr.2: 74523576-74523576 on Build GRCh38
rs749907332 Chr.2: 74523468-74523468 on Build GRCh38
rs201705642 Chr.2: 74523437-74523437 on Build GRCh38
rs751033879 Chr.2: 74523261-74523261 on Build GRCh38
rs748114940 Chr.2: 74523295-74523295 on Build GRCh38
rs770233601 Chr.2: 74523292-74523295 on Build GRCh38
rs765998279 Chr.2: 74523494-74523494 on Build GRCh38
rs780765276 Chr.2: 74523291-74523291 on Build GRCh38
rs758380492 Chr.2: 74523551-74523551 on Build GRCh38
rs771135773 Chr.2: 74523333-74523333 on Build GRCh38
rs367789352 Chr.2: 74523560-74523560 on Build GRCh38
rs573451528 Chr.2: 74523402-74523402 on Build GRCh38
rs190988345 Chr.2: 74523650-74523650 on Build GRCh38
rs371651616 Chr.2: 74523305-74523305 on Build GRCh38
rs201047482 Chr.2: 74523465-74523465 on Build GRCh38
rs989928401 Chr.2: 74523710-74523710 on Build GRCh38
rs781401427 Chr.2: 74523435-74523435 on Build GRCh38
rs369535167 Chr.2: 74523277-74523277 on Build GRCh38
rs769780881 Chr.2: 74523568-74523568 on Build GRCh38
rs866282693 Chr.2: 74523363-74523365 on Build GRCh38
rs143706179 Chr.2: 74523370-74523370 on Build GRCh38
rs770357979 Chr.2: 74523468-74523470 on Build GRCh38
rs777353299 Chr.2: 74523357-74523358 on Build GRCh38
rs778924021 Chr.2: 74523421-74523421 on Build GRCh38
rs745977901 Chr.2: 74523436-74523436 on Build GRCh38
rs774772212 Chr.2: 74523557-74523564 on Build GRCh38
rs749983331 Chr.2: 74523401-74523401 on Build GRCh38
rs752586868 Chr.2: 74523540-74523540 on Build GRCh38
rs929775250 Chr.2: 74523666-74523666 on Build GRCh38
rs562369621 Chr.2: 74523625-74523625 on Build GRCh38
rs374870900 Chr.2: 74523362-74523362 on Build GRCh38
rs753619516 Chr.2: 74523408-74523408 on Build GRCh38
rs1000944255 Chr.2: 74523549-74523549 on Build GRCh38
rs761246637 Chr.2: 74523387-74523387 on Build GRCh38
rs910887018 Chr.2: 74523629-74523629 on Build GRCh38
rs748813867 Chr.2: 74523426-74523426 on Build GRCh38
rs762195332 Chr.2: 74523578-74523578 on Build GRCh38
rs777952294 Chr.2: 74523255-74523255 on Build GRCh38
rs942485807 Chr.2: 74523634-74523634 on Build GRCh38
rs982881353 Chr.2: 74523624-74523624 on Build GRCh38
rs773034844 Chr.2: 74523314-74523314 on Build GRCh38
rs768926128 Chr.2: 74523577-74523577 on Build GRCh38
rs747022988 Chr.2: 74523318-74523318 on Build GRCh38
rs150732366 Chr.2: 74523381-74523381 on Build GRCh38
rs181044530 Chr.2: 74523253-74523253 on Build GRCh38
rs754738930 Chr.2: 74523415-74523415 on Build GRCh38
rs753433578 Chr.2: 74523508-74523508 on Build GRCh38
rs769014157 Chr.2: 74523440-74523440 on Build GRCh38
rs185264067 Chr.2: 74523374-74523374 on Build GRCh38
rs1033738144 Chr.2: 74523517-74523517 on Build GRCh38
rs749467915 Chr.2: 74523572-74523572 on Build GRCh38
rs756861841 Chr.2: 74523702-74523702 on Build GRCh38
rs745915437 Chr.2: 74523559-74523559 on Build GRCh38
rs533094538 Chr.2: 74523654-74523654 on Build GRCh38
rs759787740 Chr.2: 74523347-74523347 on Build GRCh38
rs767012140 Chr.2: 74523396-74523396 on Build GRCh38
rs955075894 Chr.2: 74523598-74523598 on Build GRCh38

View Full Product Details