Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75560589-75561080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1A
Entrez Gene ID
84141
Gene Name
eva-1 homolog A, regulator of programmed cell death
Gene Aliases
FAM176A, TMEM166
UniGene
Hs.302346
Gene Chromosome Location
Chr.2: 75492318-75569722 on Build GRCh38
Transcripts
NM_001135032.1 NM_032181.2

SNP Detail

SNP ID Chromosome Location
rs893904224 Chr.2: 75560800-75560800 on Build GRCh38
rs58578233 Chr.2: 75560642-75560642 on Build GRCh38
rs940269706 Chr.2: 75560738-75560738 on Build GRCh38
rs113609413 Chr.2: 75560625-75560625 on Build GRCh38
rs1028053983 Chr.2: 75560976-75560976 on Build GRCh38
rs952410084 Chr.2: 75560712-75560712 on Build GRCh38
rs796662814 Chr.2: 75561001-75561001 on Build GRCh38
rs928237810 Chr.2: 75561055-75561055 on Build GRCh38
rs900998218 Chr.2: 75560920-75560920 on Build GRCh38
rs908782363 Chr.2: 75560735-75560735 on Build GRCh38
rs984216890 Chr.2: 75560730-75560730 on Build GRCh38
rs928121583 Chr.2: 75560772-75560772 on Build GRCh38
rs555139120 Chr.2: 75561018-75561018 on Build GRCh38
rs1019423493 Chr.2: 75560611-75560611 on Build GRCh38
rs1019371610 Chr.2: 75560876-75560876 on Build GRCh38
rs568255854 Chr.2: 75561046-75561046 on Build GRCh38
rs1015288465 Chr.2: 75561020-75561020 on Build GRCh38
rs921046770 Chr.2: 75560656-75560656 on Build GRCh38
rs11538749 Chr.2: 75560846-75560846 on Build GRCh38
rs931407806 Chr.2: 75560866-75560866 on Build GRCh38
rs1041187421 Chr.2: 75560963-75560963 on Build GRCh38
rs997123197 Chr.2: 75560942-75560942 on Build GRCh38
rs998730477 Chr.2: 75560868-75560868 on Build GRCh38
rs991421874 Chr.2: 75561058-75561058 on Build GRCh38
rs900101032 Chr.2: 75560972-75560972 on Build GRCh38
rs996256858 Chr.2: 75560643-75560643 on Build GRCh38
rs113295675 Chr.2: 75560922-75560922 on Build GRCh38
rs771919880 Chr.2: 75560618-75560618 on Build GRCh38
rs533277400 Chr.2: 75560710-75560710 on Build GRCh38
rs892892103 Chr.2: 75560648-75560648 on Build GRCh38
rs533692484 Chr.2: 75560994-75560994 on Build GRCh38
rs908755285 Chr.2: 75560903-75560903 on Build GRCh38
rs1011296969 Chr.2: 75560690-75560690 on Build GRCh38
rs550626205 Chr.2: 75560632-75560632 on Build GRCh38
rs938062618 Chr.2: 75560790-75560790 on Build GRCh38
rs528997840 Chr.2: 75560627-75560627 on Build GRCh38
rs903083129 Chr.2: 75560860-75560860 on Build GRCh38
rs1010892473 Chr.2: 75560817-75560817 on Build GRCh38
rs377693990 Chr.2: 75560643-75560644 on Build GRCh38
rs952599206 Chr.2: 75560978-75560978 on Build GRCh38
rs983775704 Chr.2: 75561016-75561016 on Build GRCh38
rs568735660 Chr.2: 75560644-75560644 on Build GRCh38
rs398090402 Chr.2: 75560646-75560646 on Build GRCh38
rs938193556 Chr.2: 75561057-75561057 on Build GRCh38
rs369751818 Chr.2: 75560761-75560761 on Build GRCh38
rs982824203 Chr.2: 75560878-75560878 on Build GRCh38
rs1046579149 Chr.2: 75560752-75560752 on Build GRCh38
rs1050094406 Chr.2: 75560980-75560980 on Build GRCh38
rs115603825 Chr.2: 75560791-75560791 on Build GRCh38
rs991916838 Chr.2: 75560831-75560831 on Build GRCh38
rs982370589 Chr.2: 75561048-75561048 on Build GRCh38

View Full Product Details