Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75662514-75663018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCFC2
Entrez Gene ID
6936
Gene Name
GC-rich sequence DNA-binding factor 2
Gene Aliases
C2orf3, DNABF, GCF, TCF9
UniGene
Hs.303808
Gene Chromosome Location
Chr.2: 75662655-75710986 on Build GRCh38
Transcripts
NM_001201334.1 NM_003203.4 XM_005264520.3 XM_011533074.2

SNP Detail

SNP ID Chromosome Location
rs972982029 Chr.2: 75662690-75662690 on Build GRCh38
rs3082417 Chr.2: 75662605-75662606 on Build GRCh38
rs1014022265 Chr.2: 75662905-75662908 on Build GRCh38
rs771822150 Chr.2: 75662992-75662992 on Build GRCh38
rs1037110222 Chr.2: 75662967-75662967 on Build GRCh38
rs984122924 Chr.2: 75662645-75662645 on Build GRCh38
rs5832171 Chr.2: 75662602-75662603 on Build GRCh38
rs567173416 Chr.2: 75662919-75662919 on Build GRCh38
rs188919747 Chr.2: 75662977-75662977 on Build GRCh38
rs544911575 Chr.2: 75662756-75662756 on Build GRCh38
rs76738743 Chr.2: 75662881-75662881 on Build GRCh38
rs1063988 Chr.2: 75662837-75662837 on Build GRCh38
rs922730812 Chr.2: 75662691-75662691 on Build GRCh38
rs1016913486 Chr.2: 75662917-75662917 on Build GRCh38
rs1022742094 Chr.2: 75662662-75662662 on Build GRCh38
rs757715380 Chr.2: 75662930-75662930 on Build GRCh38
rs767730058 Chr.2: 75662915-75662915 on Build GRCh38
rs1037545366 Chr.2: 75662538-75662538 on Build GRCh38
rs17741121 Chr.2: 75662646-75662646 on Build GRCh38
rs543279168 Chr.2: 75662574-75662574 on Build GRCh38
rs910283674 Chr.2: 75662787-75662787 on Build GRCh38
rs1006054783 Chr.2: 75662916-75662917 on Build GRCh38
rs968981683 Chr.2: 75662714-75662714 on Build GRCh38
rs941562413 Chr.2: 75662957-75662957 on Build GRCh38
rs955823977 Chr.2: 75662813-75662813 on Build GRCh38
rs115374124 Chr.2: 75662974-75662974 on Build GRCh38
rs532030470 Chr.2: 75662868-75662868 on Build GRCh38
rs897150095 Chr.2: 75662568-75662568 on Build GRCh38
rs1052976614 Chr.2: 75662885-75662885 on Build GRCh38
rs904217227 Chr.2: 75662858-75662858 on Build GRCh38
rs531966434 Chr.2: 75662600-75662600 on Build GRCh38
rs895773603 Chr.2: 75662903-75662903 on Build GRCh38
rs1031873797 Chr.2: 75662793-75662793 on Build GRCh38
rs879027983 Chr.2: 75662905-75662905 on Build GRCh38
rs73936764 Chr.2: 75662920-75662920 on Build GRCh38
rs548501175 Chr.2: 75662954-75662954 on Build GRCh38
rs779468650 Chr.2: 75662937-75662937 on Build GRCh38
rs943142521 Chr.2: 75662790-75662790 on Build GRCh38
rs932861064 Chr.2: 75662947-75662947 on Build GRCh38
rs17011635 Chr.2: 75662780-75662780 on Build GRCh38
rs199648088 Chr.2: 75662920-75662921 on Build GRCh38
rs911386328 Chr.2: 75662986-75662986 on Build GRCh38
rs202107416 Chr.2: 75662588-75662588 on Build GRCh38
rs970569521 Chr.2: 75662634-75662634 on Build GRCh38
rs922281045 Chr.2: 75662925-75662925 on Build GRCh38
rs529963383 Chr.2: 75662948-75662948 on Build GRCh38
rs530582058 Chr.2: 75662818-75662818 on Build GRCh38
rs1023246971 Chr.2: 75662914-75662915 on Build GRCh38
rs373576816 Chr.2: 75662602-75662602 on Build GRCh38
rs961596913 Chr.2: 75662674-75662674 on Build GRCh38
rs1042702000 Chr.2: 75662826-75662827 on Build GRCh38

View Full Product Details