Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84547141-84547669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
UniGene
Hs.249972
Gene Chromosome Location
Chr.2: 84459588-84819589 on Build GRCh38
Transcripts
NM_001370.1 XM_006711956.2 XM_011532649.2 XM_011532650.2 XM_011532652.1 XM_011532653.1 XM_011532654.1 XM_011532655.2 XM_011532657.1 XM_011532660.1 XM_017003521.1 XM_017003522.1 XM_017003523.1 XM_017003524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1409720 c.1181T>G p.F394C Chr.2: 84547607-84547607 on Build GRCh38
4388720 c.1055G>A p.R352Q Chr.2: 84547392-84547392 on Build GRCh38
1023244 c.937C>T p.R313* Chr.2: 84547274-84547274 on Build GRCh38
4936532 c.1090C>T p.R364* Chr.2: 84547516-84547516 on Build GRCh38
7246359 c.1101A>G p.A367A Chr.2: 84547527-84547527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942414865 Chr.2: 84547280-84547280 on Build GRCh38
rs977616716 Chr.2: 84547192-84547192 on Build GRCh38
rs762684833 Chr.2: 84547637-84547637 on Build GRCh38
rs752965638 Chr.2: 84547469-84547469 on Build GRCh38
rs751518371 Chr.2: 84547378-84547378 on Build GRCh38
rs780554299 Chr.2: 84547283-84547283 on Build GRCh38
rs759180925 Chr.2: 84547229-84547229 on Build GRCh38
rs747357611 Chr.2: 84547381-84547381 on Build GRCh38
rs978260141 Chr.2: 84547183-84547183 on Build GRCh38
rs201462081 Chr.2: 84547273-84547273 on Build GRCh38
rs923446938 Chr.2: 84547193-84547193 on Build GRCh38
rs780629101 Chr.2: 84547520-84547520 on Build GRCh38
rs114989346 Chr.2: 84547630-84547630 on Build GRCh38
rs775093942 Chr.2: 84547334-84547334 on Build GRCh38
rs531016943 Chr.2: 84547501-84547501 on Build GRCh38
rs955084404 Chr.2: 84547215-84547215 on Build GRCh38
rs373968372 Chr.2: 84547275-84547275 on Build GRCh38
rs999127335 Chr.2: 84547523-84547523 on Build GRCh38
rs1052288738 Chr.2: 84547345-84547345 on Build GRCh38
rs763975231 Chr.2: 84547373-84547373 on Build GRCh38
rs954865988 Chr.2: 84547582-84547582 on Build GRCh38
rs747560481 Chr.2: 84547502-84547502 on Build GRCh38
rs1022282290 Chr.2: 84547470-84547470 on Build GRCh38
rs866819036 Chr.2: 84547613-84547613 on Build GRCh38
rs768969450 Chr.2: 84547462-84547462 on Build GRCh38
rs563527813 Chr.2: 84547488-84547488 on Build GRCh38
rs1025162423 Chr.2: 84547548-84547548 on Build GRCh38
rs1055920327 Chr.2: 84547416-84547416 on Build GRCh38
rs1036839300 Chr.2: 84547431-84547431 on Build GRCh38
rs758836169 Chr.2: 84547212-84547212 on Build GRCh38
rs1054829013 Chr.2: 84547475-84547475 on Build GRCh38
rs898445754 Chr.2: 84547444-84547444 on Build GRCh38
rs1038688272 Chr.2: 84547335-84547335 on Build GRCh38
rs529015397 Chr.2: 84547588-84547588 on Build GRCh38
rs545338994 Chr.2: 84547484-84547484 on Build GRCh38
rs763465058 Chr.2: 84547419-84547419 on Build GRCh38
rs141384152 Chr.2: 84547564-84547564 on Build GRCh38
rs933454792 Chr.2: 84547331-84547331 on Build GRCh38
rs367544135 Chr.2: 84547639-84547639 on Build GRCh38
rs867472657 Chr.2: 84547516-84547516 on Build GRCh38
rs966457262 Chr.2: 84547553-84547553 on Build GRCh38
rs755213863 Chr.2: 84547456-84547456 on Build GRCh38
rs566972871 Chr.2: 84547392-84547392 on Build GRCh38
rs543025934 Chr.2: 84547254-84547254 on Build GRCh38
rs372206908 Chr.2: 84547213-84547213 on Build GRCh38
rs200299504 Chr.2: 84547565-84547565 on Build GRCh38
rs770563696 Chr.2: 84547517-84547517 on Build GRCh38
rs1002880052 Chr.2: 84547391-84547391 on Build GRCh38
rs919875288 Chr.2: 84547336-84547336 on Build GRCh38
rs34059681 Chr.2: 84547209-84547209 on Build GRCh38
rs533653812 Chr.2: 84547379-84547379 on Build GRCh38

View Full Product Details