Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84548154-84548666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
UniGene
Hs.249972
Gene Chromosome Location
Chr.2: 84459588-84819589 on Build GRCh38
Transcripts
NM_001370.1 XM_006711956.2 XM_011532649.2 XM_011532650.2 XM_011532652.1 XM_011532653.1 XM_011532654.1 XM_011532655.2 XM_011532657.1 XM_011532660.1 XM_017003521.1 XM_017003522.1 XM_017003523.1 XM_017003524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453821 c.1314G>A p.T438T Chr.2: 84548415-84548415 on Build GRCh38
4924621 c.1247A>T p.Y416F Chr.2: 84548348-84548348 on Build GRCh38
6092508 c.1232A>G p.H411R Chr.2: 84548333-84548333 on Build GRCh38
1023246 c.1267A>G p.T423A Chr.2: 84548368-84548368 on Build GRCh38
6092507 c.1232A>G p.H411R Chr.2: 84548333-84548333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778351682 Chr.2: 84548280-84548280 on Build GRCh38
rs755669216 Chr.2: 84548342-84548342 on Build GRCh38
rs947241920 Chr.2: 84548517-84548517 on Build GRCh38
rs771837943 Chr.2: 84548373-84548373 on Build GRCh38
rs1027751650 Chr.2: 84548550-84548550 on Build GRCh38
rs757779722 Chr.2: 84548260-84548262 on Build GRCh38
rs559262254 Chr.2: 84548500-84548500 on Build GRCh38
rs191349552 Chr.2: 84548594-84548594 on Build GRCh38
rs773224592 Chr.2: 84548375-84548375 on Build GRCh38
rs757723655 Chr.2: 84548419-84548419 on Build GRCh38
rs752333165 Chr.2: 84548577-84548577 on Build GRCh38
rs758863666 Chr.2: 84548440-84548441 on Build GRCh38
rs780373359 Chr.2: 84548444-84548445 on Build GRCh38
rs561353874 Chr.2: 84548352-84548352 on Build GRCh38
rs532962134 Chr.2: 84548521-84548521 on Build GRCh38
rs745326515 Chr.2: 84548340-84548340 on Build GRCh38
rs774188153 Chr.2: 84548370-84548370 on Build GRCh38
rs962040036 Chr.2: 84548536-84548536 on Build GRCh38
rs991084571 Chr.2: 84548575-84548575 on Build GRCh38
rs767860081 Chr.2: 84548433-84548433 on Build GRCh38
rs1051998472 Chr.2: 84548196-84548196 on Build GRCh38
rs934964765 Chr.2: 84548353-84548353 on Build GRCh38
rs750984738 Chr.2: 84548438-84548438 on Build GRCh38
rs765724240 Chr.2: 84548370-84548371 on Build GRCh38
rs916513837 Chr.2: 84548259-84548259 on Build GRCh38
rs779547478 Chr.2: 84548343-84548343 on Build GRCh38
rs771952808 Chr.2: 84548461-84548461 on Build GRCh38
rs890727930 Chr.2: 84548496-84548496 on Build GRCh38
rs750748861 Chr.2: 84548418-84548419 on Build GRCh38
rs765400269 Chr.2: 84548393-84548393 on Build GRCh38
rs756333599 Chr.2: 84548584-84548584 on Build GRCh38
rs775853186 Chr.2: 84548410-84548410 on Build GRCh38
rs60315854 Chr.2: 84548549-84548549 on Build GRCh38
rs759512799 Chr.2: 84548390-84548390 on Build GRCh38
rs376004002 Chr.2: 84548417-84548417 on Build GRCh38
rs958765411 Chr.2: 84548553-84548553 on Build GRCh38
rs747889923 Chr.2: 84548459-84548459 on Build GRCh38
rs1008235899 Chr.2: 84548227-84548227 on Build GRCh38
rs768433275 Chr.2: 84548354-84548354 on Build GRCh38
rs748924683 Chr.2: 84548452-84548452 on Build GRCh38
rs753015014 Chr.2: 84548253-84548253 on Build GRCh38
rs186889514 Chr.2: 84548190-84548190 on Build GRCh38
rs755545781 Chr.2: 84548439-84548439 on Build GRCh38
rs924713746 Chr.2: 84548573-84548573 on Build GRCh38
rs902169852 Chr.2: 84548349-84548349 on Build GRCh38
rs753329575 Chr.2: 84548243-84548244 on Build GRCh38
rs982441899 Chr.2: 84548315-84548315 on Build GRCh38
rs996315605 Chr.2: 84548445-84548445 on Build GRCh38
rs781726778 Chr.2: 84548308-84548308 on Build GRCh38
rs916413614 Chr.2: 84548581-84548581 on Build GRCh38
rs1051735287 Chr.2: 84548634-84548635 on Build GRCh38
rs148515102 Chr.2: 84548372-84548372 on Build GRCh38
rs890737523 Chr.2: 84548216-84548216 on Build GRCh38
rs934633763 Chr.2: 84548597-84548597 on Build GRCh38
rs759693869 Chr.2: 84548250-84548250 on Build GRCh38
rs528982800 Chr.2: 84548446-84548446 on Build GRCh38
rs752102602 Chr.2: 84548299-84548299 on Build GRCh38
rs199761452 Chr.2: 84548249-84548249 on Build GRCh38
rs1046525243 Chr.2: 84548302-84548302 on Build GRCh38
rs547217422 Chr.2: 84548455-84548455 on Build GRCh38
rs757784873 Chr.2: 84548301-84548301 on Build GRCh38
rs758806535 Chr.2: 84548265-84548265 on Build GRCh38
rs757879883 Chr.2: 84548414-84548414 on Build GRCh38
rs766432991 Chr.2: 84548248-84548248 on Build GRCh38
rs761804506 Chr.2: 84548242-84548242 on Build GRCh38
rs372850589 Chr.2: 84548415-84548415 on Build GRCh38

View Full Product Details