Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85369558-85370058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD3
Entrez Gene ID
84173
Gene Name
ELMO domain containing 3
Gene Aliases
DFNB88, LST3, RBED1, RBM29
UniGene
Hs.269990
Gene Chromosome Location
Chr.2: 85354394-85391752 on Build GRCh38
Transcripts
NM_001135021.1 NM_001135022.1 NM_001135023.1 NM_032213.4 XM_005264596.3 XM_005264597.3 XM_005264598.3 XM_017005078.1 XM_017005079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7270712 c.360+8G>C p.? Chr.2: 85369838-85369838 on Build GRCh38
7215100 c.313A>G p.S105G Chr.2: 85369783-85369783 on Build GRCh38
3001646 c.298C>T p.Q100* Chr.2: 85369768-85369768 on Build GRCh38
7215099 c.313A>G p.S105G Chr.2: 85369783-85369783 on Build GRCh38
6467454 c.360+4T>C p.? Chr.2: 85369834-85369834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944247646 Chr.2: 85369613-85369613 on Build GRCh38
rs774846077 Chr.2: 85369776-85369778 on Build GRCh38
rs1008833256 Chr.2: 85369799-85369799 on Build GRCh38
rs376215791 Chr.2: 85369719-85369719 on Build GRCh38
rs143305924 Chr.2: 85369794-85369794 on Build GRCh38
rs550629639 Chr.2: 85369823-85369823 on Build GRCh38
rs896796575 Chr.2: 85369624-85369624 on Build GRCh38
rs888206292 Chr.2: 85369942-85369942 on Build GRCh38
rs1021596219 Chr.2: 85370030-85370030 on Build GRCh38
rs368463560 Chr.2: 85369727-85369727 on Build GRCh38
rs751033079 Chr.2: 85369873-85369873 on Build GRCh38
rs565687440 Chr.2: 85369909-85369909 on Build GRCh38
rs999611404 Chr.2: 85369585-85369585 on Build GRCh38
rs765643099 Chr.2: 85369853-85369853 on Build GRCh38
rs769769449 Chr.2: 85369788-85369788 on Build GRCh38
rs376021304 Chr.2: 85369821-85369821 on Build GRCh38
rs1048163331 Chr.2: 85369938-85369938 on Build GRCh38
rs964056108 Chr.2: 85369645-85369645 on Build GRCh38
rs139909907 Chr.2: 85369670-85369670 on Build GRCh38
rs951980893 Chr.2: 85369684-85369684 on Build GRCh38
rs1030906519 Chr.2: 85370010-85370010 on Build GRCh38
rs929638837 Chr.2: 85369913-85369913 on Build GRCh38
rs370601760 Chr.2: 85369863-85369863 on Build GRCh38
rs182130373 Chr.2: 85369686-85369686 on Build GRCh38
rs759663862 Chr.2: 85369815-85369815 on Build GRCh38
rs979113053 Chr.2: 85370018-85370018 on Build GRCh38
rs78809694 Chr.2: 85369783-85369783 on Build GRCh38
rs970675198 Chr.2: 85369717-85369717 on Build GRCh38
rs368160223 Chr.2: 85369789-85369789 on Build GRCh38
rs776906391 Chr.2: 85369835-85369835 on Build GRCh38
rs3821021 Chr.2: 85369616-85369616 on Build GRCh38
rs140138704 Chr.2: 85369767-85369767 on Build GRCh38
rs1050463981 Chr.2: 85369583-85369583 on Build GRCh38
rs766575688 Chr.2: 85369748-85369748 on Build GRCh38
rs183307680 Chr.2: 85369943-85369943 on Build GRCh38
rs779888534 Chr.2: 85369907-85369907 on Build GRCh38
rs756870288 Chr.2: 85369778-85369778 on Build GRCh38
rs947712766 Chr.2: 85369747-85369747 on Build GRCh38
rs73945721 Chr.2: 85370024-85370024 on Build GRCh38
rs771063445 Chr.2: 85369816-85369816 on Build GRCh38
rs780921189 Chr.2: 85369782-85369782 on Build GRCh38
rs951928776 Chr.2: 85369906-85369906 on Build GRCh38
rs753265000 Chr.2: 85369768-85369768 on Build GRCh38
rs201461413 Chr.2: 85369808-85369808 on Build GRCh38
rs1003492160 Chr.2: 85369987-85369987 on Build GRCh38
rs760840170 Chr.2: 85369732-85369732 on Build GRCh38
rs140453777 Chr.2: 85369812-85369812 on Build GRCh38
rs559344384 Chr.2: 85369672-85369672 on Build GRCh38
rs1026670399 Chr.2: 85369666-85369666 on Build GRCh38
rs890052562 Chr.2: 85369608-85369608 on Build GRCh38
rs192679508 Chr.2: 85369730-85369730 on Build GRCh38
rs758919811 Chr.2: 85369771-85369771 on Build GRCh38
rs188058240 Chr.2: 85369726-85369726 on Build GRCh38
rs201516145 Chr.2: 85369665-85369665 on Build GRCh38
rs775802928 Chr.2: 85369859-85369859 on Build GRCh38
rs780868527 Chr.2: 85369706-85369706 on Build GRCh38
rs778271812 Chr.2: 85369638-85369638 on Build GRCh38
rs557561806 Chr.2: 85369982-85369982 on Build GRCh38
rs906331470 Chr.2: 85369985-85369985 on Build GRCh38
rs1014868108 Chr.2: 85369942-85369942 on Build GRCh38
rs371610953 Chr.2: 85369862-85369862 on Build GRCh38
rs745597070 Chr.2: 85369710-85369710 on Build GRCh38
rs759829048 Chr.2: 85369760-85369760 on Build GRCh38
rs759744234 Chr.2: 85369838-85369838 on Build GRCh38
rs777191100 Chr.2: 85369773-85369773 on Build GRCh38
rs768595869 Chr.2: 85369796-85369796 on Build GRCh38
rs143407189 Chr.2: 85370025-85370025 on Build GRCh38
rs374016114 Chr.2: 85369868-85369868 on Build GRCh38
rs1031168985 Chr.2: 85369586-85369586 on Build GRCh38
rs576119003 Chr.2: 85370023-85370023 on Build GRCh38
rs891204818 Chr.2: 85369607-85369607 on Build GRCh38
rs199798354 Chr.2: 85369775-85369775 on Build GRCh38
rs41289949 Chr.2: 85369888-85369888 on Build GRCh38
rs62162717 Chr.2: 85369713-85369713 on Build GRCh38

View Full Product Details