Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85551294-85551799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGCX
Entrez Gene ID
2677
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
VKCFD1
UniGene
Hs.77719
Gene Chromosome Location
Chr.2: 85544720-85561534 on Build GRCh38
Transcripts
NM_000821.6 NM_001142269.3 XM_005264259.4 XM_011532764.2 XM_011532765.2 XM_017003803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1409788 c.1628T>G p.F543C Chr.2: 85551592-85551592 on Build GRCh38
1690756 c.1687G>A p.E563K Chr.2: 85551533-85551533 on Build GRCh38
1023363 c.1623G>T p.E541D Chr.2: 85551597-85551597 on Build GRCh38
4686855 c.1735delA p.M579fs*43 Chr.2: 85551485-85551485 on Build GRCh38
6835617 c.1652C>G p.T551S Chr.2: 85551568-85551568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765982989 Chr.2: 85551775-85551775 on Build GRCh38
rs764561796 Chr.2: 85551630-85551630 on Build GRCh38
rs750038705 Chr.2: 85551499-85551499 on Build GRCh38
rs1023389128 Chr.2: 85551488-85551488 on Build GRCh38
rs533941307 Chr.2: 85551736-85551736 on Build GRCh38
rs866055223 Chr.2: 85551578-85551578 on Build GRCh38
rs371085760 Chr.2: 85551766-85551766 on Build GRCh38
rs182585478 Chr.2: 85551427-85551427 on Build GRCh38
rs754910632 Chr.2: 85551569-85551569 on Build GRCh38
rs763609921 Chr.2: 85551449-85551449 on Build GRCh38
rs369103875 Chr.2: 85551636-85551636 on Build GRCh38
rs747081496 Chr.2: 85551592-85551592 on Build GRCh38
rs756773111 Chr.2: 85551474-85551474 on Build GRCh38
rs764661229 Chr.2: 85551455-85551455 on Build GRCh38
rs868590120 Chr.2: 85551740-85551740 on Build GRCh38
rs776151639 Chr.2: 85551444-85551444 on Build GRCh38
rs947143910 Chr.2: 85551584-85551584 on Build GRCh38
rs1026836371 Chr.2: 85551739-85551739 on Build GRCh38
rs774876815 Chr.2: 85551633-85551633 on Build GRCh38
rs982136375 Chr.2: 85551513-85551513 on Build GRCh38
rs368490782 Chr.2: 85551635-85551635 on Build GRCh38
rs776066132 Chr.2: 85551616-85551616 on Build GRCh38
rs749542014 Chr.2: 85551611-85551611 on Build GRCh38
rs762037951 Chr.2: 85551343-85551343 on Build GRCh38
rs952239466 Chr.2: 85551424-85551424 on Build GRCh38
rs1041007131 Chr.2: 85551700-85551700 on Build GRCh38
rs763349020 Chr.2: 85551618-85551618 on Build GRCh38
rs530359354 Chr.2: 85551404-85551404 on Build GRCh38
rs779769047 Chr.2: 85551550-85551550 on Build GRCh38
rs765437436 Chr.2: 85551440-85551440 on Build GRCh38
rs767099313 Chr.2: 85551477-85551477 on Build GRCh38
rs121909678 Chr.2: 85551548-85551548 on Build GRCh38
rs568978484 Chr.2: 85551449-85551450 on Build GRCh38
rs566828705 Chr.2: 85551674-85551674 on Build GRCh38
rs982677546 Chr.2: 85551493-85551493 on Build GRCh38
rs375606200 Chr.2: 85551605-85551605 on Build GRCh38
rs988507919 Chr.2: 85551560-85551560 on Build GRCh38
rs928807484 Chr.2: 85551536-85551536 on Build GRCh38
rs963830268 Chr.2: 85551378-85551378 on Build GRCh38
rs757621394 Chr.2: 85551457-85551459 on Build GRCh38
rs776995247 Chr.2: 85551607-85551607 on Build GRCh38
rs866846044 Chr.2: 85551426-85551426 on Build GRCh38
rs889482335 Chr.2: 85551647-85551647 on Build GRCh38
rs749076123 Chr.2: 85551551-85551551 on Build GRCh38
rs770124279 Chr.2: 85551615-85551615 on Build GRCh38
rs750090532 Chr.2: 85551652-85551652 on Build GRCh38
rs778886787 Chr.2: 85551575-85551575 on Build GRCh38
rs896978190 Chr.2: 85551703-85551703 on Build GRCh38
rs914295002 Chr.2: 85551561-85551561 on Build GRCh38
rs755908721 Chr.2: 85551500-85551500 on Build GRCh38
rs750814417 Chr.2: 85551538-85551538 on Build GRCh38
rs751073943 Chr.2: 85551459-85551459 on Build GRCh38
rs993969999 Chr.2: 85551734-85551734 on Build GRCh38
rs1026427051 Chr.2: 85551376-85551376 on Build GRCh38
rs894304281 Chr.2: 85551727-85551727 on Build GRCh38
rs766986633 Chr.2: 85551645-85551645 on Build GRCh38

View Full Product Details