Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86790651-86791150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8A
Entrez Gene ID
925
Gene Name
CD8a molecule
Gene Aliases
CD8, Leu2, MAL, p32
UniGene
Hs.85258
Gene Chromosome Location
Chr.2: 86784605-86808396 on Build GRCh38
Transcripts
NM_001145873.1 NM_001768.6 NM_171827.3 NR_027353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597535 c.50G>C p.R17P Chr.2: 86790899-86790899 on Build GRCh38
6217809 c.75G>A p.R25R Chr.2: 86790656-86790656 on Build GRCh38
6158996 c.183G>A p.R61R Chr.2: 86790671-86790671 on Build GRCh38
4670565 c.51C>T p.H17H Chr.2: 86790680-86790680 on Build GRCh38
7279525 c.164_165insT p.L55fs*169 Chr.2: 86790784-86790785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555064884 Chr.2: 86790859-86790860 on Build GRCh38
rs2944248 Chr.2: 86790710-86790710 on Build GRCh38
rs779333377 Chr.2: 86790723-86790723 on Build GRCh38
rs748395953 Chr.2: 86790724-86790724 on Build GRCh38
rs916842463 Chr.2: 86790932-86790932 on Build GRCh38
rs778945617 Chr.2: 86790825-86790825 on Build GRCh38
rs758849884 Chr.2: 86790981-86790981 on Build GRCh38
rs762602332 Chr.2: 86790759-86790759 on Build GRCh38
rs768733381 Chr.2: 86790720-86790721 on Build GRCh38
rs766551828 Chr.2: 86790788-86790788 on Build GRCh38
rs769505299 Chr.2: 86790895-86790895 on Build GRCh38
rs36227194 Chr.2: 86790866-86790866 on Build GRCh38
rs553681715 Chr.2: 86790682-86790682 on Build GRCh38
rs567647961 Chr.2: 86791012-86791012 on Build GRCh38
rs746557780 Chr.2: 86790734-86790734 on Build GRCh38
rs903849626 Chr.2: 86790987-86790987 on Build GRCh38
rs2944249 Chr.2: 86790689-86790689 on Build GRCh38
rs769152003 Chr.2: 86791085-86791085 on Build GRCh38
rs375219408 Chr.2: 86790881-86790881 on Build GRCh38
rs939495922 Chr.2: 86790928-86790928 on Build GRCh38
rs935419379 Chr.2: 86791026-86791026 on Build GRCh38
rs142374955 Chr.2: 86790842-86790842 on Build GRCh38
rs979496521 Chr.2: 86790997-86790997 on Build GRCh38
rs763179704 Chr.2: 86790925-86790925 on Build GRCh38
rs757772155 Chr.2: 86791036-86791036 on Build GRCh38
rs951166741 Chr.2: 86790902-86790902 on Build GRCh38
rs764996911 Chr.2: 86790669-86790669 on Build GRCh38
rs1038067695 Chr.2: 86791109-86791109 on Build GRCh38
rs1052668153 Chr.2: 86790840-86790840 on Build GRCh38
rs762732617 Chr.2: 86790745-86790746 on Build GRCh38
rs756309877 Chr.2: 86790784-86790784 on Build GRCh38
rs755449765 Chr.2: 86790715-86790715 on Build GRCh38
rs1001238999 Chr.2: 86790875-86790875 on Build GRCh38
rs776525176 Chr.2: 86790721-86790734 on Build GRCh38
rs868233441 Chr.2: 86790777-86790777 on Build GRCh38
rs562063776 Chr.2: 86790698-86790698 on Build GRCh38
rs753982338 Chr.2: 86790916-86790916 on Build GRCh38
rs759178072 Chr.2: 86790745-86790745 on Build GRCh38
rs775365870 Chr.2: 86790752-86790752 on Build GRCh38
rs745410119 Chr.2: 86791069-86791069 on Build GRCh38
rs955282672 Chr.2: 86790906-86790906 on Build GRCh38
rs2944247 Chr.2: 86790711-86790711 on Build GRCh38
rs926956723 Chr.2: 86791077-86791077 on Build GRCh38
rs935512606 Chr.2: 86790793-86790793 on Build GRCh38
rs769759152 Chr.2: 86790723-86790733 on Build GRCh38
rs752128882 Chr.2: 86791033-86791033 on Build GRCh38
rs776318667 Chr.2: 86790740-86790740 on Build GRCh38
rs531213531 Chr.2: 86790690-86790690 on Build GRCh38
rs765738788 Chr.2: 86790752-86790753 on Build GRCh38
rs773543693 Chr.2: 86790731-86790731 on Build GRCh38
rs190362172 Chr.2: 86790967-86790967 on Build GRCh38
rs999604488 Chr.2: 86790790-86790790 on Build GRCh38
rs908060166 Chr.2: 86790904-86790904 on Build GRCh38
rs200750291 Chr.2: 86790780-86790780 on Build GRCh38
rs772724090 Chr.2: 86790730-86790731 on Build GRCh38
rs758358202 Chr.2: 86790680-86790680 on Build GRCh38
rs987124060 Chr.2: 86790931-86790931 on Build GRCh38
rs964073700 Chr.2: 86790973-86790973 on Build GRCh38
rs559739267 Chr.2: 86790859-86790859 on Build GRCh38
rs768312279 Chr.2: 86790898-86790898 on Build GRCh38
rs935404610 Chr.2: 86791002-86791002 on Build GRCh38
rs1045371693 Chr.2: 86790862-86790862 on Build GRCh38
rs752835045 Chr.2: 86790974-86790974 on Build GRCh38
rs764119617 Chr.2: 86790760-86790760 on Build GRCh38
rs770400583 Chr.2: 86790736-86790736 on Build GRCh38
rs36226877 Chr.2: 86791073-86791073 on Build GRCh38
rs551587694 Chr.2: 86790877-86790877 on Build GRCh38
rs752636744 Chr.2: 86790671-86790671 on Build GRCh38
rs767497169 Chr.2: 86790911-86790911 on Build GRCh38
rs761836461 Chr.2: 86790902-86790902 on Build GRCh38
rs770488602 Chr.2: 86790868-86790868 on Build GRCh38
rs764727034 Chr.2: 86790751-86790751 on Build GRCh38
rs747917576 Chr.2: 86790721-86790727 on Build GRCh38
rs763876643 Chr.2: 86790687-86790687 on Build GRCh38
rs36227196 Chr.2: 86791055-86791055 on Build GRCh38
rs1044151528 Chr.2: 86790979-86790979 on Build GRCh38
rs759486070 Chr.2: 86790926-86790926 on Build GRCh38
rs772647566 Chr.2: 86790729-86790729 on Build GRCh38
rs972617795 Chr.2: 86790700-86790700 on Build GRCh38
rs949680466 Chr.2: 86790844-86790844 on Build GRCh38
rs753127935 Chr.2: 86790830-86790830 on Build GRCh38
rs756222034 Chr.2: 86790691-86790691 on Build GRCh38
rs774961230 Chr.2: 86790896-86790896 on Build GRCh38
rs779805078 Chr.2: 86791112-86791112 on Build GRCh38
rs755257543 Chr.2: 86790808-86790808 on Build GRCh38
rs1039517899 Chr.2: 86791086-86791086 on Build GRCh38
rs918087813 Chr.2: 86791053-86791053 on Build GRCh38
rs762845623 Chr.2: 86790897-86790897 on Build GRCh38
rs36227195 Chr.2: 86790899-86790899 on Build GRCh38
rs758650184 Chr.2: 86790835-86790835 on Build GRCh38
rs754177445 Chr.2: 86790789-86790789 on Build GRCh38
rs542978694 Chr.2: 86790697-86790697 on Build GRCh38
rs535787896 Chr.2: 86791051-86791051 on Build GRCh38
rs937037375 Chr.2: 86791108-86791108 on Build GRCh38

View Full Product Details