Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86822216-86822754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8B
Entrez Gene ID
926
Gene Name
CD8b molecule
Gene Aliases
CD8B1, LEU2, LY3, LYT3, P37
UniGene
Hs.405667
Gene Chromosome Location
Chr.2: 86815337-86861924 on Build GRCh38
Transcripts
NM_001178100.1 NM_172101.3 NM_172102.3 NM_172213.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630809 c.630A>G p.L210L Chr.2: 86822365-86822365 on Build GRCh38
341912 c.621-1G>C p.? Chr.2: 86822375-86822375 on Build GRCh38
5553034 c.640G>T p.E214* Chr.2: 86822355-86822355 on Build GRCh38
3840195 c.653G>C p.R218T Chr.2: 86822342-86822342 on Build GRCh38
5957185 c.640G>C p.E214Q Chr.2: 86822355-86822355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189858788 Chr.2: 86822512-86822512 on Build GRCh38
rs568650339 Chr.2: 86822627-86822627 on Build GRCh38
rs185150031 Chr.2: 86822274-86822274 on Build GRCh38
rs979558262 Chr.2: 86822578-86822578 on Build GRCh38
rs555961390 Chr.2: 86822255-86822255 on Build GRCh38
rs751339636 Chr.2: 86822370-86822370 on Build GRCh38
rs1037795161 Chr.2: 86822409-86822409 on Build GRCh38
rs757043155 Chr.2: 86822382-86822382 on Build GRCh38
rs954969432 Chr.2: 86822529-86822529 on Build GRCh38
rs894213628 Chr.2: 86822294-86822294 on Build GRCh38
rs781036151 Chr.2: 86822385-86822385 on Build GRCh38
rs942074985 Chr.2: 86822697-86822697 on Build GRCh38
rs547462808 Chr.2: 86822658-86822658 on Build GRCh38
rs114490030 Chr.2: 86822573-86822573 on Build GRCh38
rs955590009 Chr.2: 86822461-86822461 on Build GRCh38
rs979222675 Chr.2: 86822279-86822279 on Build GRCh38
rs538310032 Chr.2: 86822460-86822460 on Build GRCh38
rs117448899 Chr.2: 86822320-86822320 on Build GRCh38
rs767657960 Chr.2: 86822297-86822297 on Build GRCh38
rs552497747 Chr.2: 86822268-86822268 on Build GRCh38
rs754152123 Chr.2: 86822410-86822410 on Build GRCh38
rs768789467 Chr.2: 86822391-86822391 on Build GRCh38
rs113049563 Chr.2: 86822681-86822681 on Build GRCh38
rs956745380 Chr.2: 86822621-86822621 on Build GRCh38
rs764511442 Chr.2: 86822412-86822413 on Build GRCh38
rs764745206 Chr.2: 86822530-86822530 on Build GRCh38
rs758130558 Chr.2: 86822328-86822328 on Build GRCh38
rs911329494 Chr.2: 86822510-86822510 on Build GRCh38
rs768695442 Chr.2: 86822285-86822285 on Build GRCh38
rs949277563 Chr.2: 86822646-86822646 on Build GRCh38
rs926579869 Chr.2: 86822692-86822692 on Build GRCh38
rs774612892 Chr.2: 86822290-86822290 on Build GRCh38
rs757833423 Chr.2: 86822551-86822551 on Build GRCh38
rs978750969 Chr.2: 86822444-86822444 on Build GRCh38
rs1047456996 Chr.2: 86822665-86822665 on Build GRCh38
rs906903967 Chr.2: 86822246-86822246 on Build GRCh38
rs917821483 Chr.2: 86822645-86822645 on Build GRCh38
rs748166265 Chr.2: 86822403-86822403 on Build GRCh38
rs768991715 Chr.2: 86822474-86822474 on Build GRCh38
rs1045075558 Chr.2: 86822336-86822336 on Build GRCh38
rs200594911 Chr.2: 86822302-86822302 on Build GRCh38
rs745787914 Chr.2: 86822389-86822389 on Build GRCh38
rs897978354 Chr.2: 86822449-86822449 on Build GRCh38
rs754865738 Chr.2: 86822584-86822584 on Build GRCh38
rs370157479 Chr.2: 86822544-86822544 on Build GRCh38
rs752468668 Chr.2: 86822326-86822326 on Build GRCh38
rs761052893 Chr.2: 86822303-86822303 on Build GRCh38
rs182563312 Chr.2: 86822575-86822575 on Build GRCh38
rs368173321 Chr.2: 86822317-86822317 on Build GRCh38
rs760987777 Chr.2: 86822424-86822424 on Build GRCh38
rs749365534 Chr.2: 86822281-86822281 on Build GRCh38
rs773445818 Chr.2: 86822412-86822412 on Build GRCh38
rs1007827408 Chr.2: 86822545-86822545 on Build GRCh38
rs375100155 Chr.2: 86822293-86822293 on Build GRCh38
rs560253502 Chr.2: 86822592-86822592 on Build GRCh38
rs1022485488 Chr.2: 86822305-86822305 on Build GRCh38
rs905207361 Chr.2: 86822373-86822373 on Build GRCh38
rs907105572 Chr.2: 86822683-86822683 on Build GRCh38
rs370723539 Chr.2: 86822399-86822399 on Build GRCh38
rs545810678 Chr.2: 86822364-86822364 on Build GRCh38

View Full Product Details