Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86844530-86845038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8B
Entrez Gene ID
926
Gene Name
CD8b molecule
Gene Aliases
CD8B1, LEU2, LY3, LYT3, P37
UniGene
Hs.405667
Gene Chromosome Location
Chr.2: 86815337-86861924 on Build GRCh38
Transcripts
NM_001178100.1 NM_004931.4 NM_172101.3 NM_172102.3 NM_172213.3 XM_011533164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5502497 c.82C>T p.L28L Chr.2: 86844643-86844643 on Build GRCh38
4808929 c.81-7A>T p.? Chr.2: 86844651-86844651 on Build GRCh38
4758484 c.67C>T p.R23C Chr.2: 86844935-86844935 on Build GRCh38
225997 c.590G>A p.R197Q Chr.2: 86844952-86844952 on Build GRCh38
4758482 c.607C>T p.R203C Chr.2: 86844935-86844935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761177816 Chr.2: 86844916-86844916 on Build GRCh38
rs534704182 Chr.2: 86844871-86844871 on Build GRCh38
rs879200713 Chr.2: 86844573-86844573 on Build GRCh38
rs376413559 Chr.2: 86844883-86844883 on Build GRCh38
rs776195149 Chr.2: 86844879-86844879 on Build GRCh38
rs73947279 Chr.2: 86844668-86844668 on Build GRCh38
rs766345301 Chr.2: 86844631-86844631 on Build GRCh38
rs191596977 Chr.2: 86844764-86844764 on Build GRCh38
rs773063562 Chr.2: 86844982-86844985 on Build GRCh38
rs935700824 Chr.2: 86844563-86844563 on Build GRCh38
rs746071107 Chr.2: 86844965-86844965 on Build GRCh38
rs369409124 Chr.2: 86844979-86844979 on Build GRCh38
rs767229548 Chr.2: 86844920-86844920 on Build GRCh38
rs979798127 Chr.2: 86844839-86844839 on Build GRCh38
rs867165384 Chr.2: 86844732-86844732 on Build GRCh38
rs777905487 Chr.2: 86844662-86844662 on Build GRCh38
rs867396401 Chr.2: 86844819-86844819 on Build GRCh38
rs766148662 Chr.2: 86844937-86844937 on Build GRCh38
rs970338870 Chr.2: 86844807-86844807 on Build GRCh38
rs575741141 Chr.2: 86844798-86844798 on Build GRCh38
rs370644609 Chr.2: 86845008-86845008 on Build GRCh38
rs142328928 Chr.2: 86844955-86844955 on Build GRCh38
rs769706834 Chr.2: 86844879-86844880 on Build GRCh38
rs867221419 Chr.2: 86844926-86844926 on Build GRCh38
rs746268761 Chr.2: 86844870-86844870 on Build GRCh38
rs896979651 Chr.2: 86844760-86844760 on Build GRCh38
rs897219748 Chr.2: 86844956-86844956 on Build GRCh38
rs751687141 Chr.2: 86844953-86844953 on Build GRCh38
rs528519899 Chr.2: 86844669-86844669 on Build GRCh38
rs1032709587 Chr.2: 86845015-86845015 on Build GRCh38
rs772821597 Chr.2: 86844922-86844922 on Build GRCh38
rs565267006 Chr.2: 86844698-86844698 on Build GRCh38
rs567412420 Chr.2: 86844845-86844845 on Build GRCh38
rs563876847 Chr.2: 86844767-86844767 on Build GRCh38
rs773924473 Chr.2: 86844915-86844915 on Build GRCh38
rs552731173 Chr.2: 86844872-86844872 on Build GRCh38
rs551674183 Chr.2: 86844707-86844707 on Build GRCh38
rs1050157914 Chr.2: 86844664-86844664 on Build GRCh38
rs549150361 Chr.2: 86844803-86844803 on Build GRCh38
rs74262969 Chr.2: 86844651-86844651 on Build GRCh38
rs758587941 Chr.2: 86844651-86844651 on Build GRCh38
rs771411407 Chr.2: 86844779-86844779 on Build GRCh38
rs151294448 Chr.2: 86844934-86844934 on Build GRCh38
rs540316799 Chr.2: 86844680-86844680 on Build GRCh38
rs957133757 Chr.2: 86844851-86844851 on Build GRCh38
rs1015405057 Chr.2: 86844754-86844754 on Build GRCh38
rs977285940 Chr.2: 86844906-86844906 on Build GRCh38
rs988549373 Chr.2: 86844855-86844855 on Build GRCh38
rs749555970 Chr.2: 86845004-86845004 on Build GRCh38
rs868292878 Chr.2: 86844714-86844714 on Build GRCh38
rs533104573 Chr.2: 86844706-86844706 on Build GRCh38
rs185702450 Chr.2: 86844597-86844597 on Build GRCh38
rs1005589024 Chr.2: 86844739-86844739 on Build GRCh38
rs781518011 Chr.2: 86844685-86844685 on Build GRCh38
rs753839701 Chr.2: 86844938-86844938 on Build GRCh38
rs370737681 Chr.2: 86844704-86844704 on Build GRCh38
rs780567389 Chr.2: 86844980-86844980 on Build GRCh38
rs932672302 Chr.2: 86844661-86844661 on Build GRCh38
rs768974293 Chr.2: 86844905-86844905 on Build GRCh38
rs758466427 Chr.2: 86844659-86844659 on Build GRCh38
rs145406454 Chr.2: 86844950-86844950 on Build GRCh38
rs914056597 Chr.2: 86844602-86844602 on Build GRCh38
rs765031706 Chr.2: 86844952-86844952 on Build GRCh38
rs368299070 Chr.2: 86844891-86844891 on Build GRCh38
rs781250481 Chr.2: 86844650-86844651 on Build GRCh38
rs374072931 Chr.2: 86844853-86844853 on Build GRCh38
rs901398165 Chr.2: 86844652-86844652 on Build GRCh38
rs932889775 Chr.2: 86844935-86844935 on Build GRCh38
rs769227047 Chr.2: 86845009-86845009 on Build GRCh38

View Full Product Details