Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86846218-86846714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD8B
Entrez Gene ID
926
Gene Name
CD8b molecule
Gene Aliases
CD8B1, LEU2, LY3, LYT3, P37
UniGene
Hs.405667
Gene Chromosome Location
Chr.2: 86815337-86861924 on Build GRCh38
Transcripts
NM_001178100.1 NM_004931.4 NM_172101.3 NM_172102.3 NM_172213.3 XM_011533164.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6092456 c.563G>T p.G188V Chr.2: 86846704-86846704 on Build GRCh38
1136739 c.573C>G p.I191M Chr.2: 86846694-86846694 on Build GRCh38
477725 c.573C>G p.I191M Chr.2: 86846694-86846694 on Build GRCh38
6092455 c.563G>T p.G188V Chr.2: 86846704-86846704 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005921801 Chr.2: 86846523-86846523 on Build GRCh38
rs538346835 Chr.2: 86846338-86846338 on Build GRCh38
rs898682399 Chr.2: 86846584-86846584 on Build GRCh38
rs373336316 Chr.2: 86846645-86846645 on Build GRCh38
rs868856862 Chr.2: 86846285-86846285 on Build GRCh38
rs1050058299 Chr.2: 86846516-86846516 on Build GRCh38
rs4971887 Chr.2: 86846620-86846620 on Build GRCh38
rs1003321045 Chr.2: 86846634-86846634 on Build GRCh38
rs749886822 Chr.2: 86846612-86846612 on Build GRCh38
rs192831627 Chr.2: 86846340-86846340 on Build GRCh38
rs186971649 Chr.2: 86846334-86846334 on Build GRCh38
rs771549269 Chr.2: 86846653-86846653 on Build GRCh38
rs914391180 Chr.2: 86846311-86846311 on Build GRCh38
rs750104710 Chr.2: 86846519-86846519 on Build GRCh38
rs1042038169 Chr.2: 86846461-86846461 on Build GRCh38
rs1025869187 Chr.2: 86846240-86846240 on Build GRCh38
rs1126874 Chr.2: 86846694-86846694 on Build GRCh38
rs4971892 Chr.2: 86846522-86846522 on Build GRCh38
rs182353204 Chr.2: 86846286-86846286 on Build GRCh38
rs139322949 Chr.2: 86846504-86846504 on Build GRCh38
rs573405954 Chr.2: 86846254-86846254 on Build GRCh38
rs937401117 Chr.2: 86846281-86846281 on Build GRCh38
rs575641181 Chr.2: 86846558-86846558 on Build GRCh38
rs779248365 Chr.2: 86846635-86846635 on Build GRCh38
rs1026321634 Chr.2: 86846625-86846625 on Build GRCh38
rs971824532 Chr.2: 86846245-86846245 on Build GRCh38
rs760318358 Chr.2: 86846683-86846683 on Build GRCh38
rs971639249 Chr.2: 86846626-86846626 on Build GRCh38
rs765632842 Chr.2: 86846325-86846326 on Build GRCh38
rs570920228 Chr.2: 86846288-86846288 on Build GRCh38
rs562857903 Chr.2: 86846532-86846532 on Build GRCh38
rs773974148 Chr.2: 86846329-86846330 on Build GRCh38
rs368510294 Chr.2: 86846297-86846297 on Build GRCh38
rs901736215 Chr.2: 86846474-86846474 on Build GRCh38
rs138398564 Chr.2: 86846657-86846657 on Build GRCh38
rs981429238 Chr.2: 86846252-86846252 on Build GRCh38
rs761514443 Chr.2: 86846333-86846333 on Build GRCh38
rs932983142 Chr.2: 86846478-86846478 on Build GRCh38

View Full Product Details