Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88525396-88525836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX37
Entrez Gene ID
200523
Gene Name
testis expressed 37
Gene Aliases
C2orf51, TSC21
UniGene
Hs.132104
Gene Chromosome Location
Chr.2: 88524623-88529585 on Build GRCh38
Transcripts
NM_152670.2 XM_005264188.3 XM_011532691.1 XM_011532692.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3910753 c.18C>T p.Y6Y Chr.2: 88525662-88525662 on Build GRCh38
6800310 c.4G>C p.A2P Chr.2: 88525648-88525648 on Build GRCh38
319077 c.10G>A p.V4M Chr.2: 88525654-88525654 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918219447 Chr.2: 88525764-88525764 on Build GRCh38
rs747014480 Chr.2: 88525625-88525625 on Build GRCh38
rs760002964 Chr.2: 88525624-88525624 on Build GRCh38
rs751313464 Chr.2: 88525648-88525648 on Build GRCh38
rs1002134425 Chr.2: 88525539-88525539 on Build GRCh38
rs768503033 Chr.2: 88525625-88525626 on Build GRCh38
rs970358470 Chr.2: 88525522-88525522 on Build GRCh38
rs985572841 Chr.2: 88525499-88525499 on Build GRCh38
rs769207786 Chr.2: 88525720-88525720 on Build GRCh38
rs976285508 Chr.2: 88525482-88525482 on Build GRCh38
rs750348762 Chr.2: 88525665-88525665 on Build GRCh38
rs929888814 Chr.2: 88525545-88525545 on Build GRCh38
rs561067750 Chr.2: 88525799-88525799 on Build GRCh38
rs1001831470 Chr.2: 88525741-88525741 on Build GRCh38
rs1053583586 Chr.2: 88525455-88525455 on Build GRCh38
rs1033980428 Chr.2: 88525544-88525544 on Build GRCh38
rs747708605 Chr.2: 88525698-88525703 on Build GRCh38
rs781151725 Chr.2: 88525650-88525651 on Build GRCh38
rs371985758 Chr.2: 88525502-88525502 on Build GRCh38
rs772315735 Chr.2: 88525602-88525602 on Build GRCh38
rs879099869 Chr.2: 88525586-88525586 on Build GRCh38
rs762599468 Chr.2: 88525638-88525638 on Build GRCh38
rs62159803 Chr.2: 88525686-88525686 on Build GRCh38
rs749036831 Chr.2: 88525798-88525798 on Build GRCh38
rs758561576 Chr.2: 88525685-88525685 on Build GRCh38
rs1011605801 Chr.2: 88525662-88525662 on Build GRCh38
rs34724508 Chr.2: 88525496-88525496 on Build GRCh38
rs775748607 Chr.2: 88525637-88525637 on Build GRCh38
rs775864753 Chr.2: 88525708-88525708 on Build GRCh38
rs906206781 Chr.2: 88525740-88525740 on Build GRCh38
rs560102367 Chr.2: 88525798-88525799 on Build GRCh38
rs771058676 Chr.2: 88525612-88525612 on Build GRCh38
rs878967739 Chr.2: 88525798-88525799 on Build GRCh38
rs577813909 Chr.2: 88525664-88525664 on Build GRCh38
rs551571556 Chr.2: 88525442-88525442 on Build GRCh38
rs910906632 Chr.2: 88525500-88525500 on Build GRCh38
rs780616725 Chr.2: 88525566-88525566 on Build GRCh38
rs929898870 Chr.2: 88525789-88525789 on Build GRCh38
rs184110782 Chr.2: 88525593-88525593 on Build GRCh38
rs779700734 Chr.2: 88525471-88525471 on Build GRCh38
rs1033641258 Chr.2: 88525748-88525748 on Build GRCh38
rs191189330 Chr.2: 88525460-88525460 on Build GRCh38
rs777192277 Chr.2: 88525614-88525614 on Build GRCh38
rs74663053 Chr.2: 88525797-88525800 on Build GRCh38
rs570141863 Chr.2: 88525456-88525456 on Build GRCh38
rs763395095 Chr.2: 88525711-88525711 on Build GRCh38
rs374816585 Chr.2: 88525663-88525663 on Build GRCh38
rs770172792 Chr.2: 88525635-88525635 on Build GRCh38
rs567586678 Chr.2: 88525552-88525552 on Build GRCh38
rs1024629604 Chr.2: 88525508-88525508 on Build GRCh38
rs368597509 Chr.2: 88525716-88525716 on Build GRCh38
rs373883734 Chr.2: 88525743-88525743 on Build GRCh38
rs560922527 Chr.2: 88525798-88525798 on Build GRCh38
rs34809896 Chr.2: 88525811-88525812 on Build GRCh38
rs753583503 Chr.2: 88525677-88525677 on Build GRCh38
rs201974018 Chr.2: 88525598-88525598 on Build GRCh38
rs374413211 Chr.2: 88525703-88525703 on Build GRCh38
rs933762305 Chr.2: 88525498-88525498 on Build GRCh38
rs761401624 Chr.2: 88525652-88525652 on Build GRCh38
rs899069075 Chr.2: 88525749-88525749 on Build GRCh38
rs973744006 Chr.2: 88525745-88525745 on Build GRCh38
rs763851340 Chr.2: 88525644-88525644 on Build GRCh38
rs371925857 Chr.2: 88525698-88525698 on Build GRCh38
rs1020574997 Chr.2: 88525443-88525443 on Build GRCh38
rs990416790 Chr.2: 88525438-88525438 on Build GRCh38
rs569544611 Chr.2: 88525799-88525800 on Build GRCh38
rs994708474 Chr.2: 88525755-88525755 on Build GRCh38
rs770206809 Chr.2: 88525706-88525706 on Build GRCh38
rs779862356 Chr.2: 88525676-88525676 on Build GRCh38
rs1045551147 Chr.2: 88525569-88525569 on Build GRCh38
rs1026546983 Chr.2: 88525758-88525758 on Build GRCh38
rs373147477 Chr.2: 88525582-88525582 on Build GRCh38
rs748064098 Chr.2: 88525576-88525576 on Build GRCh38
rs769789212 Chr.2: 88525623-88525623 on Build GRCh38
rs556741219 Chr.2: 88525702-88525702 on Build GRCh38
rs747107273 Chr.2: 88525610-88525610 on Build GRCh38
rs756023928 Chr.2: 88525673-88525673 on Build GRCh38
rs940995006 Chr.2: 88525501-88525501 on Build GRCh38
rs777766619 Chr.2: 88525607-88525607 on Build GRCh38

View Full Product Details