Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95410764-95411260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAHD2A
Entrez Gene ID
51011
Gene Name
fumarylacetoacetate hydrolase domain containing 2A
Gene Aliases
CGI-105
UniGene
Hs.531247
Gene Chromosome Location
Chr.2: 95402691-95416702 on Build GRCh38
Transcripts
NM_016044.2 XM_011511282.2 XM_011511283.2 XM_011511284.2 XM_011511285.2 XM_011511286.2 XM_011511287.2 XM_011511288.2 XM_011511289.2 XM_011511290.2 XM_017004236.1 XM_017004237.1 XM_017004238.1 XM_017004239.1 XM_017004240.1 XM_017004241.1 XM_017004242.1 XM_017004243.1 XM_017004244.1 XM_017004245.1 XM_017004246.1 XM_017004247.1 XM_017004248.1 XM_017004249.1 XM_017004250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657988 c.541C>T p.H181Y Chr.2: 95410882-95410882 on Build GRCh38
5021308 c.580C>T p.R194C Chr.2: 95410921-95410921 on Build GRCh38
4096314 c.581G>A p.R194H Chr.2: 95410922-95410922 on Build GRCh38
1409992 c.633C>T p.F211F Chr.2: 95410974-95410974 on Build GRCh38
5059119 c.571G>T p.V191L Chr.2: 95410912-95410912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546817338 Chr.2: 95410916-95410916 on Build GRCh38
rs771772969 Chr.2: 95410914-95410914 on Build GRCh38
rs769273179 Chr.2: 95410831-95410833 on Build GRCh38
rs60804719 Chr.2: 95410971-95410971 on Build GRCh38
rs771572341 Chr.2: 95410832-95410832 on Build GRCh38
rs1053076583 Chr.2: 95411122-95411122 on Build GRCh38
rs756885595 Chr.2: 95411043-95411043 on Build GRCh38
rs1814305 Chr.2: 95410822-95410822 on Build GRCh38
rs3860489 Chr.2: 95410789-95410789 on Build GRCh38
rs964858096 Chr.2: 95410966-95410966 on Build GRCh38
rs768303991 Chr.2: 95411056-95411056 on Build GRCh38
rs934652928 Chr.2: 95411116-95411116 on Build GRCh38
rs780713910 Chr.2: 95411044-95411044 on Build GRCh38
rs554582796 Chr.2: 95411051-95411051 on Build GRCh38
rs894584573 Chr.2: 95410825-95410825 on Build GRCh38
rs760845347 Chr.2: 95410969-95410969 on Build GRCh38
rs536395913 Chr.2: 95410888-95410888 on Build GRCh38
rs1045335 Chr.2: 95410941-95410941 on Build GRCh38
rs755439238 Chr.2: 95410907-95410907 on Build GRCh38
rs1045337 Chr.2: 95410953-95410953 on Build GRCh38
rs753166477 Chr.2: 95410999-95410999 on Build GRCh38
rs750604619 Chr.2: 95410967-95410967 on Build GRCh38
rs1043473186 Chr.2: 95410846-95410846 on Build GRCh38
rs764994730 Chr.2: 95410874-95410874 on Build GRCh38
rs780676808 Chr.2: 95410891-95410891 on Build GRCh38
rs1017443793 Chr.2: 95410940-95410940 on Build GRCh38
rs767567489 Chr.2: 95410884-95410884 on Build GRCh38
rs201416966 Chr.2: 95410909-95410909 on Build GRCh38
rs150916987 Chr.2: 95410977-95410977 on Build GRCh38
rs754363054 Chr.2: 95410906-95410906 on Build GRCh38
rs778744172 Chr.2: 95410823-95410823 on Build GRCh38
rs890414297 Chr.2: 95411144-95411144 on Build GRCh38
rs755571148 Chr.2: 95410975-95410975 on Build GRCh38
rs537832044 Chr.2: 95411048-95411048 on Build GRCh38
rs370293938 Chr.2: 95411058-95411058 on Build GRCh38
rs192125504 Chr.2: 95411079-95411079 on Build GRCh38
rs1045330 Chr.2: 95410931-95410931 on Build GRCh38
rs759494586 Chr.2: 95410924-95410924 on Build GRCh38
rs776548589 Chr.2: 95410860-95410860 on Build GRCh38
rs138025378 Chr.2: 95410912-95410912 on Build GRCh38
rs772957599 Chr.2: 95410939-95410939 on Build GRCh38
rs867389679 Chr.2: 95411236-95411236 on Build GRCh38
rs563016350 Chr.2: 95411210-95411210 on Build GRCh38
rs140892236 Chr.2: 95410868-95410868 on Build GRCh38
rs200799023 Chr.2: 95410974-95410974 on Build GRCh38
rs2320435 Chr.2: 95410804-95410804 on Build GRCh38
rs199659432 Chr.2: 95410962-95410962 on Build GRCh38
rs995051527 Chr.2: 95411212-95411212 on Build GRCh38
rs575517943 Chr.2: 95411003-95411003 on Build GRCh38
rs745318280 Chr.2: 95411045-95411045 on Build GRCh38
rs1045332 Chr.2: 95410934-95410934 on Build GRCh38
rs904580162 Chr.2: 95411102-95411102 on Build GRCh38
rs762127494 Chr.2: 95410876-95410876 on Build GRCh38
rs141385169 Chr.2: 95410875-95410875 on Build GRCh38
rs746621447 Chr.2: 95411033-95411033 on Build GRCh38
rs2874267 Chr.2: 95411139-95411139 on Build GRCh38
rs762039821 Chr.2: 95411017-95411017 on Build GRCh38
rs769681317 Chr.2: 95410926-95410926 on Build GRCh38
rs540282029 Chr.2: 95411078-95411078 on Build GRCh38
rs938855672 Chr.2: 95410809-95410809 on Build GRCh38
rs758690715 Chr.2: 95411006-95411006 on Build GRCh38
rs770963609 Chr.2: 95410857-95410857 on Build GRCh38
rs546397810 Chr.2: 95411148-95411148 on Build GRCh38
rs566500770 Chr.2: 95410921-95410921 on Build GRCh38
rs187604324 Chr.2: 95411061-95411061 on Build GRCh38
rs772786422 Chr.2: 95410837-95410837 on Build GRCh38
rs770319428 Chr.2: 95410922-95410922 on Build GRCh38
rs183839404 Chr.2: 95411097-95411097 on Build GRCh38
rs1017878532 Chr.2: 95411181-95411181 on Build GRCh38
rs948844783 Chr.2: 95411095-95411095 on Build GRCh38
rs369791868 Chr.2: 95410824-95410824 on Build GRCh38
rs983038698 Chr.2: 95410979-95410979 on Build GRCh38
rs775267251 Chr.2: 95410930-95410930 on Build GRCh38
rs749174031 Chr.2: 95411053-95411053 on Build GRCh38
rs866828561 Chr.2: 95411131-95411131 on Build GRCh38
rs374507627 Chr.2: 95410890-95410890 on Build GRCh38
rs777526179 Chr.2: 95411031-95411031 on Build GRCh38
rs760048250 Chr.2: 95410852-95410852 on Build GRCh38
rs1045333 Chr.2: 95410935-95410935 on Build GRCh38

View Full Product Details