Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96132418-96132916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
UniGene
Hs.447993
Gene Chromosome Location
Chr.2: 96122817-96143059 on Build GRCh38
Transcripts
NM_001002036.3 XM_011511205.2 XM_011511206.2 XM_011511207.2 XM_011511208.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259526 c.507G>A p.A169A Chr.2: 96132670-96132670 on Build GRCh38
227435 c.625G>A p.E209K Chr.2: 96132552-96132552 on Build GRCh38
6159315 c.467G>C p.S156T Chr.2: 96132710-96132710 on Build GRCh38
3584061 c.501C>T p.S167S Chr.2: 96132676-96132676 on Build GRCh38
6159316 c.590C>A p.A197D Chr.2: 96132587-96132587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375008194 Chr.2: 96132601-96132601 on Build GRCh38
rs78275861 Chr.2: 96132712-96132712 on Build GRCh38
rs976753473 Chr.2: 96132469-96132469 on Build GRCh38
rs761136005 Chr.2: 96132642-96132642 on Build GRCh38
rs752897981 Chr.2: 96132499-96132499 on Build GRCh38
rs944963077 Chr.2: 96132879-96132879 on Build GRCh38
rs528556110 Chr.2: 96132768-96132769 on Build GRCh38
rs748724913 Chr.2: 96132569-96132569 on Build GRCh38
rs372867360 Chr.2: 96132751-96132751 on Build GRCh38
rs536523896 Chr.2: 96132885-96132885 on Build GRCh38
rs138165928 Chr.2: 96132671-96132671 on Build GRCh38
rs779697688 Chr.2: 96132526-96132526 on Build GRCh38
rs763746689 Chr.2: 96132742-96132742 on Build GRCh38
rs375060410 Chr.2: 96132763-96132763 on Build GRCh38
rs772118993 Chr.2: 96132553-96132553 on Build GRCh38
rs377277711 Chr.2: 96132489-96132489 on Build GRCh38
rs751579283 Chr.2: 96132579-96132579 on Build GRCh38
rs376251705 Chr.2: 96132505-96132505 on Build GRCh38
rs768755515 Chr.2: 96132614-96132614 on Build GRCh38
rs767533400 Chr.2: 96132636-96132636 on Build GRCh38
rs1010238716 Chr.2: 96132687-96132687 on Build GRCh38
rs766783182 Chr.2: 96132647-96132647 on Build GRCh38
rs752138502 Chr.2: 96132517-96132517 on Build GRCh38
rs1168966 Chr.2: 96132771-96132771 on Build GRCh38
rs766795734 Chr.2: 96132709-96132709 on Build GRCh38
rs139152188 Chr.2: 96132617-96132617 on Build GRCh38
rs778436747 Chr.2: 96132664-96132664 on Build GRCh38
rs148818932 Chr.2: 96132580-96132580 on Build GRCh38
rs367836103 Chr.2: 96132473-96132473 on Build GRCh38
rs368781391 Chr.2: 96132591-96132591 on Build GRCh38
rs151115044 Chr.2: 96132582-96132582 on Build GRCh38
rs754143947 Chr.2: 96132648-96132648 on Build GRCh38
rs1034857703 Chr.2: 96132766-96132766 on Build GRCh38
rs765548772 Chr.2: 96132493-96132493 on Build GRCh38
rs541100535 Chr.2: 96132521-96132521 on Build GRCh38
rs201337537 Chr.2: 96132549-96132549 on Build GRCh38
rs780360443 Chr.2: 96132583-96132583 on Build GRCh38
rs747604715 Chr.2: 96132592-96132592 on Build GRCh38
rs142432881 Chr.2: 96132734-96132734 on Build GRCh38
rs983151067 Chr.2: 96132822-96132822 on Build GRCh38
rs976246062 Chr.2: 96132892-96132892 on Build GRCh38
rs764416739 Chr.2: 96132662-96132662 on Build GRCh38
rs757683346 Chr.2: 96132519-96132519 on Build GRCh38
rs115782187 Chr.2: 96132490-96132490 on Build GRCh38
rs750956004 Chr.2: 96132640-96132640 on Build GRCh38
rs748161965 Chr.2: 96132552-96132552 on Build GRCh38
rs41320144 Chr.2: 96132566-96132566 on Build GRCh38
rs371727585 Chr.2: 96132594-96132594 on Build GRCh38
rs143172084 Chr.2: 96132567-96132567 on Build GRCh38
rs766071395 Chr.2: 96132613-96132613 on Build GRCh38
rs944942846 Chr.2: 96132464-96132464 on Build GRCh38
rs746895908 Chr.2: 96132600-96132600 on Build GRCh38
rs759833663 Chr.2: 96132713-96132713 on Build GRCh38
rs746228775 Chr.2: 96132523-96132523 on Build GRCh38
rs368868764 Chr.2: 96132770-96132770 on Build GRCh38
rs764030100 Chr.2: 96132506-96132506 on Build GRCh38
rs369122887 Chr.2: 96132670-96132670 on Build GRCh38
rs908111517 Chr.2: 96132837-96132837 on Build GRCh38
rs768248187 Chr.2: 96132532-96132532 on Build GRCh38
rs769572571 Chr.2: 96132680-96132680 on Build GRCh38
rs182722283 Chr.2: 96132586-96132586 on Build GRCh38
rs764556124 Chr.2: 96132560-96132560 on Build GRCh38
rs777864306 Chr.2: 96132595-96132595 on Build GRCh38
rs774149667 Chr.2: 96132615-96132615 on Build GRCh38
rs754304646 Chr.2: 96132758-96132758 on Build GRCh38
rs567686090 Chr.2: 96132811-96132811 on Build GRCh38
rs201045455 Chr.2: 96132667-96132667 on Build GRCh38
rs949740122 Chr.2: 96132694-96132694 on Build GRCh38
rs951779763 Chr.2: 96132815-96132815 on Build GRCh38
rs200103398 Chr.2: 96132665-96132665 on Build GRCh38
rs756407081 Chr.2: 96132524-96132524 on Build GRCh38
rs909361287 Chr.2: 96132853-96132853 on Build GRCh38
rs575107981 Chr.2: 96132655-96132655 on Build GRCh38
rs775876067 Chr.2: 96132575-96132575 on Build GRCh38
rs765288893 Chr.2: 96132721-96132721 on Build GRCh38
rs777378437 Chr.2: 96132669-96132669 on Build GRCh38
rs750874250 Chr.2: 96132581-96132581 on Build GRCh38
rs1014150375 Chr.2: 96132554-96132554 on Build GRCh38
rs959788586 Chr.2: 96132877-96132877 on Build GRCh38
rs908144098 Chr.2: 96132620-96132620 on Build GRCh38
rs753572086 Chr.2: 96132585-96132585 on Build GRCh38
rs757806364 Chr.2: 96132764-96132765 on Build GRCh38
rs61735195 Chr.2: 96132701-96132701 on Build GRCh38
rs957721103 Chr.2: 96132443-96132443 on Build GRCh38
rs147042054 Chr.2: 96132632-96132632 on Build GRCh38
rs758093607 Chr.2: 96132740-96132740 on Build GRCh38
rs573922224 Chr.2: 96132474-96132474 on Build GRCh38
rs922127491 Chr.2: 96132895-96132895 on Build GRCh38
rs145986421 Chr.2: 96132702-96132702 on Build GRCh38
rs761232259 Chr.2: 96132706-96132706 on Build GRCh38
rs1026356757 Chr.2: 96132808-96132808 on Build GRCh38
rs370913603 Chr.2: 96132527-96132527 on Build GRCh38
rs996165744 Chr.2: 96132559-96132559 on Build GRCh38
rs1055209461 Chr.2: 96132446-96132446 on Build GRCh38
rs910545878 Chr.2: 96132449-96132449 on Build GRCh38
rs930932621 Chr.2: 96132488-96132488 on Build GRCh38
rs774455538 Chr.2: 96132692-96132692 on Build GRCh38
rs185826096 Chr.2: 96132679-96132679 on Build GRCh38

View Full Product Details