Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96133875-96134376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
UniGene
Hs.447993
Gene Chromosome Location
Chr.2: 96122817-96143059 on Build GRCh38
Transcripts
NM_001002036.3 XM_011511205.2 XM_011511206.2 XM_011511207.2 XM_011511208.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3584063 c.337+1G>A p.? Chr.2: 96133964-96133964 on Build GRCh38
6092775 c.331A>G p.K111E Chr.2: 96133971-96133971 on Build GRCh38
3584064 c.296G>A p.S99N Chr.2: 96134006-96134006 on Build GRCh38
1742812 c.324C>A p.L108L Chr.2: 96133978-96133978 on Build GRCh38
4955884 c.303C>T p.V101V Chr.2: 96133999-96133999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536002330 Chr.2: 96134209-96134209 on Build GRCh38
rs745964188 Chr.2: 96133929-96133929 on Build GRCh38
rs142627630 Chr.2: 96134005-96134005 on Build GRCh38
rs762252298 Chr.2: 96134103-96134103 on Build GRCh38
rs780727002 Chr.2: 96133940-96133940 on Build GRCh38
rs998961152 Chr.2: 96134044-96134044 on Build GRCh38
rs746605320 Chr.2: 96134084-96134084 on Build GRCh38
rs567350273 Chr.2: 96134185-96134185 on Build GRCh38
rs769181108 Chr.2: 96133948-96133948 on Build GRCh38
rs138696994 Chr.2: 96134032-96134032 on Build GRCh38
rs1004575645 Chr.2: 96134336-96134336 on Build GRCh38
rs374993114 Chr.2: 96133930-96133930 on Build GRCh38
rs150180534 Chr.2: 96133988-96133988 on Build GRCh38
rs550355362 Chr.2: 96134098-96134098 on Build GRCh38
rs906790801 Chr.2: 96133907-96133907 on Build GRCh38
rs772942732 Chr.2: 96134074-96134074 on Build GRCh38
rs750561567 Chr.2: 96133999-96133999 on Build GRCh38
rs1052653090 Chr.2: 96134184-96134184 on Build GRCh38
rs760249581 Chr.2: 96133965-96133965 on Build GRCh38
rs752524466 Chr.2: 96133983-96133983 on Build GRCh38
rs894070878 Chr.2: 96134195-96134195 on Build GRCh38
rs766963684 Chr.2: 96134121-96134121 on Build GRCh38
rs759439566 Chr.2: 96133978-96133978 on Build GRCh38
rs758535926 Chr.2: 96134075-96134076 on Build GRCh38
rs375664604 Chr.2: 96134049-96134049 on Build GRCh38
rs973673037 Chr.2: 96134048-96134048 on Build GRCh38
rs765320695 Chr.2: 96134348-96134348 on Build GRCh38
rs753806992 Chr.2: 96133976-96133976 on Build GRCh38
rs960107172 Chr.2: 96134151-96134151 on Build GRCh38
rs927974731 Chr.2: 96134145-96134146 on Build GRCh38
rs777247621 Chr.2: 96134133-96134133 on Build GRCh38
rs1050609451 Chr.2: 96133912-96133912 on Build GRCh38
rs148110898 Chr.2: 96133966-96133966 on Build GRCh38
rs757480696 Chr.2: 96133922-96133922 on Build GRCh38
rs916660726 Chr.2: 96134123-96134124 on Build GRCh38
rs935554308 Chr.2: 96134167-96134167 on Build GRCh38
rs538516410 Chr.2: 96134288-96134288 on Build GRCh38
rs373539231 Chr.2: 96133913-96133913 on Build GRCh38
rs749857645 Chr.2: 96134007-96134007 on Build GRCh38
rs189011409 Chr.2: 96133950-96133950 on Build GRCh38
rs991554393 Chr.2: 96134180-96134180 on Build GRCh38
rs933093994 Chr.2: 96133901-96133901 on Build GRCh38
rs772271351 Chr.2: 96134066-96134066 on Build GRCh38
rs781489619 Chr.2: 96133998-96133998 on Build GRCh38
rs771746691 Chr.2: 96133954-96133954 on Build GRCh38
rs772677204 Chr.2: 96133960-96133960 on Build GRCh38
rs552534177 Chr.2: 96134225-96134225 on Build GRCh38
rs909114470 Chr.2: 96134353-96134353 on Build GRCh38
rs755524379 Chr.2: 96134023-96134023 on Build GRCh38
rs948174700 Chr.2: 96134130-96134130 on Build GRCh38
rs200408159 Chr.2: 96134087-96134087 on Build GRCh38
rs558444024 Chr.2: 96134294-96134294 on Build GRCh38
rs376266579 Chr.2: 96134138-96134138 on Build GRCh38
rs900212077 Chr.2: 96134246-96134246 on Build GRCh38
rs954380263 Chr.2: 96134300-96134300 on Build GRCh38
rs143027302 Chr.2: 96134094-96134094 on Build GRCh38
rs572533534 Chr.2: 96134229-96134229 on Build GRCh38
rs781187649 Chr.2: 96133927-96133927 on Build GRCh38
rs750866527 Chr.2: 96134061-96134063 on Build GRCh38
rs749770140 Chr.2: 96133946-96133946 on Build GRCh38
rs114425276 Chr.2: 96134345-96134345 on Build GRCh38
rs532481120 Chr.2: 96133993-96133993 on Build GRCh38
rs1051952481 Chr.2: 96133906-96133906 on Build GRCh38
rs995934780 Chr.2: 96134280-96134280 on Build GRCh38
rs759987083 Chr.2: 96134239-96134239 on Build GRCh38
rs375249891 Chr.2: 96133949-96133949 on Build GRCh38
rs765068047 Chr.2: 96133980-96133980 on Build GRCh38

View Full Product Details