Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96265993-96266484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
UniGene
Hs.12109
Gene Chromosome Location
Chr.2: 96266146-96274179 on Build GRCh38
Transcripts
NM_004804.2 NM_001193304.2 NM_017849.3 XM_017004451.1

Gene Symbol
TMEM127
Entrez Gene ID
55654
Gene Name
transmembrane protein 127
Gene Aliases
-
UniGene
Hs.164303
Gene Chromosome Location
Chr.2: 96248514-96266014 on Build GRCh38
Transcripts
NM_004804.2 NM_001193304.2 NM_017849.3 XM_017004451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407056 c.7G>T p.D3Y Chr.2: 96266357-96266357 on Build GRCh38
3695645 c.29G>A p.R10H Chr.2: 96266379-96266379 on Build GRCh38
6306804 c.56G>T p.C19F Chr.2: 96266406-96266406 on Build GRCh38
6403227 c.5A>C p.K2T Chr.2: 96266355-96266355 on Build GRCh38
6405321 c.106G>A p.G36S Chr.2: 96266456-96266456 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008656555 Chr.2: 96266464-96266464 on Build GRCh38
rs903722571 Chr.2: 96266379-96266379 on Build GRCh38
rs1038476400 Chr.2: 96266119-96266119 on Build GRCh38
rs766122023 Chr.2: 96266458-96266458 on Build GRCh38
rs753777793 Chr.2: 96266325-96266325 on Build GRCh38
rs1032968217 Chr.2: 96266336-96266336 on Build GRCh38
rs917975874 Chr.2: 96266215-96266215 on Build GRCh38
rs539465368 Chr.2: 96266123-96266123 on Build GRCh38
rs773403813 Chr.2: 96266446-96266446 on Build GRCh38
rs183324092 Chr.2: 96266311-96266311 on Build GRCh38
rs993963655 Chr.2: 96266077-96266077 on Build GRCh38
rs866408622 Chr.2: 96266097-96266097 on Build GRCh38
rs929839012 Chr.2: 96266393-96266393 on Build GRCh38
rs569518818 Chr.2: 96266238-96266238 on Build GRCh38
rs926590957 Chr.2: 96266233-96266233 on Build GRCh38
rs1017926087 Chr.2: 96266281-96266281 on Build GRCh38
rs961578084 Chr.2: 96266269-96266269 on Build GRCh38
rs964287509 Chr.2: 96266169-96266169 on Build GRCh38
rs549269750 Chr.2: 96266427-96266427 on Build GRCh38
rs774429632 Chr.2: 96266428-96266428 on Build GRCh38
rs781708415 Chr.2: 96266049-96266049 on Build GRCh38
rs1039425806 Chr.2: 96266435-96266435 on Build GRCh38
rs1025083472 Chr.2: 96266343-96266343 on Build GRCh38
rs974216366 Chr.2: 96266270-96266270 on Build GRCh38
rs772201213 Chr.2: 96266433-96266433 on Build GRCh38
rs570448081 Chr.2: 96266118-96266118 on Build GRCh38
rs1018493552 Chr.2: 96266137-96266137 on Build GRCh38
rs1003199437 Chr.2: 96266208-96266208 on Build GRCh38
rs921223616 Chr.2: 96266066-96266066 on Build GRCh38
rs1009417664 Chr.2: 96266220-96266220 on Build GRCh38
rs929760358 Chr.2: 96266249-96266249 on Build GRCh38
rs759160802 Chr.2: 96266205-96266205 on Build GRCh38
rs541537065 Chr.2: 96266278-96266278 on Build GRCh38
rs764756576 Chr.2: 96266338-96266338 on Build GRCh38
rs192322868 Chr.2: 96266206-96266206 on Build GRCh38
rs543450676 Chr.2: 96266359-96266359 on Build GRCh38
rs926374916 Chr.2: 96266332-96266332 on Build GRCh38
rs1032546313 Chr.2: 96266188-96266188 on Build GRCh38
rs905751787 Chr.2: 96266041-96266041 on Build GRCh38
rs555550230 Chr.2: 96266246-96266246 on Build GRCh38
rs968096757 Chr.2: 96266040-96266040 on Build GRCh38
rs1038683286 Chr.2: 96266240-96266240 on Build GRCh38
rs376741594 Chr.2: 96266432-96266432 on Build GRCh38
rs199793823 Chr.2: 96266406-96266406 on Build GRCh38
rs758056557 Chr.2: 96266344-96266344 on Build GRCh38
rs1046806368 Chr.2: 96266261-96266261 on Build GRCh38
rs760931648 Chr.2: 96266448-96266448 on Build GRCh38
rs980080554 Chr.2: 96266187-96266187 on Build GRCh38
rs919804072 Chr.2: 96266386-96266386 on Build GRCh38
rs765606392 Chr.2: 96266214-96266214 on Build GRCh38
rs2301707 Chr.2: 96266108-96266108 on Build GRCh38
rs754904404 Chr.2: 96266328-96266328 on Build GRCh38
rs576453050 Chr.2: 96266222-96266222 on Build GRCh38
rs1028399196 Chr.2: 96266242-96266242 on Build GRCh38
rs764724164 Chr.2: 96266418-96266418 on Build GRCh38
rs746716248 Chr.2: 96266360-96266360 on Build GRCh38
rs955585056 Chr.2: 96266243-96266243 on Build GRCh38
rs1045248299 Chr.2: 96266312-96266312 on Build GRCh38
rs572415505 Chr.2: 96266250-96266250 on Build GRCh38
rs890794289 Chr.2: 96266115-96266115 on Build GRCh38
rs939033128 Chr.2: 96266335-96266335 on Build GRCh38
rs1007815122 Chr.2: 96266116-96266116 on Build GRCh38
rs980746701 Chr.2: 96266229-96266229 on Build GRCh38
rs890112199 Chr.2: 96266226-96266250 on Build GRCh38
rs898307338 Chr.2: 96266076-96266076 on Build GRCh38
rs374164270 Chr.2: 96266383-96266383 on Build GRCh38
rs1044571 Chr.2: 96266289-96266289 on Build GRCh38
rs1001164235 Chr.2: 96266327-96266327 on Build GRCh38
rs185791368 Chr.2: 96266342-96266342 on Build GRCh38
rs890846936 Chr.2: 96266265-96266265 on Build GRCh38
rs980837410 Chr.2: 96266346-96266346 on Build GRCh38
rs974017610 Chr.2: 96266385-96266385 on Build GRCh38
rs765842818 Chr.2: 96266047-96266047 on Build GRCh38
rs1046856049 Chr.2: 96266398-96266398 on Build GRCh38
rs1044819245 Chr.2: 96266189-96266189 on Build GRCh38

View Full Product Details