Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96268379-96268885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
UniGene
Hs.12109
Gene Chromosome Location
Chr.2: 96266146-96274179 on Build GRCh38
Transcripts
NM_004804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987908 c.520G>A p.V174M Chr.2: 96268487-96268487 on Build GRCh38
6629158 c.601G>A p.A201T Chr.2: 96268568-96268568 on Build GRCh38
5964117 c.612G>A p.P204P Chr.2: 96268579-96268579 on Build GRCh38
3840279 c.511G>C p.D171H Chr.2: 96268478-96268478 on Build GRCh38
5005931 c.533G>A p.R178Q Chr.2: 96268500-96268500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559101787 Chr.2: 96268552-96268552 on Build GRCh38
rs746325594 Chr.2: 96268645-96268645 on Build GRCh38
rs369168627 Chr.2: 96268702-96268702 on Build GRCh38
rs946049496 Chr.2: 96268819-96268819 on Build GRCh38
rs768342930 Chr.2: 96268668-96268668 on Build GRCh38
rs746419754 Chr.2: 96268529-96268529 on Build GRCh38
rs757115923 Chr.2: 96268426-96268426 on Build GRCh38
rs757668297 Chr.2: 96268637-96268637 on Build GRCh38
rs993774739 Chr.2: 96268711-96268711 on Build GRCh38
rs375865195 Chr.2: 96268686-96268686 on Build GRCh38
rs749741492 Chr.2: 96268490-96268491 on Build GRCh38
rs771485213 Chr.2: 96268520-96268520 on Build GRCh38
rs993355025 Chr.2: 96268768-96268768 on Build GRCh38
rs952599630 Chr.2: 96268739-96268739 on Build GRCh38
rs747374312 Chr.2: 96268504-96268504 on Build GRCh38
rs977391420 Chr.2: 96268821-96268823 on Build GRCh38
rs377203527 Chr.2: 96268500-96268500 on Build GRCh38
rs936213775 Chr.2: 96268843-96268843 on Build GRCh38
rs774622752 Chr.2: 96268589-96268589 on Build GRCh38
rs371292183 Chr.2: 96268632-96268632 on Build GRCh38
rs761680697 Chr.2: 96268831-96268831 on Build GRCh38
rs140260270 Chr.2: 96268650-96268650 on Build GRCh38
rs367966046 Chr.2: 96268427-96268427 on Build GRCh38
rs144987111 Chr.2: 96268579-96268579 on Build GRCh38
rs983976562 Chr.2: 96268748-96268748 on Build GRCh38
rs1004474331 Chr.2: 96268631-96268631 on Build GRCh38
rs542568006 Chr.2: 96268452-96268452 on Build GRCh38
rs749319878 Chr.2: 96268667-96268667 on Build GRCh38
rs966288607 Chr.2: 96268675-96268675 on Build GRCh38
rs756030018 Chr.2: 96268603-96268603 on Build GRCh38
rs754584475 Chr.2: 96268613-96268613 on Build GRCh38
rs771628407 Chr.2: 96268659-96268660 on Build GRCh38
rs936196574 Chr.2: 96268609-96268609 on Build GRCh38
rs1025533131 Chr.2: 96268734-96268734 on Build GRCh38
rs774534803 Chr.2: 96268691-96268691 on Build GRCh38
rs748924142 Chr.2: 96268480-96268480 on Build GRCh38
rs753292815 Chr.2: 96268607-96268607 on Build GRCh38
rs769876006 Chr.2: 96268530-96268530 on Build GRCh38
rs571238881 Chr.2: 96268604-96268604 on Build GRCh38
rs953668969 Chr.2: 96268760-96268760 on Build GRCh38
rs779467939 Chr.2: 96268475-96268475 on Build GRCh38
rs1029195965 Chr.2: 96268724-96268724 on Build GRCh38
rs992845067 Chr.2: 96268524-96268524 on Build GRCh38
rs904753769 Chr.2: 96268583-96268583 on Build GRCh38
rs773006368 Chr.2: 96268697-96268697 on Build GRCh38
rs62153033 Chr.2: 96268766-96268766 on Build GRCh38
rs1050698715 Chr.2: 96268847-96268847 on Build GRCh38
rs192314683 Chr.2: 96268597-96268597 on Build GRCh38
rs767437368 Chr.2: 96268429-96268429 on Build GRCh38
rs939231845 Chr.2: 96268548-96268548 on Build GRCh38
rs902409423 Chr.2: 96268544-96268544 on Build GRCh38
rs927411593 Chr.2: 96268420-96268420 on Build GRCh38
rs142522111 Chr.2: 96268614-96268614 on Build GRCh38
rs946021559 Chr.2: 96268537-96268537 on Build GRCh38
rs761581891 Chr.2: 96268590-96268590 on Build GRCh38
rs563370317 Chr.2: 96268405-96268405 on Build GRCh38
rs775568417 Chr.2: 96268664-96268664 on Build GRCh38
rs199846474 Chr.2: 96268578-96268578 on Build GRCh38
rs199831944 Chr.2: 96268565-96268565 on Build GRCh38
rs1042341646 Chr.2: 96268531-96268531 on Build GRCh38
rs772000473 Chr.2: 96268445-96268445 on Build GRCh38
rs368621207 Chr.2: 96268527-96268527 on Build GRCh38
rs1027780080 Chr.2: 96268612-96268612 on Build GRCh38
rs147727121 Chr.2: 96268417-96268417 on Build GRCh38
rs760596766 Chr.2: 96268699-96268699 on Build GRCh38
rs184147889 Chr.2: 96268639-96268639 on Build GRCh38
rs767347306 Chr.2: 96268596-96268596 on Build GRCh38
rs977443952 Chr.2: 96268826-96268826 on Build GRCh38
rs754601899 Chr.2: 96268498-96268498 on Build GRCh38
rs1019316162 Chr.2: 96268652-96268652 on Build GRCh38
rs553306687 Chr.2: 96268814-96268814 on Build GRCh38
rs767259141 Chr.2: 96268695-96268695 on Build GRCh38
rs747856302 Chr.2: 96268622-96268622 on Build GRCh38
rs914612786 Chr.2: 96268781-96268781 on Build GRCh38
rs746370859 Chr.2: 96268703-96268703 on Build GRCh38

View Full Product Details