Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96550383-96550876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID5A
Entrez Gene ID
10865
Gene Name
AT-rich interaction domain 5A
Gene Aliases
MRF-1, MRF1, RP11-363D14
UniGene
Hs.920
Gene Chromosome Location
Chr.2: 96536719-96552638 on Build GRCh38
Transcripts
NM_001319085.1 NM_001319087.1 NM_001319092.1 NM_001319093.1 NM_001319094.1 NM_001319096.1 NM_212481.2 XM_011510499.2 XM_017003183.1 XM_017003184.1 XM_017003185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1738599 c.497C>G p.A166G Chr.2: 96550660-96550660 on Build GRCh38
7286265 c.509delG p.D172fs*20 Chr.2: 96550672-96550672 on Build GRCh38
7356418 c.529G>T p.E177* Chr.2: 96550692-96550692 on Build GRCh38
5684145 c.430C>T p.R144W Chr.2: 96550593-96550593 on Build GRCh38
1130712 c.544G>A p.A182T Chr.2: 96550707-96550707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948147179 Chr.2: 96550536-96550536 on Build GRCh38
rs927804135 Chr.2: 96550498-96550498 on Build GRCh38
rs1037768745 Chr.2: 96550473-96550473 on Build GRCh38
rs747867101 Chr.2: 96550558-96550558 on Build GRCh38
rs377372638 Chr.2: 96550743-96550743 on Build GRCh38
rs36003446 Chr.2: 96550631-96550631 on Build GRCh38
rs958334522 Chr.2: 96550463-96550463 on Build GRCh38
rs1015610553 Chr.2: 96550809-96550809 on Build GRCh38
rs1004286060 Chr.2: 96550409-96550409 on Build GRCh38
rs12472484 Chr.2: 96550525-96550525 on Build GRCh38
rs745688261 Chr.2: 96550491-96550491 on Build GRCh38
rs56060938 Chr.2: 96550692-96550692 on Build GRCh38
rs768268780 Chr.2: 96550526-96550526 on Build GRCh38
rs150763874 Chr.2: 96550662-96550662 on Build GRCh38
rs377598098 Chr.2: 96550559-96550559 on Build GRCh38
rs887148104 Chr.2: 96550586-96550586 on Build GRCh38
rs1046221390 Chr.2: 96550488-96550488 on Build GRCh38
rs939994257 Chr.2: 96550850-96550850 on Build GRCh38
rs368409361 Chr.2: 96550622-96550622 on Build GRCh38
rs551015377 Chr.2: 96550805-96550805 on Build GRCh38
rs776037854 Chr.2: 96550595-96550595 on Build GRCh38
rs1041069339 Chr.2: 96550851-96550851 on Build GRCh38
rs1047660532 Chr.2: 96550765-96550765 on Build GRCh38
rs930557898 Chr.2: 96550748-96550748 on Build GRCh38
rs201900040 Chr.2: 96550555-96550555 on Build GRCh38
rs370634774 Chr.2: 96550571-96550571 on Build GRCh38
rs937872926 Chr.2: 96550506-96550506 on Build GRCh38
rs746222389 Chr.2: 96550704-96550704 on Build GRCh38
rs113371580 Chr.2: 96550430-96550430 on Build GRCh38
rs768085834 Chr.2: 96550671-96550671 on Build GRCh38
rs777474043 Chr.2: 96550700-96550700 on Build GRCh38
rs989764612 Chr.2: 96550689-96550689 on Build GRCh38
rs913578836 Chr.2: 96550478-96550478 on Build GRCh38
rs963382779 Chr.2: 96550824-96550824 on Build GRCh38
rs567940439 Chr.2: 96550808-96550808 on Build GRCh38
rs1044233098 Chr.2: 96550565-96550565 on Build GRCh38
rs775911272 Chr.2: 96550707-96550707 on Build GRCh38
rs749830072 Chr.2: 96550715-96550715 on Build GRCh38
rs778370882 Chr.2: 96550534-96550534 on Build GRCh38
rs780034963 Chr.2: 96550720-96550720 on Build GRCh38
rs143379982 Chr.2: 96550429-96550429 on Build GRCh38
rs759938225 Chr.2: 96550589-96550589 on Build GRCh38
rs770284184 Chr.2: 96550593-96550593 on Build GRCh38
rs770161130 Chr.2: 96550705-96550705 on Build GRCh38
rs768567479 Chr.2: 96550716-96550716 on Build GRCh38
rs766405325 Chr.2: 96550668-96550668 on Build GRCh38
rs770481563 Chr.2: 96550781-96550781 on Build GRCh38
rs762886555 Chr.2: 96550616-96550616 on Build GRCh38
rs983682197 Chr.2: 96550800-96550800 on Build GRCh38
rs945100094 Chr.2: 96550481-96550481 on Build GRCh38
rs771152996 Chr.2: 96550566-96550566 on Build GRCh38
rs754407768 Chr.2: 96550676-96550676 on Build GRCh38
rs989107614 Chr.2: 96550469-96550469 on Build GRCh38
rs761510345 Chr.2: 96550723-96550723 on Build GRCh38
rs748682799 Chr.2: 96550671-96550672 on Build GRCh38
rs1004235214 Chr.2: 96550604-96550604 on Build GRCh38
rs145969024 Chr.2: 96550696-96550696 on Build GRCh38
rs750211527 Chr.2: 96550637-96550637 on Build GRCh38
rs927676001 Chr.2: 96550702-96550702 on Build GRCh38
rs921485341 Chr.2: 96550461-96550461 on Build GRCh38
rs200249887 Chr.2: 96550453-96550454 on Build GRCh38
rs951431061 Chr.2: 96550404-96550404 on Build GRCh38
rs762113269 Chr.2: 96550628-96550628 on Build GRCh38
rs760208081 Chr.2: 96550632-96550632 on Build GRCh38
rs997400713 Chr.2: 96550644-96550644 on Build GRCh38
rs748802848 Chr.2: 96550524-96550524 on Build GRCh38
rs887097179 Chr.2: 96550837-96550837 on Build GRCh38
rs761835947 Chr.2: 96550728-96550728 on Build GRCh38
rs1019738719 Chr.2: 96550420-96550420 on Build GRCh38
rs973799370 Chr.2: 96550853-96550853 on Build GRCh38
rs551537636 Chr.2: 96550677-96550677 on Build GRCh38

View Full Product Details