Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96647687-96648195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542844 c.34G>T p.A12S Chr.2: 96647791-96647791 on Build GRCh38
6542843 c.244G>T p.A82S Chr.2: 96647791-96647791 on Build GRCh38
6509577 c.31C>A p.L11M Chr.2: 96647788-96647788 on Build GRCh38
1661836 c.125C>T p.T42I Chr.2: 96647882-96647882 on Build GRCh38
6509576 c.241C>A p.L81M Chr.2: 96647788-96647788 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954228627 Chr.2: 96647725-96647725 on Build GRCh38
rs930190353 Chr.2: 96647936-96647936 on Build GRCh38
rs912775241 Chr.2: 96647742-96647742 on Build GRCh38
rs147475359 Chr.2: 96647850-96647850 on Build GRCh38
rs767752398 Chr.2: 96647765-96647765 on Build GRCh38
rs140096817 Chr.2: 96648080-96648080 on Build GRCh38
rs912180534 Chr.2: 96647949-96647949 on Build GRCh38
rs899576230 Chr.2: 96647972-96647972 on Build GRCh38
rs779884686 Chr.2: 96647805-96647805 on Build GRCh38
rs186703140 Chr.2: 96648071-96648071 on Build GRCh38
rs540309562 Chr.2: 96647939-96647939 on Build GRCh38
rs757432678 Chr.2: 96647764-96647764 on Build GRCh38
rs966420671 Chr.2: 96647718-96647718 on Build GRCh38
rs920217568 Chr.2: 96647933-96647933 on Build GRCh38
rs979203750 Chr.2: 96647720-96647720 on Build GRCh38
rs1032509990 Chr.2: 96647722-96647722 on Build GRCh38
rs370201394 Chr.2: 96648048-96648048 on Build GRCh38
rs907233802 Chr.2: 96648170-96648170 on Build GRCh38
rs747610167 Chr.2: 96647869-96647869 on Build GRCh38
rs549939569 Chr.2: 96648100-96648100 on Build GRCh38
rs200413243 Chr.2: 96647828-96647828 on Build GRCh38
rs781272286 Chr.2: 96647948-96647948 on Build GRCh38
rs892366017 Chr.2: 96647768-96647768 on Build GRCh38
rs937809642 Chr.2: 96647942-96647942 on Build GRCh38
rs771551036 Chr.2: 96647878-96647878 on Build GRCh38
rs562084378 Chr.2: 96648019-96648019 on Build GRCh38
rs943699482 Chr.2: 96647959-96647959 on Build GRCh38
rs944308479 Chr.2: 96647774-96647774 on Build GRCh38
rs534047381 Chr.2: 96647711-96647711 on Build GRCh38
rs1020217171 Chr.2: 96648099-96648099 on Build GRCh38
rs958899326 Chr.2: 96647908-96647908 on Build GRCh38
rs527977956 Chr.2: 96648034-96648034 on Build GRCh38
rs902203744 Chr.2: 96648039-96648039 on Build GRCh38
rs896379586 Chr.2: 96647955-96647955 on Build GRCh38
rs780271307 Chr.2: 96648109-96648109 on Build GRCh38
rs563000521 Chr.2: 96647757-96647757 on Build GRCh38
rs1039467898 Chr.2: 96647960-96647960 on Build GRCh38
rs267599498 Chr.2: 96647879-96647879 on Build GRCh38
rs748320252 Chr.2: 96648079-96648079 on Build GRCh38
rs1042042270 Chr.2: 96647986-96647986 on Build GRCh38
rs60378028 Chr.2: 96647714-96647714 on Build GRCh38
rs750625807 Chr.2: 96647772-96647772 on Build GRCh38
rs1009857179 Chr.2: 96648086-96648086 on Build GRCh38
rs778853167 Chr.2: 96647841-96647841 on Build GRCh38
rs756355909 Chr.2: 96647790-96647790 on Build GRCh38
rs971506895 Chr.2: 96647832-96647832 on Build GRCh38
rs770015092 Chr.2: 96648134-96648134 on Build GRCh38
rs755083165 Chr.2: 96647914-96647914 on Build GRCh38
rs542258168 Chr.2: 96647915-96647915 on Build GRCh38
rs749950 Chr.2: 96647737-96647737 on Build GRCh38
rs752289110 Chr.2: 96647731-96647732 on Build GRCh38
rs749949 Chr.2: 96647713-96647713 on Build GRCh38
rs1010253244 Chr.2: 96647811-96647811 on Build GRCh38
rs368496444 Chr.2: 96647978-96647978 on Build GRCh38
rs1013933578 Chr.2: 96647976-96647976 on Build GRCh38
rs769536108 Chr.2: 96648094-96648094 on Build GRCh38

View Full Product Details